Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39743932-39744465 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTHD1
Entrez Gene ID
150350
Gene Name
ENTH domain containing 1
Gene Aliases
CACNA1I, dJ370M22.3
UniGene
Hs.474869
Gene Chromosome Location
Chr.22: 39743044-39893884 on Build GRCh38
Transcripts
NM_152512.3 XM_006724149.3 XM_006724150.3 XM_006724151.1 XM_011529927.2 XM_011529928.2 XM_011529930.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7237126 c.1464C>G p.L488L Chr.22: 39744039-39744039 on Build GRCh38
1714409 c.1313C>T p.S438L Chr.22: 39744190-39744190 on Build GRCh38
3554753 c.1223C>T p.S408L Chr.22: 39744280-39744280 on Build GRCh38
7236789 c.1421C>T p.S474F Chr.22: 39744082-39744082 on Build GRCh38
6276454 c.1315C>A p.P439T Chr.22: 39744188-39744188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762736817 Chr.22: 39743990-39743990 on Build GRCh38
rs868164759 Chr.22: 39744281-39744281 on Build GRCh38
rs752651560 Chr.22: 39744241-39744241 on Build GRCh38
rs191007110 Chr.22: 39744294-39744294 on Build GRCh38
rs771522264 Chr.22: 39744047-39744047 on Build GRCh38
rs1032110332 Chr.22: 39744077-39744077 on Build GRCh38
rs529502860 Chr.22: 39744188-39744188 on Build GRCh38
rs761355836 Chr.22: 39744085-39744085 on Build GRCh38
rs532011940 Chr.22: 39744172-39744172 on Build GRCh38
rs758936453 Chr.22: 39744324-39744324 on Build GRCh38
rs780779007 Chr.22: 39744328-39744328 on Build GRCh38
rs574689277 Chr.22: 39744423-39744423 on Build GRCh38
rs200008989 Chr.22: 39744298-39744298 on Build GRCh38
rs376116655 Chr.22: 39744053-39744053 on Build GRCh38
rs868771517 Chr.22: 39744178-39744178 on Build GRCh38
rs768337439 Chr.22: 39744177-39744177 on Build GRCh38
rs114978051 Chr.22: 39744132-39744132 on Build GRCh38
rs751508929 Chr.22: 39743959-39743960 on Build GRCh38
rs746563037 Chr.22: 39744056-39744056 on Build GRCh38
rs747664310 Chr.22: 39744332-39744332 on Build GRCh38
rs139301765 Chr.22: 39744037-39744037 on Build GRCh38
rs769284855 Chr.22: 39744214-39744214 on Build GRCh38
rs918254917 Chr.22: 39744220-39744220 on Build GRCh38
rs767590468 Chr.22: 39744285-39744285 on Build GRCh38
rs780907956 Chr.22: 39744076-39744076 on Build GRCh38
rs546690722 Chr.22: 39743984-39743984 on Build GRCh38
rs751467701 Chr.22: 39744131-39744131 on Build GRCh38
rs116668695 Chr.22: 39744176-39744176 on Build GRCh38
rs745523909 Chr.22: 39744126-39744126 on Build GRCh38
rs768245834 Chr.22: 39744061-39744061 on Build GRCh38
rs759413013 Chr.22: 39744259-39744259 on Build GRCh38
rs904635503 Chr.22: 39744033-39744033 on Build GRCh38
rs904668138 Chr.22: 39744366-39744366 on Build GRCh38
rs547486923 Chr.22: 39744193-39744193 on Build GRCh38
rs373242296 Chr.22: 39744032-39744032 on Build GRCh38
rs200214610 Chr.22: 39744041-39744041 on Build GRCh38
rs201138603 Chr.22: 39743971-39743971 on Build GRCh38
rs757758541 Chr.22: 39744307-39744307 on Build GRCh38
rs762974291 Chr.22: 39744117-39744117 on Build GRCh38
rs1031933339 Chr.22: 39744010-39744010 on Build GRCh38
rs115302874 Chr.22: 39744238-39744238 on Build GRCh38
rs115673469 Chr.22: 39744223-39744223 on Build GRCh38
rs149999735 Chr.22: 39744118-39744118 on Build GRCh38
rs764933165 Chr.22: 39743983-39743983 on Build GRCh38
rs143498985 Chr.22: 39744161-39744161 on Build GRCh38
rs765680600 Chr.22: 39744319-39744319 on Build GRCh38
rs114600441 Chr.22: 39744186-39744186 on Build GRCh38
rs751594535 Chr.22: 39744017-39744017 on Build GRCh38
rs1000703327 Chr.22: 39744395-39744395 on Build GRCh38
rs556145635 Chr.22: 39744399-39744399 on Build GRCh38
rs374392116 Chr.22: 39744244-39744244 on Build GRCh38
rs992713516 Chr.22: 39744099-39744099 on Build GRCh38
rs776491903 Chr.22: 39744076-39744076 on Build GRCh38
rs772844860 Chr.22: 39744115-39744115 on Build GRCh38
rs538138761 Chr.22: 39744386-39744386 on Build GRCh38
rs767573970 Chr.22: 39744133-39744133 on Build GRCh38
rs757672725 Chr.22: 39744162-39744162 on Build GRCh38
rs112945971 Chr.22: 39744158-39744158 on Build GRCh38
rs757056332 Chr.22: 39744073-39744073 on Build GRCh38
rs78556357 Chr.22: 39744439-39744439 on Build GRCh38
rs752857604 Chr.22: 39744031-39744031 on Build GRCh38
rs747792238 Chr.22: 39744192-39744193 on Build GRCh38
rs761705077 Chr.22: 39743982-39743982 on Build GRCh38
rs760433786 Chr.22: 39743968-39743968 on Build GRCh38
rs759366808 Chr.22: 39744102-39744102 on Build GRCh38
rs999477775 Chr.22: 39743998-39743998 on Build GRCh38
rs980493689 Chr.22: 39744060-39744060 on Build GRCh38
rs751463026 Chr.22: 39744098-39744098 on Build GRCh38
rs375328695 Chr.22: 39743976-39743976 on Build GRCh38
rs1007771858 Chr.22: 39744027-39744027 on Build GRCh38
rs568312009 Chr.22: 39744064-39744064 on Build GRCh38
rs561938047 Chr.22: 39744185-39744185 on Build GRCh38
rs762780840 Chr.22: 39744239-39744239 on Build GRCh38
rs775390948 Chr.22: 39744292-39744292 on Build GRCh38
rs369391771 Chr.22: 39744022-39744022 on Build GRCh38
rs755622430 Chr.22: 39744334-39744334 on Build GRCh38
rs756222786 Chr.22: 39744153-39744153 on Build GRCh38
rs775073970 Chr.22: 39743963-39743963 on Build GRCh38
rs771713637 Chr.22: 39744356-39744356 on Build GRCh38
rs766276952 Chr.22: 39744091-39744091 on Build GRCh38
rs766480780 Chr.22: 39744011-39744011 on Build GRCh38
rs755031768 Chr.22: 39744020-39744020 on Build GRCh38
rs769388346 Chr.22: 39744129-39744129 on Build GRCh38
rs891802024 Chr.22: 39744433-39744433 on Build GRCh38
rs1039634295 Chr.22: 39744330-39744330 on Build GRCh38
rs370409541 Chr.22: 39744145-39744145 on Build GRCh38
rs752978671 Chr.22: 39744135-39744135 on Build GRCh38
rs114891067 Chr.22: 39743979-39743979 on Build GRCh38
rs960585734 Chr.22: 39744127-39744127 on Build GRCh38
rs750977073 Chr.22: 39744323-39744323 on Build GRCh38
rs943589573 Chr.22: 39744299-39744299 on Build GRCh38

View Full Product Details