Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39995047-39995537 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83F
Entrez Gene ID
113828
Gene Name
family with sequence similarity 83 member F
Gene Aliases
-
UniGene
Hs.197680
Gene Chromosome Location
Chr.22: 39994949-40030039 on Build GRCh38
Transcripts
NM_138435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6214298 c.134C>G p.A45G Chr.22: 39995176-39995176 on Build GRCh38
6208454 c.441G>A p.A147A Chr.22: 39995483-39995483 on Build GRCh38
5368248 c.124G>A p.G42S Chr.22: 39995166-39995166 on Build GRCh38
6539655 c.202C>G p.R68G Chr.22: 39995244-39995244 on Build GRCh38
6840259 c.286C>G p.P96A Chr.22: 39995328-39995328 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182864330 Chr.22: 39995327-39995327 on Build GRCh38
rs780242455 Chr.22: 39995337-39995337 on Build GRCh38
rs761716229 Chr.22: 39995466-39995466 on Build GRCh38
rs776338603 Chr.22: 39995458-39995458 on Build GRCh38
rs770351704 Chr.22: 39995246-39995246 on Build GRCh38
rs982213513 Chr.22: 39995126-39995126 on Build GRCh38
rs144508688 Chr.22: 39995228-39995228 on Build GRCh38
rs557221818 Chr.22: 39995181-39995181 on Build GRCh38
rs918263276 Chr.22: 39995348-39995348 on Build GRCh38
rs756411323 Chr.22: 39995410-39995410 on Build GRCh38
rs750330123 Chr.22: 39995333-39995333 on Build GRCh38
rs758559986 Chr.22: 39995335-39995335 on Build GRCh38
rs909723107 Chr.22: 39995189-39995189 on Build GRCh38
rs1000804355 Chr.22: 39995283-39995283 on Build GRCh38
rs751830504 Chr.22: 39995360-39995360 on Build GRCh38
rs780438842 Chr.22: 39995113-39995113 on Build GRCh38
rs970905927 Chr.22: 39995406-39995406 on Build GRCh38
rs1031449599 Chr.22: 39995288-39995288 on Build GRCh38
rs763776467 Chr.22: 39995318-39995318 on Build GRCh38
rs771590239 Chr.22: 39995429-39995429 on Build GRCh38
rs571485923 Chr.22: 39995112-39995112 on Build GRCh38
rs376460601 Chr.22: 39995441-39995441 on Build GRCh38
rs746493818 Chr.22: 39995445-39995445 on Build GRCh38
rs753612158 Chr.22: 39995322-39995322 on Build GRCh38
rs942536384 Chr.22: 39995415-39995415 on Build GRCh38
rs1013621621 Chr.22: 39995490-39995490 on Build GRCh38
rs760193367 Chr.22: 39995310-39995310 on Build GRCh38
rs752114815 Chr.22: 39995302-39995303 on Build GRCh38
rs942589325 Chr.22: 39995233-39995233 on Build GRCh38
rs554283947 Chr.22: 39995244-39995244 on Build GRCh38
rs903820686 Chr.22: 39995282-39995282 on Build GRCh38
rs903412810 Chr.22: 39995303-39995308 on Build GRCh38
rs986965846 Chr.22: 39995372-39995372 on Build GRCh38
rs759059352 Chr.22: 39995279-39995279 on Build GRCh38
rs371318214 Chr.22: 39995081-39995081 on Build GRCh38
rs765061473 Chr.22: 39995499-39995499 on Build GRCh38
rs773314476 Chr.22: 39995507-39995507 on Build GRCh38
rs538891826 Chr.22: 39995166-39995166 on Build GRCh38
rs1033107729 Chr.22: 39995318-39995319 on Build GRCh38
rs548156672 Chr.22: 39995241-39995241 on Build GRCh38
rs951293296 Chr.22: 39995443-39995443 on Build GRCh38
rs781431457 Chr.22: 39995324-39995325 on Build GRCh38
rs867751149 Chr.22: 39995484-39995484 on Build GRCh38
rs757799046 Chr.22: 39995316-39995317 on Build GRCh38
rs973732994 Chr.22: 39995345-39995345 on Build GRCh38
rs149259541 Chr.22: 39995096-39995096 on Build GRCh38
rs575537665 Chr.22: 39995187-39995187 on Build GRCh38
rs1052424041 Chr.22: 39995462-39995462 on Build GRCh38
rs984444386 Chr.22: 39995459-39995459 on Build GRCh38
rs768263209 Chr.22: 39995451-39995451 on Build GRCh38
rs1025107041 Chr.22: 39995319-39995319 on Build GRCh38
rs771502116 Chr.22: 39995290-39995290 on Build GRCh38
rs903768311 Chr.22: 39995450-39995450 on Build GRCh38
rs539932773 Chr.22: 39995328-39995328 on Build GRCh38
rs929726175 Chr.22: 39995171-39995171 on Build GRCh38
rs1050922952 Chr.22: 39995211-39995211 on Build GRCh38
rs534821105 Chr.22: 39995087-39995087 on Build GRCh38
rs747134916 Chr.22: 39995133-39995133 on Build GRCh38
rs749752916 Chr.22: 39995425-39995425 on Build GRCh38
rs748518685 Chr.22: 39995374-39995374 on Build GRCh38
rs936605840 Chr.22: 39995452-39995452 on Build GRCh38
rs778256073 Chr.22: 39995413-39995413 on Build GRCh38
rs909497332 Chr.22: 39995120-39995120 on Build GRCh38
rs1013072222 Chr.22: 39995385-39995385 on Build GRCh38
rs755205792 Chr.22: 39995361-39995361 on Build GRCh38
rs918327880 Chr.22: 39995169-39995169 on Build GRCh38
rs750601731 Chr.22: 39995099-39995099 on Build GRCh38
rs775236815 Chr.22: 39995306-39995306 on Build GRCh38
rs372029034 Chr.22: 39995366-39995366 on Build GRCh38
rs890958462 Chr.22: 39995224-39995224 on Build GRCh38
rs918491307 Chr.22: 39995141-39995141 on Build GRCh38
rs941002016 Chr.22: 39995164-39995164 on Build GRCh38
rs758541793 Chr.22: 39995111-39995111 on Build GRCh38
rs781363933 Chr.22: 39995216-39995216 on Build GRCh38
rs998186104 Chr.22: 39995297-39995297 on Build GRCh38
rs892523775 Chr.22: 39995296-39995296 on Build GRCh38
rs773695733 Chr.22: 39995247-39995247 on Build GRCh38

View Full Product Details