Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 39995438-39995925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM83F
Entrez Gene ID
113828
Gene Name
family with sequence similarity 83 member F
Gene Aliases
-
UniGene
Hs.197680
Gene Chromosome Location
Chr.22: 39994949-40030039 on Build GRCh38
Transcripts
NM_138435.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6208454 c.441G>A p.A147A Chr.22: 39995483-39995483 on Build GRCh38
4548335 c.444G>A p.P148P Chr.22: 39995486-39995486 on Build GRCh38
6298462 c.396G>T p.V132V Chr.22: 39995438-39995438 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs571446715 Chr.22: 39995839-39995839 on Build GRCh38
rs761716229 Chr.22: 39995466-39995466 on Build GRCh38
rs776338603 Chr.22: 39995458-39995458 on Build GRCh38
rs905300423 Chr.22: 39995524-39995524 on Build GRCh38
rs546684674 Chr.22: 39995833-39995833 on Build GRCh38
rs1046460790 Chr.22: 39995745-39995745 on Build GRCh38
rs148762006 Chr.22: 39995624-39995624 on Build GRCh38
rs951113749 Chr.22: 39995569-39995569 on Build GRCh38
rs934088726 Chr.22: 39995560-39995560 on Build GRCh38
rs922664363 Chr.22: 39995559-39995559 on Build GRCh38
rs955009329 Chr.22: 39995874-39995874 on Build GRCh38
rs986523654 Chr.22: 39995588-39995588 on Build GRCh38
rs1054627529 Chr.22: 39995614-39995614 on Build GRCh38
rs1013621621 Chr.22: 39995490-39995490 on Build GRCh38
rs1003710249 Chr.22: 39995661-39995661 on Build GRCh38
rs903190786 Chr.22: 39995579-39995579 on Build GRCh38
rs141513158 Chr.22: 39995640-39995640 on Build GRCh38
rs765061473 Chr.22: 39995499-39995499 on Build GRCh38
rs1024444627 Chr.22: 39995636-39995636 on Build GRCh38
rs922442254 Chr.22: 39995803-39995803 on Build GRCh38
rs1016850832 Chr.22: 39995726-39995726 on Build GRCh38
rs773314476 Chr.22: 39995507-39995507 on Build GRCh38
rs912296897 Chr.22: 39995609-39995609 on Build GRCh38
rs562724280 Chr.22: 39995617-39995617 on Build GRCh38
rs1026981198 Chr.22: 39995870-39995870 on Build GRCh38
rs56293563 Chr.22: 39995591-39995591 on Build GRCh38
rs1002358458 Chr.22: 39995525-39995525 on Build GRCh38
rs1039588782 Chr.22: 39995557-39995557 on Build GRCh38
rs1002304619 Chr.22: 39995755-39995755 on Build GRCh38
rs983824969 Chr.22: 39995710-39995710 on Build GRCh38
rs1025277122 Chr.22: 39995567-39995567 on Build GRCh38
rs942482593 Chr.22: 39995620-39995620 on Build GRCh38
rs867751149 Chr.22: 39995484-39995484 on Build GRCh38
rs977913538 Chr.22: 39995851-39995851 on Build GRCh38
rs1037832542 Chr.22: 39995820-39995820 on Build GRCh38
rs913858106 Chr.22: 39995707-39995707 on Build GRCh38
rs544430186 Chr.22: 39995615-39995615 on Build GRCh38
rs984444386 Chr.22: 39995459-39995459 on Build GRCh38
rs1052424041 Chr.22: 39995462-39995462 on Build GRCh38
rs147517818 Chr.22: 39995735-39995735 on Build GRCh38
rs758846476 Chr.22: 39995521-39995521 on Build GRCh38
rs1008296026 Chr.22: 39995897-39995897 on Build GRCh38
rs753940111 Chr.22: 39995817-39995817 on Build GRCh38
rs754909455 Chr.22: 39995522-39995522 on Build GRCh38
rs1041691812 Chr.22: 39995574-39995574 on Build GRCh38
rs1016452598 Chr.22: 39995541-39995541 on Build GRCh38
rs561110126 Chr.22: 39995721-39995721 on Build GRCh38
rs963597547 Chr.22: 39995553-39995553 on Build GRCh38
rs894758839 Chr.22: 39995616-39995616 on Build GRCh38
rs1000413418 Chr.22: 39995585-39995585 on Build GRCh38
rs1054409324 Chr.22: 39995864-39995864 on Build GRCh38
rs975188040 Chr.22: 39995622-39995622 on Build GRCh38
rs1052773516 Chr.22: 39995669-39995669 on Build GRCh38
rs755071875 Chr.22: 39995573-39995573 on Build GRCh38
rs905247950 Chr.22: 39995752-39995752 on Build GRCh38
rs751632286 Chr.22: 39995558-39995558 on Build GRCh38
rs577105797 Chr.22: 39995554-39995554 on Build GRCh38
rs993716872 Chr.22: 39995837-39995837 on Build GRCh38
rs951115073 Chr.22: 39995702-39995702 on Build GRCh38
rs924758968 Chr.22: 39995859-39995859 on Build GRCh38
rs935986769 Chr.22: 39995861-39995861 on Build GRCh38

View Full Product Details