Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 43530644-43531138 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFCAB6
Entrez Gene ID
64800
Gene Name
EF-hand calcium binding domain 6
Gene Aliases
DJBP, HSCBCIP1, dJ185D5.1
UniGene
Hs.658996
Gene Chromosome Location
Chr.22: 43528549-43812403 on Build GRCh38
Transcripts
NM_022785.3 NM_198856.2 XM_005261704.2 XM_005261705.3 XM_011530316.1 XM_011530317.2 XM_011530318.2 XM_011530319.2 XM_011530320.2 XM_011530321.2 XM_011530322.2 XM_011530323.2 XM_011530325.2 XM_011530327.2 XM_011530329.1 XM_017028910.1 XM_017028911.1 XM_017028912.1 NR_046563.1

Gene Symbol
EFCAB6-AS1
Entrez Gene ID
100874197
Gene Name
EFCAB6 antisense RNA 1
Gene Aliases
-
UniGene
Hs.627997
Gene Chromosome Location
Chr.22: 43516254-43536605 on Build GRCh38
Transcripts
NM_022785.3 NM_198856.2 XM_005261704.2 XM_005261705.3 XM_011530316.1 XM_011530317.2 XM_011530318.2 XM_011530319.2 XM_011530320.2 XM_011530321.2 XM_011530322.2 XM_011530323.2 XM_011530325.2 XM_011530327.2 XM_011530329.1 XM_017028910.1 XM_017028911.1 XM_017028912.1 NR_046563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6651933 c.4298T>C p.V1433A Chr.22: 43530900-43530900 on Build GRCh38
3390270 c.4365C>T p.S1455S Chr.22: 43530833-43530833 on Build GRCh38
3555137 c.4237G>A p.E1413K Chr.22: 43530961-43530961 on Build GRCh38
3555136 c.4335C>T p.S1445S Chr.22: 43530863-43530863 on Build GRCh38
3555135 c.4372G>A p.D1458N Chr.22: 43530826-43530826 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752691879 Chr.22: 43530967-43530967 on Build GRCh38
rs541720424 Chr.22: 43530799-43530799 on Build GRCh38
rs761597238 Chr.22: 43530859-43530859 on Build GRCh38
rs1006935458 Chr.22: 43530905-43530905 on Build GRCh38
rs781458260 Chr.22: 43530780-43530780 on Build GRCh38
rs898971715 Chr.22: 43530957-43530957 on Build GRCh38
rs547196409 Chr.22: 43530962-43530962 on Build GRCh38
rs76475527 Chr.22: 43530978-43530987 on Build GRCh38
rs71313354 Chr.22: 43531016-43531017 on Build GRCh38
rs750687549 Chr.22: 43530826-43530826 on Build GRCh38
rs956576858 Chr.22: 43531057-43531057 on Build GRCh38
rs771631087 Chr.22: 43530798-43530798 on Build GRCh38
rs201816198 Chr.22: 43530849-43530849 on Build GRCh38
rs151055081 Chr.22: 43530878-43530878 on Build GRCh38
rs868031539 Chr.22: 43530939-43530939 on Build GRCh38
rs768007652 Chr.22: 43530856-43530856 on Build GRCh38
rs141167337 Chr.22: 43530918-43530918 on Build GRCh38
rs141680193 Chr.22: 43530706-43530706 on Build GRCh38
rs769545422 Chr.22: 43530727-43530727 on Build GRCh38
rs1029579323 Chr.22: 43530848-43530848 on Build GRCh38
rs769389964 Chr.22: 43531009-43531009 on Build GRCh38
rs151282295 Chr.22: 43530835-43530835 on Build GRCh38
rs373487037 Chr.22: 43531087-43531087 on Build GRCh38
rs761698490 Chr.22: 43530815-43530815 on Build GRCh38
rs745591218 Chr.22: 43530841-43530841 on Build GRCh38
rs750632684 Chr.22: 43530770-43530770 on Build GRCh38
rs911189201 Chr.22: 43531114-43531114 on Build GRCh38
rs775990059 Chr.22: 43530938-43530938 on Build GRCh38
rs539590253 Chr.22: 43530710-43530710 on Build GRCh38
rs144797247 Chr.22: 43530833-43530833 on Build GRCh38
rs776633265 Chr.22: 43530858-43530858 on Build GRCh38
rs929580202 Chr.22: 43530928-43530928 on Build GRCh38
rs185173041 Chr.22: 43530673-43530673 on Build GRCh38
rs992614985 Chr.22: 43530772-43530772 on Build GRCh38
rs915086992 Chr.22: 43530876-43530876 on Build GRCh38
rs935141947 Chr.22: 43530702-43530702 on Build GRCh38
rs149993265 Chr.22: 43530867-43530867 on Build GRCh38
rs746574690 Chr.22: 43530921-43530921 on Build GRCh38
rs140939056 Chr.22: 43530828-43530828 on Build GRCh38
rs373313660 Chr.22: 43530812-43530812 on Build GRCh38
rs1017559007 Chr.22: 43530764-43530764 on Build GRCh38
rs773194460 Chr.22: 43530760-43530760 on Build GRCh38
rs933970627 Chr.22: 43530880-43530880 on Build GRCh38
rs764067760 Chr.22: 43530811-43530811 on Build GRCh38
rs980898350 Chr.22: 43530894-43530894 on Build GRCh38
rs751667777 Chr.22: 43530829-43530829 on Build GRCh38
rs140830170 Chr.22: 43531085-43531085 on Build GRCh38
rs200105929 Chr.22: 43530972-43530972 on Build GRCh38
rs756690707 Chr.22: 43530971-43530971 on Build GRCh38
rs754300535 Chr.22: 43530975-43530975 on Build GRCh38
rs879015288 Chr.22: 43531036-43531036 on Build GRCh38
rs756578495 Chr.22: 43530893-43530893 on Build GRCh38
rs201765619 Chr.22: 43530802-43530802 on Build GRCh38
rs753309592 Chr.22: 43530890-43530890 on Build GRCh38
rs36016693 Chr.22: 43530980-43530981 on Build GRCh38
rs556052088 Chr.22: 43530712-43530712 on Build GRCh38
rs981045981 Chr.22: 43531092-43531092 on Build GRCh38
rs778147732 Chr.22: 43530903-43530903 on Build GRCh38
rs200656934 Chr.22: 43530861-43530861 on Build GRCh38
rs762796463 Chr.22: 43530769-43530769 on Build GRCh38
rs372164113 Chr.22: 43530774-43530774 on Build GRCh38
rs1051535754 Chr.22: 43531110-43531110 on Build GRCh38
rs976682500 Chr.22: 43530670-43530670 on Build GRCh38
rs568096824 Chr.22: 43530946-43530946 on Build GRCh38
rs762874796 Chr.22: 43530952-43530952 on Build GRCh38
rs376979584 Chr.22: 43530983-43530983 on Build GRCh38
rs529660338 Chr.22: 43530907-43530907 on Build GRCh38
rs555989485 Chr.22: 43530977-43530977 on Build GRCh38
rs35982593 Chr.22: 43530845-43530845 on Build GRCh38
rs764872032 Chr.22: 43530819-43530819 on Build GRCh38
rs773235865 Chr.22: 43530951-43530951 on Build GRCh38
rs757483999 Chr.22: 43530971-43530975 on Build GRCh38
rs988088845 Chr.22: 43531078-43531078 on Build GRCh38
rs569508466 Chr.22: 43531025-43531025 on Build GRCh38
rs763315688 Chr.22: 43530869-43530869 on Build GRCh38
rs766287775 Chr.22: 43530953-43530953 on Build GRCh38
rs963290884 Chr.22: 43530765-43530765 on Build GRCh38
rs889863960 Chr.22: 43530882-43530882 on Build GRCh38
rs62232049 Chr.22: 43530981-43530981 on Build GRCh38
rs767370813 Chr.22: 43530795-43530795 on Build GRCh38
rs751045331 Chr.22: 43530851-43530851 on Build GRCh38
rs374932960 Chr.22: 43530787-43530787 on Build GRCh38
rs954570418 Chr.22: 43530928-43530929 on Build GRCh38
rs751792593 Chr.22: 43530881-43530881 on Build GRCh38
rs780805261 Chr.22: 43530984-43530984 on Build GRCh38
rs398040516 Chr.22: 43531019-43531020 on Build GRCh38
rs778101008 Chr.22: 43530840-43530840 on Build GRCh38
rs144891068 Chr.22: 43530919-43530919 on Build GRCh38
rs190077873 Chr.22: 43531038-43531038 on Build GRCh38
rs1022526145 Chr.22: 43531079-43531079 on Build GRCh38
rs770018138 Chr.22: 43530784-43530784 on Build GRCh38
rs979847396 Chr.22: 43531032-43531032 on Build GRCh38
rs755092656 Chr.22: 43530883-43530883 on Build GRCh38
rs1018332079 Chr.22: 43530818-43530818 on Build GRCh38
rs62232048 Chr.22: 43530978-43530978 on Build GRCh38
rs201426447 Chr.22: 43530771-43530771 on Build GRCh38
rs751791814 Chr.22: 43530929-43530933 on Build GRCh38
rs781738975 Chr.22: 43530830-43530830 on Build GRCh38
rs907582700 Chr.22: 43531032-43531042 on Build GRCh38
rs563974290 Chr.22: 43530888-43530888 on Build GRCh38
rs760641608 Chr.22: 43530807-43530807 on Build GRCh38
rs747633660 Chr.22: 43530991-43530991 on Build GRCh38
rs376668505 Chr.22: 43530965-43530965 on Build GRCh38
rs375580423 Chr.22: 43531103-43531103 on Build GRCh38
rs769176388 Chr.22: 43530776-43530776 on Build GRCh38
rs1045438907 Chr.22: 43530751-43530751 on Build GRCh38
rs1029337369 Chr.22: 43531052-43531052 on Build GRCh38
rs771677982 Chr.22: 43530844-43530844 on Build GRCh38
rs766211358 Chr.22: 43530955-43530955 on Build GRCh38
rs926706076 Chr.22: 43531101-43531101 on Build GRCh38
rs774062183 Chr.22: 43530785-43530785 on Build GRCh38
rs527255915 Chr.22: 43530956-43530956 on Build GRCh38
rs926737361 Chr.22: 43530900-43530900 on Build GRCh38
rs926141220 Chr.22: 43531033-43531033 on Build GRCh38
rs768223676 Chr.22: 43530926-43530926 on Build GRCh38
rs796793248 Chr.22: 43530730-43530731 on Build GRCh38

View Full Product Details