Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 45729339-45729846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATXN10
Entrez Gene ID
25814
Gene Name
ataxin 10
Gene Aliases
E46L, HUMEEP, SCA10
UniGene
Hs.475125
Gene Chromosome Location
Chr.22: 45671798-45845307 on Build GRCh38
Transcripts
NM_001167621.1 NM_013236.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7128744 c.729-1G>A p.? Chr.22: 45729424-45729424 on Build GRCh38
6608679 c.729-8delT p.? Chr.22: 45729417-45729417 on Build GRCh38
7285145 c.753A>G p.I251M Chr.22: 45729449-45729449 on Build GRCh38
4104804 c.764C>T p.T255M Chr.22: 45729460-45729460 on Build GRCh38
4104805 c.797C>G p.P266R Chr.22: 45729493-45729493 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs776429944 Chr.22: 45729731-45729731 on Build GRCh38
rs368814580 Chr.22: 45729581-45729581 on Build GRCh38
rs771885827 Chr.22: 45729452-45729452 on Build GRCh38
rs937532232 Chr.22: 45729434-45729434 on Build GRCh38
rs763991549 Chr.22: 45729512-45729512 on Build GRCh38
rs769181505 Chr.22: 45729539-45729539 on Build GRCh38
rs532347943 Chr.22: 45729817-45729817 on Build GRCh38
rs748465267 Chr.22: 45729695-45729695 on Build GRCh38
rs756105988 Chr.22: 45729551-45729552 on Build GRCh38
rs745375792 Chr.22: 45729527-45729527 on Build GRCh38
rs189025930 Chr.22: 45729786-45729786 on Build GRCh38
rs148992434 Chr.22: 45729566-45729566 on Build GRCh38
rs960397 Chr.22: 45729693-45729693 on Build GRCh38
rs763121141 Chr.22: 45729486-45729486 on Build GRCh38
rs377292388 Chr.22: 45729634-45729634 on Build GRCh38
rs138453547 Chr.22: 45729562-45729562 on Build GRCh38
rs765408301 Chr.22: 45729479-45729479 on Build GRCh38
rs777263532 Chr.22: 45729464-45729464 on Build GRCh38
rs568068318 Chr.22: 45729478-45729478 on Build GRCh38
rs749115201 Chr.22: 45729757-45729758 on Build GRCh38
rs749939716 Chr.22: 45729617-45729617 on Build GRCh38
rs745873539 Chr.22: 45729633-45729633 on Build GRCh38
rs967751097 Chr.22: 45729718-45729718 on Build GRCh38
rs775448122 Chr.22: 45729754-45729754 on Build GRCh38
rs773185560 Chr.22: 45729484-45729484 on Build GRCh38
rs765937326 Chr.22: 45729788-45729788 on Build GRCh38
rs760443327 Chr.22: 45729583-45729583 on Build GRCh38
rs144613992 Chr.22: 45729416-45729416 on Build GRCh38
rs954893372 Chr.22: 45729378-45729378 on Build GRCh38
rs750427928 Chr.22: 45729756-45729756 on Build GRCh38
rs927568754 Chr.22: 45729784-45729784 on Build GRCh38
rs368442867 Chr.22: 45729792-45729792 on Build GRCh38
rs751303669 Chr.22: 45729657-45729657 on Build GRCh38
rs778197338 Chr.22: 45729702-45729702 on Build GRCh38
rs752429212 Chr.22: 45729504-45729504 on Build GRCh38
rs994243932 Chr.22: 45729455-45729455 on Build GRCh38
rs758050698 Chr.22: 45729623-45729623 on Build GRCh38
rs754698393 Chr.22: 45729662-45729662 on Build GRCh38
rs763903931 Chr.22: 45729596-45729596 on Build GRCh38
rs1057473358 Chr.22: 45729448-45729448 on Build GRCh38
rs761572224 Chr.22: 45729741-45729741 on Build GRCh38
rs200043904 Chr.22: 45729404-45729404 on Build GRCh38
rs769759482 Chr.22: 45729734-45729734 on Build GRCh38
rs369463744 Chr.22: 45729629-45729629 on Build GRCh38
rs768568088 Chr.22: 45729758-45729759 on Build GRCh38
rs376280245 Chr.22: 45729497-45729497 on Build GRCh38
rs988074604 Chr.22: 45729379-45729379 on Build GRCh38
rs761766866 Chr.22: 45729466-45729466 on Build GRCh38
rs779822907 Chr.22: 45729656-45729656 on Build GRCh38
rs5764830 Chr.22: 45729766-45729766 on Build GRCh38
rs757097542 Chr.22: 45729559-45729559 on Build GRCh38
rs375099471 Chr.22: 45729546-45729546 on Build GRCh38
rs745479298 Chr.22: 45729447-45729447 on Build GRCh38
rs117607950 Chr.22: 45729616-45729616 on Build GRCh38
rs762529402 Chr.22: 45729782-45729782 on Build GRCh38
rs770274640 Chr.22: 45729697-45729697 on Build GRCh38
rs368066531 Chr.22: 45729395-45729395 on Build GRCh38
rs541097591 Chr.22: 45729707-45729707 on Build GRCh38
rs756836344 Chr.22: 45729442-45729442 on Build GRCh38
rs952423500 Chr.22: 45729805-45729805 on Build GRCh38
rs141434930 Chr.22: 45729600-45729600 on Build GRCh38
rs937627645 Chr.22: 45729815-45729815 on Build GRCh38
rs747374366 Chr.22: 45729532-45729532 on Build GRCh38
rs149859910 Chr.22: 45729460-45729460 on Build GRCh38
rs772191665 Chr.22: 45729730-45729730 on Build GRCh38
rs373925397 Chr.22: 45729380-45729380 on Build GRCh38
rs202151442 Chr.22: 45729505-45729505 on Build GRCh38
rs145859375 Chr.22: 45729520-45729520 on Build GRCh38
rs778412063 Chr.22: 45729444-45729444 on Build GRCh38
rs1043333507 Chr.22: 45729370-45729370 on Build GRCh38
rs757155741 Chr.22: 45729612-45729612 on Build GRCh38
rs201298547 Chr.22: 45729558-45729558 on Build GRCh38
rs768152255 Chr.22: 45729551-45729551 on Build GRCh38
rs186780419 Chr.22: 45729582-45729582 on Build GRCh38
rs771050655 Chr.22: 45729714-45729714 on Build GRCh38
rs753866615 Chr.22: 45729519-45729519 on Build GRCh38
rs528589993 Chr.22: 45729413-45729413 on Build GRCh38
rs748731082 Chr.22: 45729428-45729428 on Build GRCh38
rs777697084 Chr.22: 45729427-45729427 on Build GRCh38
rs779910802 Chr.22: 45729531-45729531 on Build GRCh38
rs775931227 Chr.22: 45729454-45729454 on Build GRCh38
rs779799565 Chr.22: 45729693-45729695 on Build GRCh38
rs778449268 Chr.22: 45729521-45729521 on Build GRCh38
rs566477401 Chr.22: 45729715-45729715 on Build GRCh38
rs1018193075 Chr.22: 45729735-45729735 on Build GRCh38
rs547165095 Chr.22: 45729425-45729425 on Build GRCh38
rs577503398 Chr.22: 45729529-45729529 on Build GRCh38
rs919601839 Chr.22: 45729500-45729500 on Build GRCh38
rs1021133966 Chr.22: 45729377-45729377 on Build GRCh38
rs759719422 Chr.22: 45729724-45729724 on Build GRCh38
rs1009782450 Chr.22: 45729363-45729363 on Build GRCh38
rs548270176 Chr.22: 45729461-45729461 on Build GRCh38
rs949316184 Chr.22: 45729548-45729548 on Build GRCh38
rs776980825 Chr.22: 45729543-45729543 on Build GRCh38
rs893821940 Chr.22: 45729677-45729678 on Build GRCh38
rs750697028 Chr.22: 45729393-45729393 on Build GRCh38

View Full Product Details