Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46051740-46052275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR34
Entrez Gene ID
55267
Gene Name
proline rich 34
Gene Aliases
C22orf26
UniGene
Hs.567529
Gene Chromosome Location
Chr.22: 46050459-46054144 on Build GRCh38
Transcripts
NM_018280.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4419794 c.410A>G p.Q137R Chr.22: 46051834-46051834 on Build GRCh38
3396408 c.396T>A p.N132K Chr.22: 46051848-46051848 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs568263722 Chr.22: 46051951-46051951 on Build GRCh38
rs995442768 Chr.22: 46051819-46051819 on Build GRCh38
rs1008195828 Chr.22: 46052106-46052106 on Build GRCh38
rs915570520 Chr.22: 46051955-46051955 on Build GRCh38
rs766794558 Chr.22: 46051955-46051956 on Build GRCh38
rs1020044486 Chr.22: 46052113-46052113 on Build GRCh38
rs981518089 Chr.22: 46052019-46052019 on Build GRCh38
rs187507363 Chr.22: 46052167-46052167 on Build GRCh38
rs928268689 Chr.22: 46052222-46052222 on Build GRCh38
rs374143698 Chr.22: 46051795-46051795 on Build GRCh38
rs1032941085 Chr.22: 46051881-46051881 on Build GRCh38
rs150118279 Chr.22: 46051858-46051858 on Build GRCh38
rs537958831 Chr.22: 46052055-46052055 on Build GRCh38
rs908581651 Chr.22: 46052176-46052177 on Build GRCh38
rs893019332 Chr.22: 46052064-46052064 on Build GRCh38
rs775165517 Chr.22: 46051875-46051875 on Build GRCh38
rs931207307 Chr.22: 46052073-46052073 on Build GRCh38
rs1032497396 Chr.22: 46052144-46052144 on Build GRCh38
rs952763739 Chr.22: 46051927-46051927 on Build GRCh38
rs565439736 Chr.22: 46052125-46052125 on Build GRCh38
rs534051146 Chr.22: 46051981-46051981 on Build GRCh38
rs115090509 Chr.22: 46051840-46051840 on Build GRCh38
rs770514272 Chr.22: 46051775-46051775 on Build GRCh38
rs970962738 Chr.22: 46052208-46052208 on Build GRCh38
rs983026159 Chr.22: 46052172-46052172 on Build GRCh38
rs765142921 Chr.22: 46051859-46051859 on Build GRCh38
rs759074947 Chr.22: 46051852-46051852 on Build GRCh38
rs575656614 Chr.22: 46052051-46052051 on Build GRCh38
rs1023804233 Chr.22: 46052182-46052182 on Build GRCh38
rs182815432 Chr.22: 46052133-46052133 on Build GRCh38
rs961169831 Chr.22: 46052226-46052226 on Build GRCh38
rs140761102 Chr.22: 46051764-46051764 on Build GRCh38
rs949483462 Chr.22: 46052026-46052026 on Build GRCh38
rs762186789 Chr.22: 46051916-46051916 on Build GRCh38
rs558725372 Chr.22: 46052036-46052036 on Build GRCh38
rs140248907 Chr.22: 46052030-46052030 on Build GRCh38
rs138519628 Chr.22: 46052027-46052027 on Build GRCh38
rs774684637 Chr.22: 46051813-46051813 on Build GRCh38
rs937134614 Chr.22: 46052048-46052048 on Build GRCh38
rs964522092 Chr.22: 46051855-46051855 on Build GRCh38
rs1011472668 Chr.22: 46052065-46052065 on Build GRCh38
rs948429325 Chr.22: 46052007-46052007 on Build GRCh38
rs898384617 Chr.22: 46051817-46051817 on Build GRCh38
rs368357852 Chr.22: 46051821-46051821 on Build GRCh38
rs555119592 Chr.22: 46052097-46052097 on Build GRCh38
rs767742979 Chr.22: 46051916-46051916 on Build GRCh38
rs577165361 Chr.22: 46052214-46052214 on Build GRCh38
rs547727725 Chr.22: 46051985-46051985 on Build GRCh38
rs547121870 Chr.22: 46051849-46051849 on Build GRCh38
rs555883946 Chr.22: 46051848-46051849 on Build GRCh38
rs889707625 Chr.22: 46052104-46052104 on Build GRCh38
rs551915555 Chr.22: 46051838-46051838 on Build GRCh38
rs986500098 Chr.22: 46051841-46051841 on Build GRCh38
rs559686846 Chr.22: 46051820-46051820 on Build GRCh38
rs59929908 Chr.22: 46051834-46051834 on Build GRCh38
rs920144382 Chr.22: 46051897-46051897 on Build GRCh38
rs967203151 Chr.22: 46051871-46051871 on Build GRCh38
rs1034639076 Chr.22: 46051800-46051800 on Build GRCh38
rs902855512 Chr.22: 46052103-46052103 on Build GRCh38
rs545032886 Chr.22: 46052168-46052168 on Build GRCh38
rs762746798 Chr.22: 46051950-46051950 on Build GRCh38
rs928453511 Chr.22: 46052035-46052035 on Build GRCh38
rs573837925 Chr.22: 46051763-46051763 on Build GRCh38
rs141796966 Chr.22: 46051915-46051916 on Build GRCh38

View Full Product Details