Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46053741-46054275 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PRR34
Entrez Gene ID
55267
Gene Name
proline rich 34
Gene Aliases
C22orf26
UniGene
Hs.567529
Gene Chromosome Location
Chr.22: 46050459-46054144 on Build GRCh38
Transcripts
NM_018280.2 NR_027034.1

Gene Symbol
PRR34-AS1
Entrez Gene ID
150381
Gene Name
PRR34 antisense RNA 1
Gene Aliases
-
UniGene
Hs.659113
Gene Chromosome Location
Chr.22: 46053846-46058522 on Build GRCh38
Transcripts
NM_018280.2 NR_027034.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3749771 c.83C>T p.P28L Chr.22: 46054011-46054011 on Build GRCh38
6047050 c.45C>T p.C15C Chr.22: 46054049-46054049 on Build GRCh38
6212434 c.14C>T p.A5V Chr.22: 46054080-46054080 on Build GRCh38
3992227 c.291C>T p.H97H Chr.22: 46053803-46053803 on Build GRCh38
399898 c.320G>C p.R107P Chr.22: 46053774-46053774 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761376202 Chr.22: 46053827-46053827 on Build GRCh38
rs997982530 Chr.22: 46054155-46054155 on Build GRCh38
rs55895702 Chr.22: 46054181-46054181 on Build GRCh38
rs561749473 Chr.22: 46053775-46053775 on Build GRCh38
rs896415177 Chr.22: 46054223-46054223 on Build GRCh38
rs751619335 Chr.22: 46053862-46053862 on Build GRCh38
rs200221204 Chr.22: 46053930-46053930 on Build GRCh38
rs1010360032 Chr.22: 46054178-46054178 on Build GRCh38
rs747819929 Chr.22: 46053975-46053975 on Build GRCh38
rs976652138 Chr.22: 46053774-46053774 on Build GRCh38
rs777358437 Chr.22: 46054023-46054023 on Build GRCh38
rs992284323 Chr.22: 46054227-46054227 on Build GRCh38
rs563508064 Chr.22: 46054116-46054116 on Build GRCh38
rs527752877 Chr.22: 46053789-46053789 on Build GRCh38
rs762139185 Chr.22: 46054140-46054140 on Build GRCh38
rs759968688 Chr.22: 46054005-46054005 on Build GRCh38
rs745742400 Chr.22: 46053801-46053801 on Build GRCh38
rs1012304553 Chr.22: 46054014-46054014 on Build GRCh38
rs112405410 Chr.22: 46054012-46054012 on Build GRCh38
rs755039357 Chr.22: 46053864-46053864 on Build GRCh38
rs577714787 Chr.22: 46054016-46054016 on Build GRCh38
rs960613840 Chr.22: 46053860-46053860 on Build GRCh38
rs887689218 Chr.22: 46054125-46054125 on Build GRCh38
rs937893975 Chr.22: 46053863-46053863 on Build GRCh38
rs1056318098 Chr.22: 46054102-46054102 on Build GRCh38
rs780746082 Chr.22: 46054013-46054013 on Build GRCh38
rs772194573 Chr.22: 46053803-46053803 on Build GRCh38
rs924097455 Chr.22: 46053933-46053933 on Build GRCh38
rs780773169 Chr.22: 46054009-46054010 on Build GRCh38
rs767528878 Chr.22: 46053875-46053875 on Build GRCh38
rs926259120 Chr.22: 46054200-46054200 on Build GRCh38
rs771222343 Chr.22: 46054028-46054028 on Build GRCh38
rs764299168 Chr.22: 46054124-46054124 on Build GRCh38
rs1039038221 Chr.22: 46054133-46054133 on Build GRCh38
rs746973500 Chr.22: 46053966-46053966 on Build GRCh38
rs767579361 Chr.22: 46054006-46054006 on Build GRCh38
rs757040984 Chr.22: 46053791-46053791 on Build GRCh38
rs768145869 Chr.22: 46053967-46053967 on Build GRCh38
rs550394595 Chr.22: 46053998-46053998 on Build GRCh38
rs1023778638 Chr.22: 46053865-46053865 on Build GRCh38
rs773532515 Chr.22: 46053996-46053996 on Build GRCh38
rs776018298 Chr.22: 46053974-46053974 on Build GRCh38
rs772521729 Chr.22: 46054085-46054085 on Build GRCh38
rs12159707 Chr.22: 46054011-46054011 on Build GRCh38
rs752779459 Chr.22: 46054008-46054008 on Build GRCh38
rs745726071 Chr.22: 46053942-46053942 on Build GRCh38
rs968930486 Chr.22: 46054192-46054192 on Build GRCh38
rs1036410427 Chr.22: 46054175-46054175 on Build GRCh38
rs763442138 Chr.22: 46053853-46053853 on Build GRCh38
rs868769748 Chr.22: 46054067-46054067 on Build GRCh38
rs917759703 Chr.22: 46054246-46054246 on Build GRCh38
rs769462537 Chr.22: 46054020-46054020 on Build GRCh38
rs765578365 Chr.22: 46053840-46053840 on Build GRCh38
rs929161908 Chr.22: 46054248-46054248 on Build GRCh38
rs959165155 Chr.22: 46054205-46054205 on Build GRCh38
rs937740434 Chr.22: 46054100-46054100 on Build GRCh38
rs766909643 Chr.22: 46053855-46053855 on Build GRCh38
rs774714903 Chr.22: 46054044-46054044 on Build GRCh38
rs185392710 Chr.22: 46054009-46054009 on Build GRCh38
rs754044519 Chr.22: 46054129-46054129 on Build GRCh38
rs993797443 Chr.22: 46053949-46053949 on Build GRCh38
rs745393787 Chr.22: 46054111-46054111 on Build GRCh38
rs935618089 Chr.22: 46053934-46053934 on Build GRCh38
rs897104408 Chr.22: 46053848-46053848 on Build GRCh38
rs1056404304 Chr.22: 46054218-46054218 on Build GRCh38
rs369307763 Chr.22: 46053872-46053872 on Build GRCh38
rs758268375 Chr.22: 46054007-46054008 on Build GRCh38
rs752900960 Chr.22: 46053901-46053901 on Build GRCh38
rs374981205 Chr.22: 46054010-46054010 on Build GRCh38
rs1030510153 Chr.22: 46054166-46054166 on Build GRCh38
rs760781615 Chr.22: 46053779-46053780 on Build GRCh38
rs887743015 Chr.22: 46053976-46053976 on Build GRCh38
rs946425847 Chr.22: 46054039-46054039 on Build GRCh38
rs372768937 Chr.22: 46054094-46054094 on Build GRCh38
rs1023792520 Chr.22: 46054107-46054121 on Build GRCh38
rs765272321 Chr.22: 46053780-46053781 on Build GRCh38
rs547499372 Chr.22: 46053910-46053910 on Build GRCh38
rs1006258678 Chr.22: 46054128-46054128 on Build GRCh38
rs747647754 Chr.22: 46054018-46054018 on Build GRCh38
rs934893693 Chr.22: 46053780-46053780 on Build GRCh38
rs766262494 Chr.22: 46054141-46054141 on Build GRCh38
rs1034733144 Chr.22: 46053856-46053856 on Build GRCh38
rs776304261 Chr.22: 46053820-46053820 on Build GRCh38
rs376335148 Chr.22: 46053787-46053787 on Build GRCh38
rs559208308 Chr.22: 46054008-46054009 on Build GRCh38
rs1043419154 Chr.22: 46053825-46053825 on Build GRCh38
rs773620461 Chr.22: 46053850-46053850 on Build GRCh38
rs763769965 Chr.22: 46054008-46054008 on Build GRCh38
rs748973634 Chr.22: 46054027-46054027 on Build GRCh38
rs1056687579 Chr.22: 46053868-46053868 on Build GRCh38
rs775137475 Chr.22: 46053814-46053814 on Build GRCh38
rs769395466 Chr.22: 46053978-46053978 on Build GRCh38
rs956364448 Chr.22: 46054174-46054174 on Build GRCh38
rs956829489 Chr.22: 46054080-46054080 on Build GRCh38
rs199581388 Chr.22: 46053961-46053961 on Build GRCh38
rs756890481 Chr.22: 46054010-46054010 on Build GRCh38
rs917714504 Chr.22: 46054114-46054114 on Build GRCh38
rs778581399 Chr.22: 46053797-46053797 on Build GRCh38
rs929769003 Chr.22: 46054182-46054182 on Build GRCh38
rs778707515 Chr.22: 46054235-46054235 on Build GRCh38
rs968260981 Chr.22: 46053927-46053927 on Build GRCh38
rs570240816 Chr.22: 46054003-46054003 on Build GRCh38
rs760075737 Chr.22: 46054083-46054083 on Build GRCh38
rs752674927 Chr.22: 46053837-46053837 on Build GRCh38
rs746558867 Chr.22: 46053817-46053817 on Build GRCh38
rs979090528 Chr.22: 46054196-46054196 on Build GRCh38
rs768072686 Chr.22: 46053819-46053819 on Build GRCh38
rs758380292 Chr.22: 46053943-46053943 on Build GRCh38
rs766754694 Chr.22: 46053999-46053999 on Build GRCh38

View Full Product Details