Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46361770-46362282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR1
Entrez Gene ID
9620
Gene Name
cadherin EGF LAG seven-pass G-type receptor 1
Gene Aliases
ADGRC1, CDHF9, FMI2, HFMI2, ME2
UniGene
Hs.252387
Gene Chromosome Location
Chr.22: 46360834-46537502 on Build GRCh38
Transcripts
NM_014246.1 XM_006724383.3 XM_011530554.2 XM_011530555.2

SNP Detail

SNP ID Chromosome Location
rs558537643 Chr.22: 46362066-46362066 on Build GRCh38
rs761871054 Chr.22: 46362152-46362152 on Build GRCh38
rs1053437328 Chr.22: 46361830-46361830 on Build GRCh38
rs3747249 Chr.22: 46361887-46361887 on Build GRCh38
rs929897718 Chr.22: 46362226-46362226 on Build GRCh38
rs774231244 Chr.22: 46362084-46362084 on Build GRCh38
rs967768571 Chr.22: 46362070-46362070 on Build GRCh38
rs768283250 Chr.22: 46362068-46362068 on Build GRCh38
rs978102000 Chr.22: 46362228-46362228 on Build GRCh38
rs979048083 Chr.22: 46361854-46361854 on Build GRCh38
rs772297530 Chr.22: 46362235-46362235 on Build GRCh38
rs536403104 Chr.22: 46361969-46361969 on Build GRCh38
rs769495860 Chr.22: 46361865-46361865 on Build GRCh38
rs571531942 Chr.22: 46361866-46361866 on Build GRCh38
rs532478561 Chr.22: 46362245-46362245 on Build GRCh38
rs375310374 Chr.22: 46361958-46361958 on Build GRCh38
rs942057830 Chr.22: 46362201-46362201 on Build GRCh38
rs1044722346 Chr.22: 46362253-46362253 on Build GRCh38
rs1012075686 Chr.22: 46362246-46362246 on Build GRCh38
rs560388681 Chr.22: 46362236-46362236 on Build GRCh38
rs1050528721 Chr.22: 46361893-46361893 on Build GRCh38
rs752667103 Chr.22: 46361960-46361960 on Build GRCh38
rs1036364813 Chr.22: 46362211-46362211 on Build GRCh38
rs909328742 Chr.22: 46362174-46362174 on Build GRCh38
rs915000363 Chr.22: 46362033-46362033 on Build GRCh38
rs749292820 Chr.22: 46362024-46362024 on Build GRCh38
rs893514448 Chr.22: 46361837-46361837 on Build GRCh38
rs539413475 Chr.22: 46361851-46361852 on Build GRCh38
rs79203484 Chr.22: 46362002-46362002 on Build GRCh38
rs914967800 Chr.22: 46362249-46362249 on Build GRCh38
rs953467219 Chr.22: 46362148-46362148 on Build GRCh38
rs1019378651 Chr.22: 46362162-46362162 on Build GRCh38
rs897703166 Chr.22: 46362106-46362106 on Build GRCh38
rs1000522765 Chr.22: 46362097-46362097 on Build GRCh38
rs906420928 Chr.22: 46362053-46362053 on Build GRCh38
rs573241384 Chr.22: 46361987-46361987 on Build GRCh38
rs1003898310 Chr.22: 46362065-46362065 on Build GRCh38
rs529926689 Chr.22: 46362151-46362151 on Build GRCh38
rs976331993 Chr.22: 46362123-46362123 on Build GRCh38
rs201772740 Chr.22: 46361976-46361976 on Build GRCh38
rs966716393 Chr.22: 46361990-46361990 on Build GRCh38
rs757266133 Chr.22: 46362252-46362252 on Build GRCh38
rs773067995 Chr.22: 46362250-46362250 on Build GRCh38
rs534981313 Chr.22: 46362053-46362053 on Build GRCh38
rs906442693 Chr.22: 46361872-46361872 on Build GRCh38
rs1053566472 Chr.22: 46362031-46362031 on Build GRCh38
rs576887426 Chr.22: 46362078-46362078 on Build GRCh38
rs555250416 Chr.22: 46362131-46362136 on Build GRCh38
rs953549279 Chr.22: 46362137-46362137 on Build GRCh38
rs962088568 Chr.22: 46361888-46361888 on Build GRCh38
rs947704707 Chr.22: 46362042-46362042 on Build GRCh38
rs966309123 Chr.22: 46362219-46362219 on Build GRCh38
rs973482034 Chr.22: 46361898-46361898 on Build GRCh38
rs1012568634 Chr.22: 46361864-46361864 on Build GRCh38
rs560934536 Chr.22: 46362117-46362117 on Build GRCh38
rs760283110 Chr.22: 46361858-46361859 on Build GRCh38
rs1045147688 Chr.22: 46362046-46362046 on Build GRCh38
rs977590986 Chr.22: 46361995-46361995 on Build GRCh38
rs924764467 Chr.22: 46362230-46362230 on Build GRCh38
rs1027662306 Chr.22: 46362119-46362119 on Build GRCh38
rs544308675 Chr.22: 46362105-46362105 on Build GRCh38
rs575113430 Chr.22: 46362102-46362102 on Build GRCh38
rs557141289 Chr.22: 46361922-46361922 on Build GRCh38
rs546858499 Chr.22: 46362206-46362206 on Build GRCh38
rs1003354666 Chr.22: 46361886-46361886 on Build GRCh38
rs967716300 Chr.22: 46361815-46361815 on Build GRCh38

View Full Product Details