Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 46365419-46365818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CELSR1
Entrez Gene ID
9620
Gene Name
cadherin EGF LAG seven-pass G-type receptor 1
Gene Aliases
ADGRC1, CDHF9, FMI2, HFMI2, ME2
UniGene
Hs.252387
Gene Chromosome Location
Chr.22: 46360834-46537502 on Build GRCh38
Transcripts
NM_014246.1 XM_006724383.3 XM_011530554.2 XM_011530555.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5603149 c.8345G>A p.R2782K Chr.22: 46365645-46365645 on Build GRCh38
6491623 c.8309G>A p.G2770E Chr.22: 46365681-46365681 on Build GRCh38
4630757 c.8325C>T p.G2775G Chr.22: 46365665-46365665 on Build GRCh38
4801315 c.8355C>T p.H2785H Chr.22: 46365635-46365635 on Build GRCh38
6203382 c.8404+3_8404+6delATGG p.? Chr.22: 46365580-46365583 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs966160558 Chr.22: 46365747-46365747 on Build GRCh38
rs777881067 Chr.22: 46365553-46365553 on Build GRCh38
rs754156637 Chr.22: 46365482-46365482 on Build GRCh38
rs367621773 Chr.22: 46365590-46365590 on Build GRCh38
rs999029366 Chr.22: 46365751-46365751 on Build GRCh38
rs112675709 Chr.22: 46365694-46365694 on Build GRCh38
rs376857359 Chr.22: 46365660-46365660 on Build GRCh38
rs764547876 Chr.22: 46365614-46365614 on Build GRCh38
rs561098968 Chr.22: 46365592-46365592 on Build GRCh38
rs746727825 Chr.22: 46365683-46365683 on Build GRCh38
rs148591220 Chr.22: 46365634-46365634 on Build GRCh38
rs976535538 Chr.22: 46365612-46365612 on Build GRCh38
rs764601299 Chr.22: 46365676-46365676 on Build GRCh38
rs34184256 Chr.22: 46365668-46365668 on Build GRCh38
rs758732699 Chr.22: 46365700-46365700 on Build GRCh38
rs1031147106 Chr.22: 46365759-46365759 on Build GRCh38
rs745344713 Chr.22: 46365737-46365737 on Build GRCh38
rs75999278 Chr.22: 46365477-46365477 on Build GRCh38
rs553361263 Chr.22: 46365543-46365543 on Build GRCh38
rs34191340 Chr.22: 46365671-46365671 on Build GRCh38
rs991972792 Chr.22: 46365792-46365794 on Build GRCh38
rs777804762 Chr.22: 46365654-46365654 on Build GRCh38
rs745559079 Chr.22: 46365667-46365667 on Build GRCh38
rs371505101 Chr.22: 46365622-46365622 on Build GRCh38
rs75966955 Chr.22: 46365558-46365558 on Build GRCh38
rs1014924598 Chr.22: 46365540-46365540 on Build GRCh38
rs748844245 Chr.22: 46365658-46365658 on Build GRCh38
rs972031571 Chr.22: 46365754-46365754 on Build GRCh38
rs12165943 Chr.22: 46365600-46365600 on Build GRCh38
rs910950133 Chr.22: 46365448-46365448 on Build GRCh38
rs761213758 Chr.22: 46365613-46365613 on Build GRCh38
rs776931799 Chr.22: 46365688-46365688 on Build GRCh38
rs773899714 Chr.22: 46365665-46365665 on Build GRCh38
rs775811070 Chr.22: 46365601-46365601 on Build GRCh38
rs750491166 Chr.22: 46365699-46365699 on Build GRCh38
rs142042773 Chr.22: 46365664-46365664 on Build GRCh38
rs183562040 Chr.22: 46365544-46365544 on Build GRCh38
rs951262808 Chr.22: 46365778-46365778 on Build GRCh38
rs58143064 Chr.22: 46365750-46365750 on Build GRCh38
rs757230082 Chr.22: 46365620-46365620 on Build GRCh38
rs943792337 Chr.22: 46365452-46365452 on Build GRCh38
rs746830931 Chr.22: 46365562-46365562 on Build GRCh38
rs551863118 Chr.22: 46365474-46365474 on Build GRCh38
rs889358083 Chr.22: 46365721-46365721 on Build GRCh38
rs142095871 Chr.22: 46365704-46365704 on Build GRCh38
rs777620430 Chr.22: 46365719-46365719 on Build GRCh38
rs367887796 Chr.22: 46365556-46365556 on Build GRCh38
rs762374578 Chr.22: 46365697-46365697 on Build GRCh38
rs1044750252 Chr.22: 46365606-46365606 on Build GRCh38
rs575457963 Chr.22: 46365563-46365563 on Build GRCh38
rs766668014 Chr.22: 46365703-46365703 on Build GRCh38
rs376642404 Chr.22: 46365725-46365725 on Build GRCh38
rs1028293224 Chr.22: 46365579-46365579 on Build GRCh38
rs768018062 Chr.22: 46365539-46365539 on Build GRCh38
rs867424552 Chr.22: 46365690-46365690 on Build GRCh38
rs778328341 Chr.22: 46365732-46365732 on Build GRCh38
rs755089856 Chr.22: 46365514-46365514 on Build GRCh38
rs986311574 Chr.22: 46365438-46365438 on Build GRCh38
rs893739429 Chr.22: 46365525-46365525 on Build GRCh38
rs755914412 Chr.22: 46365716-46365716 on Build GRCh38
rs536399252 Chr.22: 46365771-46365771 on Build GRCh38
rs1019118984 Chr.22: 46365744-46365744 on Build GRCh38
rs78308964 Chr.22: 46365557-46365557 on Build GRCh38
rs1054069681 Chr.22: 46365489-46365489 on Build GRCh38
rs956508332 Chr.22: 46365782-46365782 on Build GRCh38
rs1023685199 Chr.22: 46365471-46365471 on Build GRCh38
rs755992651 Chr.22: 46365464-46365464 on Build GRCh38
rs918512273 Chr.22: 46365766-46365766 on Build GRCh38
rs1007847281 Chr.22: 46365736-46365736 on Build GRCh38
rs143249423 Chr.22: 46365617-46365617 on Build GRCh38
rs781127470 Chr.22: 46365567-46365567 on Build GRCh38
rs748706515 Chr.22: 46365576-46365576 on Build GRCh38
rs756150192 Chr.22: 46365638-46365638 on Build GRCh38
rs537147540 Chr.22: 46365488-46365488 on Build GRCh38
rs544302683 Chr.22: 46365569-46365569 on Build GRCh38
rs147530261 Chr.22: 46365623-46365623 on Build GRCh38
rs199758891 Chr.22: 46365726-46365726 on Build GRCh38
rs750195830 Chr.22: 46365458-46365458 on Build GRCh38
rs200266139 Chr.22: 46365698-46365698 on Build GRCh38
rs181995022 Chr.22: 46365538-46365538 on Build GRCh38
rs567507556 Chr.22: 46365534-46365534 on Build GRCh38
rs747919597 Chr.22: 46365635-46365635 on Build GRCh38
rs557162851 Chr.22: 46365531-46365531 on Build GRCh38
rs953672689 Chr.22: 46365589-46365589 on Build GRCh38
rs904124597 Chr.22: 46365476-46365476 on Build GRCh38

View Full Product Details