Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49775892-49776422 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD1
Entrez Gene ID
23774
Gene Name
bromodomain containing 1
Gene Aliases
BRL, BRPF1, BRPF2
UniGene
Hs.127950
Gene Chromosome Location
Chr.22: 49773278-49827927 on Build GRCh38
Transcripts
NM_001304808.1 NM_001304809.1 XM_011530073.1 XM_017028714.1 XM_017028715.1 XM_017028716.1 XM_017028717.1 XM_017028718.1 XM_017028722.1 XM_017028723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3913023 c.2001C>T p.P667P Chr.22: 49776065-49776065 on Build GRCh38
1035222 c.2009C>A p.P670Q Chr.22: 49776057-49776057 on Build GRCh38
4666073 c.2779C>G p.L927V Chr.22: 49776109-49776109 on Build GRCh38
4104931 c.2819A>G p.Y940C Chr.22: 49776069-49776069 on Build GRCh38
6419597 c.1981G>T p.A661S Chr.22: 49776085-49776085 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762088706 Chr.22: 49776037-49776037 on Build GRCh38
rs534183911 Chr.22: 49775992-49775992 on Build GRCh38
rs756686235 Chr.22: 49776162-49776162 on Build GRCh38
rs561713080 Chr.22: 49776119-49776119 on Build GRCh38
rs905201434 Chr.22: 49776225-49776225 on Build GRCh38
rs905221054 Chr.22: 49776338-49776338 on Build GRCh38
rs544333332 Chr.22: 49776129-49776129 on Build GRCh38
rs1052498441 Chr.22: 49775968-49775968 on Build GRCh38
rs758892851 Chr.22: 49776293-49776293 on Build GRCh38
rs752628123 Chr.22: 49776056-49776056 on Build GRCh38
rs372292607 Chr.22: 49776132-49776132 on Build GRCh38
rs539602627 Chr.22: 49776292-49776292 on Build GRCh38
rs777588292 Chr.22: 49776024-49776024 on Build GRCh38
rs967790351 Chr.22: 49776251-49776251 on Build GRCh38
rs888512992 Chr.22: 49776218-49776218 on Build GRCh38
rs755696832 Chr.22: 49776062-49776062 on Build GRCh38
rs1041199045 Chr.22: 49776130-49776130 on Build GRCh38
rs759393550 Chr.22: 49776128-49776128 on Build GRCh38
rs920962923 Chr.22: 49776234-49776234 on Build GRCh38
rs574394226 Chr.22: 49776275-49776275 on Build GRCh38
rs780615040 Chr.22: 49776017-49776017 on Build GRCh38
rs558007967 Chr.22: 49776066-49776066 on Build GRCh38
rs746415215 Chr.22: 49776031-49776031 on Build GRCh38
rs555519288 Chr.22: 49775997-49775997 on Build GRCh38
rs532771442 Chr.22: 49775957-49775957 on Build GRCh38
rs571434728 Chr.22: 49776286-49776286 on Build GRCh38
rs939327013 Chr.22: 49776368-49776368 on Build GRCh38
rs909118674 Chr.22: 49775969-49776004 on Build GRCh38
rs1032849407 Chr.22: 49776362-49776362 on Build GRCh38
rs143063901 Chr.22: 49776113-49776113 on Build GRCh38
rs1007874885 Chr.22: 49776009-49776009 on Build GRCh38
rs567081609 Chr.22: 49775991-49775991 on Build GRCh38
rs549886481 Chr.22: 49776277-49776277 on Build GRCh38
rs193184062 Chr.22: 49776034-49776034 on Build GRCh38
rs956775934 Chr.22: 49776376-49776376 on Build GRCh38
rs369838301 Chr.22: 49776190-49776190 on Build GRCh38
rs984047544 Chr.22: 49775930-49775931 on Build GRCh38
rs537916013 Chr.22: 49776254-49776254 on Build GRCh38
rs1000845411 Chr.22: 49776358-49776358 on Build GRCh38
rs988294810 Chr.22: 49776215-49776215 on Build GRCh38
rs370656126 Chr.22: 49776041-49776041 on Build GRCh38
rs184639990 Chr.22: 49776043-49776043 on Build GRCh38
rs948234780 Chr.22: 49775931-49775931 on Build GRCh38
rs1005521072 Chr.22: 49776168-49776169 on Build GRCh38
rs1056925838 Chr.22: 49776276-49776276 on Build GRCh38
rs922847629 Chr.22: 49776270-49776270 on Build GRCh38
rs947870596 Chr.22: 49775938-49775938 on Build GRCh38
rs1023629485 Chr.22: 49776202-49776202 on Build GRCh38
rs769710560 Chr.22: 49776195-49776195 on Build GRCh38
rs28452394 Chr.22: 49775919-49775919 on Build GRCh38
rs916537024 Chr.22: 49775931-49775931 on Build GRCh38
rs935086186 Chr.22: 49775964-49775964 on Build GRCh38
rs138824 Chr.22: 49776204-49776204 on Build GRCh38
rs753955597 Chr.22: 49776187-49776187 on Build GRCh38
rs748888951 Chr.22: 49776027-49776027 on Build GRCh38
rs754929122 Chr.22: 49776005-49776005 on Build GRCh38
rs540212211 Chr.22: 49776118-49776118 on Build GRCh38
rs1022552261 Chr.22: 49776250-49776250 on Build GRCh38
rs369094496 Chr.22: 49776174-49776174 on Build GRCh38
rs375610161 Chr.22: 49776155-49776155 on Build GRCh38
rs147405126 Chr.22: 49776154-49776154 on Build GRCh38
rs369744530 Chr.22: 49776223-49776224 on Build GRCh38
rs375338194 Chr.22: 49776153-49776153 on Build GRCh38
rs199507783 Chr.22: 49776074-49776074 on Build GRCh38
rs766143354 Chr.22: 49776038-49776038 on Build GRCh38
rs112920808 Chr.22: 49776161-49776161 on Build GRCh38
rs1043295733 Chr.22: 49775969-49775969 on Build GRCh38
rs753345791 Chr.22: 49776170-49776171 on Build GRCh38
rs775569396 Chr.22: 49776142-49776142 on Build GRCh38
rs760738607 Chr.22: 49776211-49776211 on Build GRCh38
rs778121796 Chr.22: 49776377-49776377 on Build GRCh38
rs116440669 Chr.22: 49776028-49776028 on Build GRCh38
rs567334938 Chr.22: 49776243-49776243 on Build GRCh38
rs779189233 Chr.22: 49776073-49776073 on Build GRCh38
rs28539128 Chr.22: 49775937-49775937 on Build GRCh38
rs748290402 Chr.22: 49776191-49776191 on Build GRCh38
rs775771031 Chr.22: 49776341-49776341 on Build GRCh38
rs951241121 Chr.22: 49776030-49776030 on Build GRCh38
rs779692356 Chr.22: 49776171-49776187 on Build GRCh38
rs927688332 Chr.22: 49775962-49775962 on Build GRCh38
rs547261276 Chr.22: 49776179-49776179 on Build GRCh38
rs368330772 Chr.22: 49776163-49776163 on Build GRCh38
rs374693374 Chr.22: 49776042-49776042 on Build GRCh38
rs779353094 Chr.22: 49776216-49776216 on Build GRCh38
rs552065415 Chr.22: 49775980-49775980 on Build GRCh38
rs556178692 Chr.22: 49776253-49776253 on Build GRCh38
rs372337128 Chr.22: 49776164-49776164 on Build GRCh38
rs747595462 Chr.22: 49776390-49776390 on Build GRCh38
rs752219373 Chr.22: 49776019-49776019 on Build GRCh38
rs758641842 Chr.22: 49776110-49776110 on Build GRCh38
rs562228438 Chr.22: 49775951-49775951 on Build GRCh38
rs1022823989 Chr.22: 49776298-49776298 on Build GRCh38
rs781095199 Chr.22: 49776217-49776217 on Build GRCh38
rs1056886028 Chr.22: 49776371-49776371 on Build GRCh38
rs371900886 Chr.22: 49776065-49776065 on Build GRCh38
rs965525138 Chr.22: 49776386-49776386 on Build GRCh38
rs747571605 Chr.22: 49776188-49776188 on Build GRCh38
rs755737350 Chr.22: 49776168-49776168 on Build GRCh38
rs528021579 Chr.22: 49776231-49776231 on Build GRCh38
rs369554868 Chr.22: 49776210-49776210 on Build GRCh38
rs975774694 Chr.22: 49776267-49776267 on Build GRCh38
rs939854799 Chr.22: 49776255-49776255 on Build GRCh38
rs550998465 Chr.22: 49775966-49775966 on Build GRCh38
rs768789408 Chr.22: 49776116-49776116 on Build GRCh38
rs1019830526 Chr.22: 49776378-49776378 on Build GRCh38
rs756916253 Chr.22: 49776068-49776068 on Build GRCh38
rs751515699 Chr.22: 49776003-49776003 on Build GRCh38
rs779325162 Chr.22: 49776029-49776029 on Build GRCh38
rs1003409823 Chr.22: 49776171-49776171 on Build GRCh38
rs914968358 Chr.22: 49776199-49776199 on Build GRCh38
rs148224073 Chr.22: 49776114-49776114 on Build GRCh38
rs1023608166 Chr.22: 49776169-49776169 on Build GRCh38
rs925560706 Chr.22: 49775981-49775981 on Build GRCh38
rs755641784 Chr.22: 49776022-49776022 on Build GRCh38
rs912786524 Chr.22: 49776146-49776146 on Build GRCh38
rs374998959 Chr.22: 49775950-49775950 on Build GRCh38
rs770025608 Chr.22: 49776121-49776121 on Build GRCh38
rs899550803 Chr.22: 49776021-49776021 on Build GRCh38
rs1022244572 Chr.22: 49776395-49776395 on Build GRCh38
rs956849679 Chr.22: 49776236-49776236 on Build GRCh38
rs944312424 Chr.22: 49776223-49776223 on Build GRCh38
rs200471149 Chr.22: 49776035-49776035 on Build GRCh38
rs754486993 Chr.22: 49776188-49776204 on Build GRCh38
rs910048192 Chr.22: 49776281-49776281 on Build GRCh38
rs529036986 Chr.22: 49775927-49775927 on Build GRCh38
rs750194951 Chr.22: 49776031-49776033 on Build GRCh38
rs375217643 Chr.22: 49776170-49776170 on Build GRCh38
rs9628129 Chr.22: 49776122-49776122 on Build GRCh38
rs549315198 Chr.22: 49776242-49776242 on Build GRCh38
rs930144915 Chr.22: 49776216-49776217 on Build GRCh38
rs113561503 Chr.22: 49775947-49775948 on Build GRCh38
rs905317490 Chr.22: 49776349-49776349 on Build GRCh38
rs114298169 Chr.22: 49776203-49776203 on Build GRCh38

View Full Product Details