Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 49776925-49777421 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD1
Entrez Gene ID
23774
Gene Name
bromodomain containing 1
Gene Aliases
BRL, BRPF1, BRPF2
UniGene
Hs.127950
Gene Chromosome Location
Chr.22: 49773278-49827927 on Build GRCh38
Transcripts
NM_001304808.1 NM_001304809.1 XM_011530073.1 XM_017028714.1 XM_017028715.1 XM_017028716.1 XM_017028717.1 XM_017028718.1 XM_017028722.1 XM_017028723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6248572 c.1901C>A p.A634E Chr.22: 49777039-49777039 on Build GRCh38
6612617 c.2712G>A p.A904A Chr.22: 49777050-49777050 on Build GRCh38
726784 c.2610G>T p.A870A Chr.22: 49777152-49777152 on Build GRCh38
1246213 c.2675T>G p.L892R Chr.22: 49777087-49777087 on Build GRCh38
5133565 c.2666G>A p.R889H Chr.22: 49777096-49777096 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs199924628 Chr.22: 49777166-49777166 on Build GRCh38
rs865872122 Chr.22: 49777081-49777081 on Build GRCh38
rs933255083 Chr.22: 49777110-49777110 on Build GRCh38
rs369960913 Chr.22: 49777040-49777040 on Build GRCh38
rs976021624 Chr.22: 49777261-49777261 on Build GRCh38
rs773993738 Chr.22: 49777138-49777138 on Build GRCh38
rs962293055 Chr.22: 49777197-49777209 on Build GRCh38
rs918777334 Chr.22: 49777303-49777303 on Build GRCh38
rs757367533 Chr.22: 49777096-49777096 on Build GRCh38
rs902778467 Chr.22: 49777323-49777323 on Build GRCh38
rs892797481 Chr.22: 49777384-49777384 on Build GRCh38
rs752975881 Chr.22: 49777147-49777147 on Build GRCh38
rs927019569 Chr.22: 49777285-49777285 on Build GRCh38
rs769405191 Chr.22: 49777048-49777048 on Build GRCh38
rs148324843 Chr.22: 49777026-49777026 on Build GRCh38
rs751890834 Chr.22: 49777001-49777001 on Build GRCh38
rs563026361 Chr.22: 49777051-49777051 on Build GRCh38
rs1030340199 Chr.22: 49777246-49777246 on Build GRCh38
rs778234527 Chr.22: 49777192-49777192 on Build GRCh38
rs374923338 Chr.22: 49776949-49776949 on Build GRCh38
rs200866976 Chr.22: 49777027-49777027 on Build GRCh38
rs772515049 Chr.22: 49777188-49777189 on Build GRCh38
rs753084109 Chr.22: 49777018-49777018 on Build GRCh38
rs772572948 Chr.22: 49777050-49777050 on Build GRCh38
rs547589314 Chr.22: 49777231-49777231 on Build GRCh38
rs895475170 Chr.22: 49777301-49777302 on Build GRCh38
rs909881396 Chr.22: 49777288-49777288 on Build GRCh38
rs767704331 Chr.22: 49777072-49777072 on Build GRCh38
rs759903439 Chr.22: 49777066-49777066 on Build GRCh38
rs796873795 Chr.22: 49777219-49777219 on Build GRCh38
rs755295280 Chr.22: 49777002-49777002 on Build GRCh38
rs532598242 Chr.22: 49777226-49777226 on Build GRCh38
rs756220263 Chr.22: 49777020-49777020 on Build GRCh38
rs748683722 Chr.22: 49777095-49777096 on Build GRCh38
rs941540635 Chr.22: 49777161-49777161 on Build GRCh38
rs5770691 Chr.22: 49777283-49777283 on Build GRCh38
rs936978360 Chr.22: 49777156-49777156 on Build GRCh38
rs201858309 Chr.22: 49777198-49777198 on Build GRCh38
rs950195075 Chr.22: 49777307-49777307 on Build GRCh38
rs758031660 Chr.22: 49776999-49776999 on Build GRCh38
rs1009987252 Chr.22: 49777241-49777241 on Build GRCh38
rs764535212 Chr.22: 49777153-49777153 on Build GRCh38
rs141692212 Chr.22: 49777222-49777222 on Build GRCh38
rs556155486 Chr.22: 49777302-49777302 on Build GRCh38
rs1037397001 Chr.22: 49777151-49777151 on Build GRCh38
rs374731824 Chr.22: 49776996-49776996 on Build GRCh38
rs749152687 Chr.22: 49777201-49777201 on Build GRCh38
rs566630194 Chr.22: 49777242-49777242 on Build GRCh38
rs996257380 Chr.22: 49777376-49777376 on Build GRCh38
rs527458938 Chr.22: 49777260-49777260 on Build GRCh38
rs143719671 Chr.22: 49777086-49777086 on Build GRCh38
rs1006110444 Chr.22: 49776967-49776967 on Build GRCh38
rs571172880 Chr.22: 49777320-49777320 on Build GRCh38
rs1030874528 Chr.22: 49777377-49777377 on Build GRCh38
rs954040088 Chr.22: 49777270-49777270 on Build GRCh38
rs778119313 Chr.22: 49777193-49777193 on Build GRCh38
rs375440173 Chr.22: 49777074-49777074 on Build GRCh38
rs762473117 Chr.22: 49777117-49777117 on Build GRCh38
rs1045973896 Chr.22: 49777318-49777318 on Build GRCh38
rs373375061 Chr.22: 49777023-49777023 on Build GRCh38
rs747925119 Chr.22: 49777196-49777196 on Build GRCh38
rs781677385 Chr.22: 49777010-49777010 on Build GRCh38
rs531920289 Chr.22: 49777190-49777190 on Build GRCh38
rs751163081 Chr.22: 49777062-49777062 on Build GRCh38
rs538702171 Chr.22: 49777337-49777337 on Build GRCh38
rs780837839 Chr.22: 49777108-49777108 on Build GRCh38
rs1037645446 Chr.22: 49777163-49777163 on Build GRCh38
rs377610345 Chr.22: 49777210-49777210 on Build GRCh38
rs957311473 Chr.22: 49777232-49777232 on Build GRCh38
rs61449794 Chr.22: 49777279-49777279 on Build GRCh38
rs921823503 Chr.22: 49777263-49777263 on Build GRCh38
rs768514768 Chr.22: 49777039-49777039 on Build GRCh38
rs965856801 Chr.22: 49777258-49777258 on Build GRCh38
rs768641443 Chr.22: 49777102-49777102 on Build GRCh38
rs941898991 Chr.22: 49777139-49777139 on Build GRCh38
rs764586441 Chr.22: 49776997-49776997 on Build GRCh38
rs1001580973 Chr.22: 49777225-49777225 on Build GRCh38
rs779291883 Chr.22: 49777097-49777097 on Build GRCh38
rs764162132 Chr.22: 49776984-49776984 on Build GRCh38
rs114617569 Chr.22: 49777298-49777298 on Build GRCh38
rs769586081 Chr.22: 49777115-49777115 on Build GRCh38
rs201150290 Chr.22: 49776985-49776985 on Build GRCh38
rs4469 Chr.22: 49777289-49777289 on Build GRCh38
rs201557074 Chr.22: 49777276-49777277 on Build GRCh38
rs759181892 Chr.22: 49777142-49777142 on Build GRCh38
rs1015855318 Chr.22: 49776973-49776973 on Build GRCh38
rs376099852 Chr.22: 49777152-49777152 on Build GRCh38
rs779435647 Chr.22: 49776992-49776994 on Build GRCh38
rs777707723 Chr.22: 49777022-49777022 on Build GRCh38
rs147065222 Chr.22: 49777381-49777381 on Build GRCh38
rs755888959 Chr.22: 49777197-49777197 on Build GRCh38
rs138154621 Chr.22: 49777035-49777035 on Build GRCh38
rs781079239 Chr.22: 49777191-49777191 on Build GRCh38
rs1001011321 Chr.22: 49777335-49777335 on Build GRCh38
rs750732841 Chr.22: 49777175-49777175 on Build GRCh38
rs923214355 Chr.22: 49776952-49776952 on Build GRCh38
rs370621351 Chr.22: 49777059-49777059 on Build GRCh38
rs572576752 Chr.22: 49777114-49777114 on Build GRCh38
rs888644234 Chr.22: 49776960-49776960 on Build GRCh38
rs954910705 Chr.22: 49777249-49777249 on Build GRCh38
rs377653828 Chr.22: 49777041-49777041 on Build GRCh38
rs964809504 Chr.22: 49777237-49777237 on Build GRCh38
rs756562359 Chr.22: 49777076-49777076 on Build GRCh38
rs36005798 Chr.22: 49776949-49776950 on Build GRCh38
rs1049855559 Chr.22: 49777107-49777107 on Build GRCh38
rs56234215 Chr.22: 49776970-49776970 on Build GRCh38
rs754342911 Chr.22: 49777165-49777165 on Build GRCh38
rs907695621 Chr.22: 49777128-49777128 on Build GRCh38
rs542842150 Chr.22: 49777145-49777145 on Build GRCh38
rs747305183 Chr.22: 49777192-49777193 on Build GRCh38
rs114483505 Chr.22: 49777095-49777095 on Build GRCh38
rs771122476 Chr.22: 49777194-49777195 on Build GRCh38
rs369232838 Chr.22: 49776998-49776998 on Build GRCh38
rs574018747 Chr.22: 49776958-49776958 on Build GRCh38

View Full Product Details