Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10280669-10281164 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRLOS
Entrez Gene ID
100126793
Gene Name
ghrelin opposite strand/antisense RNA
Gene Aliases
GHRL-AS1, GHRLAS, NCRNA00068
UniGene
Hs.475401
Gene Chromosome Location
Chr.3: 10280952-10293449 on Build GRCh38
Transcripts
NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1 NM_014760.3

Gene Symbol
TATDN2
Entrez Gene ID
9797
Gene Name
TatD DNase domain containing 2
Gene Aliases
-
UniGene
Hs.475401
Gene Chromosome Location
Chr.3: 10248493-10281222 on Build GRCh38
Transcripts
NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1 NM_014760.3

SNP Detail

SNP ID Chromosome Location
rs952904824 Chr.3: 10280970-10280970 on Build GRCh38
rs891130859 Chr.3: 10280810-10280810 on Build GRCh38
rs922941408 Chr.3: 10280830-10280830 on Build GRCh38
rs1011857637 Chr.3: 10280842-10280842 on Build GRCh38
rs920083664 Chr.3: 10280740-10280740 on Build GRCh38
rs35541467 Chr.3: 10280732-10280732 on Build GRCh38
rs991078697 Chr.3: 10280876-10280876 on Build GRCh38
rs368694371 Chr.3: 10281006-10281006 on Build GRCh38
rs2072578 Chr.3: 10280837-10280837 on Build GRCh38
rs555251224 Chr.3: 10280869-10280869 on Build GRCh38
rs533884318 Chr.3: 10280936-10280936 on Build GRCh38
rs1011817862 Chr.3: 10281008-10281008 on Build GRCh38
rs891183241 Chr.3: 10280951-10280951 on Build GRCh38
rs994412594 Chr.3: 10280946-10280946 on Build GRCh38
rs74710207 Chr.3: 10281079-10281079 on Build GRCh38
rs928290405 Chr.3: 10280693-10280693 on Build GRCh38
rs772280433 Chr.3: 10280783-10280783 on Build GRCh38
rs1044173683 Chr.3: 10281115-10281115 on Build GRCh38
rs967560015 Chr.3: 10280839-10280839 on Build GRCh38
rs914513294 Chr.3: 10281069-10281069 on Build GRCh38
rs928341307 Chr.3: 10280853-10280853 on Build GRCh38
rs79228813 Chr.3: 10280721-10280721 on Build GRCh38
rs973409391 Chr.3: 10280722-10280722 on Build GRCh38
rs967567794 Chr.3: 10281026-10281026 on Build GRCh38
rs138861902 Chr.3: 10280783-10280783 on Build GRCh38
rs898601574 Chr.3: 10280778-10280778 on Build GRCh38
rs376779818 Chr.3: 10280873-10280873 on Build GRCh38
rs188447194 Chr.3: 10280729-10280729 on Build GRCh38
rs748048432 Chr.3: 10281062-10281062 on Build GRCh38
rs988735060 Chr.3: 10281039-10281039 on Build GRCh38
rs986073992 Chr.3: 10280979-10280979 on Build GRCh38
rs1022243249 Chr.3: 10281012-10281012 on Build GRCh38
rs915840458 Chr.3: 10280877-10280877 on Build GRCh38
rs752303451 Chr.3: 10280956-10280956 on Build GRCh38
rs1037509816 Chr.3: 10280768-10280768 on Build GRCh38
rs934347910 Chr.3: 10281035-10281035 on Build GRCh38
rs922911925 Chr.3: 10281021-10281021 on Build GRCh38
rs1008193951 Chr.3: 10280824-10280824 on Build GRCh38
rs1021776746 Chr.3: 10280838-10280838 on Build GRCh38
rs1047146338 Chr.3: 10280945-10280945 on Build GRCh38
rs1039704440 Chr.3: 10280980-10280980 on Build GRCh38
rs943015574 Chr.3: 10280777-10280777 on Build GRCh38
rs977189652 Chr.3: 10280846-10280846 on Build GRCh38
rs964424660 Chr.3: 10280983-10280983 on Build GRCh38
rs369098956 Chr.3: 10280883-10280883 on Build GRCh38
rs975846520 Chr.3: 10281002-10281002 on Build GRCh38
rs910339612 Chr.3: 10280777-10280777 on Build GRCh38
rs77548531 Chr.3: 10280714-10280714 on Build GRCh38
rs1008244868 Chr.3: 10280964-10280964 on Build GRCh38
rs34349495 Chr.3: 10280776-10280777 on Build GRCh38
rs1037970048 Chr.3: 10280903-10280903 on Build GRCh38
rs919090890 Chr.3: 10280907-10280907 on Build GRCh38
rs754865633 Chr.3: 10280776-10280776 on Build GRCh38
rs184467361 Chr.3: 10280704-10280704 on Build GRCh38
rs1018546173 Chr.3: 10280825-10280825 on Build GRCh38
rs546547772 Chr.3: 10280713-10280713 on Build GRCh38
rs35217039 Chr.3: 10280820-10280820 on Build GRCh38
rs964260894 Chr.3: 10280833-10280833 on Build GRCh38
rs1047542170 Chr.3: 10280800-10280800 on Build GRCh38
rs1002920638 Chr.3: 10280902-10280902 on Build GRCh38
rs900108401 Chr.3: 10280990-10280990 on Build GRCh38
rs536820578 Chr.3: 10280855-10280855 on Build GRCh38
rs1044457161 Chr.3: 10280870-10280870 on Build GRCh38
rs558222360 Chr.3: 10280976-10280976 on Build GRCh38

View Full Product Details