Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10285633-10286174 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRL
Entrez Gene ID
51738
Gene Name
ghrelin/obestatin prepropeptide
Gene Aliases
MTLRP
UniGene
Hs.590080
Gene Chromosome Location
Chr.3: 10285750-10292947 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1 NR_026829.1

Gene Symbol
GHRLOS
Entrez Gene ID
100126793
Gene Name
ghrelin opposite strand/antisense RNA
Gene Aliases
GHRL-AS1, GHRLAS, NCRNA00068
UniGene
Hs.475401
Gene Chromosome Location
Chr.3: 10280952-10293449 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1 NR_026829.1

Gene Symbol
LINC00852
Entrez Gene ID
84657
Gene Name
long intergenic non-protein coding RNA 852
Gene Aliases
C3orf42, GHRL-AS2, GHRLOS2, NAG73
UniGene
Hs.302131
Gene Chromosome Location
Chr.3: 10284419-10285746 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 XM_017006612.1 XM_017006613.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1 NR_026829.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4686921 c.344C>T p.A115V Chr.3: 10285885-10285885 on Build GRCh38
4686922 c.308C>T p.A103V Chr.3: 10285885-10285885 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs755187335 Chr.3: 10285893-10285893 on Build GRCh38
rs1023136475 Chr.3: 10285928-10285928 on Build GRCh38
rs369305953 Chr.3: 10285872-10285872 on Build GRCh38
rs767567218 Chr.3: 10285904-10285904 on Build GRCh38
rs757622562 Chr.3: 10285931-10285931 on Build GRCh38
rs898776092 Chr.3: 10286046-10286046 on Build GRCh38
rs551236832 Chr.3: 10285788-10285788 on Build GRCh38
rs180790269 Chr.3: 10285934-10285934 on Build GRCh38
rs1021240135 Chr.3: 10286150-10286150 on Build GRCh38
rs778484168 Chr.3: 10285677-10285677 on Build GRCh38
rs747672256 Chr.3: 10285770-10285770 on Build GRCh38
rs964117942 Chr.3: 10286131-10286131 on Build GRCh38
rs994420013 Chr.3: 10286066-10286066 on Build GRCh38
rs1025803523 Chr.3: 10285742-10285742 on Build GRCh38
rs945955425 Chr.3: 10285864-10285864 on Build GRCh38
rs957644645 Chr.3: 10285657-10285657 on Build GRCh38
rs761669571 Chr.3: 10285867-10285867 on Build GRCh38
rs766291918 Chr.3: 10286060-10286060 on Build GRCh38
rs756695811 Chr.3: 10285830-10285830 on Build GRCh38
rs970334878 Chr.3: 10285955-10285955 on Build GRCh38
rs1035850572 Chr.3: 10286000-10286000 on Build GRCh38
rs1012625361 Chr.3: 10285714-10285714 on Build GRCh38
rs558796070 Chr.3: 10286020-10286020 on Build GRCh38
rs760722097 Chr.3: 10286094-10286094 on Build GRCh38
rs768160177 Chr.3: 10285858-10285858 on Build GRCh38
rs772952663 Chr.3: 10286090-10286090 on Build GRCh38
rs534605934 Chr.3: 10285989-10285989 on Build GRCh38
rs758555486 Chr.3: 10285804-10285804 on Build GRCh38
rs1056905525 Chr.3: 10285686-10285686 on Build GRCh38
rs750221156 Chr.3: 10285794-10285794 on Build GRCh38
rs768193741 Chr.3: 10285941-10285941 on Build GRCh38
rs562343232 Chr.3: 10286085-10286085 on Build GRCh38
rs775181399 Chr.3: 10285844-10285844 on Build GRCh38
rs891303302 Chr.3: 10286093-10286093 on Build GRCh38
rs910498177 Chr.3: 10286061-10286061 on Build GRCh38
rs986570963 Chr.3: 10285805-10285805 on Build GRCh38
rs770796569 Chr.3: 10286104-10286104 on Build GRCh38
rs773190477 Chr.3: 10285879-10285879 on Build GRCh38
rs116755162 Chr.3: 10286083-10286083 on Build GRCh38
rs568932159 Chr.3: 10285828-10285828 on Build GRCh38
rs781154531 Chr.3: 10285827-10285827 on Build GRCh38
rs370534599 Chr.3: 10285930-10285930 on Build GRCh38
rs970599303 Chr.3: 10285694-10285694 on Build GRCh38
rs776146116 Chr.3: 10285863-10285863 on Build GRCh38
rs1023105281 Chr.3: 10285679-10285679 on Build GRCh38
rs930261410 Chr.3: 10286035-10286035 on Build GRCh38
rs930565885 Chr.3: 10285793-10285793 on Build GRCh38
rs201295883 Chr.3: 10285833-10285833 on Build GRCh38
rs771269632 Chr.3: 10285838-10285838 on Build GRCh38
rs751764543 Chr.3: 10286021-10286022 on Build GRCh38
rs780619940 Chr.3: 10285943-10285943 on Build GRCh38
rs1017210228 Chr.3: 10285719-10285719 on Build GRCh38
rs1011573636 Chr.3: 10286145-10286145 on Build GRCh38
rs980551343 Chr.3: 10285697-10285697 on Build GRCh38
rs914515584 Chr.3: 10285847-10285847 on Build GRCh38
rs1041515028 Chr.3: 10285659-10285659 on Build GRCh38
rs778423066 Chr.3: 10285920-10285920 on Build GRCh38
rs756321970 Chr.3: 10285919-10285919 on Build GRCh38
rs934761909 Chr.3: 10286105-10286105 on Build GRCh38
rs1011794325 Chr.3: 10285924-10285924 on Build GRCh38
rs994361172 Chr.3: 10285731-10285731 on Build GRCh38
rs542727187 Chr.3: 10286148-10286148 on Build GRCh38
rs765070591 Chr.3: 10285893-10285894 on Build GRCh38
rs766608356 Chr.3: 10285806-10285806 on Build GRCh38
rs898725190 Chr.3: 10285728-10285728 on Build GRCh38
rs901657546 Chr.3: 10286097-10286097 on Build GRCh38
rs867308216 Chr.3: 10285853-10285853 on Build GRCh38
rs1003621141 Chr.3: 10285681-10285681 on Build GRCh38
rs368192363 Chr.3: 10285817-10285817 on Build GRCh38
rs114837813 Chr.3: 10286027-10286027 on Build GRCh38
rs868519650 Chr.3: 10285739-10285739 on Build GRCh38
rs369580145 Chr.3: 10285885-10285885 on Build GRCh38
rs768341252 Chr.3: 10285660-10285660 on Build GRCh38
rs982052946 Chr.3: 10285993-10285993 on Build GRCh38
rs989805061 Chr.3: 10286141-10286141 on Build GRCh38
rs75829178 Chr.3: 10286015-10286015 on Build GRCh38
rs878990654 Chr.3: 10285859-10285859 on Build GRCh38
rs377149451 Chr.3: 10285884-10285884 on Build GRCh38
rs575305549 Chr.3: 10286107-10286107 on Build GRCh38
rs939108476 Chr.3: 10285722-10285722 on Build GRCh38
rs375772841 Chr.3: 10285849-10285849 on Build GRCh38
rs895190148 Chr.3: 10286007-10286007 on Build GRCh38
rs374584866 Chr.3: 10285933-10285933 on Build GRCh38
rs895148586 Chr.3: 10285701-10285701 on Build GRCh38
rs751795811 Chr.3: 10285807-10285807 on Build GRCh38
rs757282700 Chr.3: 10285846-10285849 on Build GRCh38
rs779171029 Chr.3: 10285932-10285932 on Build GRCh38
rs267599519 Chr.3: 10285883-10285883 on Build GRCh38
rs373274141 Chr.3: 10285910-10285910 on Build GRCh38

View Full Product Details