Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10292789-10293296 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GHRL
Entrez Gene ID
51738
Gene Name
ghrelin/obestatin prepropeptide
Gene Aliases
MTLRP
UniGene
Hs.590080
Gene Chromosome Location
Chr.3: 10285750-10292947 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1

Gene Symbol
GHRLOS
Entrez Gene ID
100126793
Gene Name
ghrelin opposite strand/antisense RNA
Gene Aliases
GHRL-AS1, GHRLAS, NCRNA00068
UniGene
Hs.475401
Gene Chromosome Location
Chr.3: 10280952-10293449 on Build GRCh38
Transcripts
NM_001134941.2 NM_001134944.1 NM_001134945.1 NM_001134946.1 NM_001302821.1 NM_001302822.1 NM_001302823.1 NM_001302824.1 NM_001302825.1 NM_016362.4 NR_126505.1 NR_004431.3 NR_024144.2 NR_024145.2 NR_073566.1 NR_073567.1 NR_073568.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3682915 c.72+10G>A p.? Chr.3: 10292832-10292832 on Build GRCh38
6286150 c.35C>A p.T12K Chr.3: 10292879-10292879 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs940838247 Chr.3: 10293237-10293237 on Build GRCh38
rs568629288 Chr.3: 10293095-10293095 on Build GRCh38
rs532575006 Chr.3: 10292954-10292955 on Build GRCh38
rs997120377 Chr.3: 10292984-10292984 on Build GRCh38
rs35631599 Chr.3: 10292959-10292960 on Build GRCh38
rs1012612975 Chr.3: 10293171-10293171 on Build GRCh38
rs796243265 Chr.3: 10293012-10293012 on Build GRCh38
rs745550678 Chr.3: 10293008-10293008 on Build GRCh38
rs375419558 Chr.3: 10293030-10293030 on Build GRCh38
rs556533493 Chr.3: 10293078-10293078 on Build GRCh38
rs552686864 Chr.3: 10292916-10292916 on Build GRCh38
rs796821661 Chr.3: 10292848-10292848 on Build GRCh38
rs570995847 Chr.3: 10292952-10292952 on Build GRCh38
rs1050090064 Chr.3: 10293252-10293252 on Build GRCh38
rs547240019 Chr.3: 10292843-10292843 on Build GRCh38
rs771520830 Chr.3: 10292954-10292972 on Build GRCh38
rs1015433155 Chr.3: 10293202-10293202 on Build GRCh38
rs781763564 Chr.3: 10292857-10292857 on Build GRCh38
rs982121553 Chr.3: 10292831-10292831 on Build GRCh38
rs149862859 Chr.3: 10292863-10292863 on Build GRCh38
rs921343582 Chr.3: 10293034-10293034 on Build GRCh38
rs956490688 Chr.3: 10293082-10293082 on Build GRCh38
rs1010339812 Chr.3: 10293087-10293087 on Build GRCh38
rs1000796435 Chr.3: 10293265-10293265 on Build GRCh38
rs1004505920 Chr.3: 10293190-10293190 on Build GRCh38
rs369032553 Chr.3: 10293043-10293043 on Build GRCh38
rs755167665 Chr.3: 10292847-10292847 on Build GRCh38
rs1032686590 Chr.3: 10292842-10292842 on Build GRCh38
rs891399969 Chr.3: 10293077-10293077 on Build GRCh38
rs971703514 Chr.3: 10292888-10292888 on Build GRCh38
rs1001143811 Chr.3: 10293047-10293047 on Build GRCh38
rs538238090 Chr.3: 10293048-10293048 on Build GRCh38
rs373029338 Chr.3: 10293076-10293076 on Build GRCh38
rs1050800553 Chr.3: 10293019-10293019 on Build GRCh38
rs903985113 Chr.3: 10293125-10293125 on Build GRCh38
rs1024368880 Chr.3: 10293172-10293172 on Build GRCh38
rs985409239 Chr.3: 10293185-10293185 on Build GRCh38
rs933019770 Chr.3: 10293249-10293249 on Build GRCh38
rs142421973 Chr.3: 10292983-10292983 on Build GRCh38
rs971414207 Chr.3: 10293111-10293111 on Build GRCh38
rs17032631 Chr.3: 10292832-10292832 on Build GRCh38
rs962555417 Chr.3: 10293214-10293214 on Build GRCh38
rs1036041463 Chr.3: 10292914-10292914 on Build GRCh38
rs756497691 Chr.3: 10292868-10292868 on Build GRCh38
rs958984453 Chr.3: 10293149-10293149 on Build GRCh38
rs901534306 Chr.3: 10292958-10292958 on Build GRCh38
rs1042995243 Chr.3: 10293271-10293271 on Build GRCh38
rs1054240947 Chr.3: 10293145-10293145 on Build GRCh38
rs766418114 Chr.3: 10292837-10292837 on Build GRCh38
rs973868124 Chr.3: 10293219-10293219 on Build GRCh38
rs769243158 Chr.3: 10293144-10293144 on Build GRCh38
rs62240120 Chr.3: 10293268-10293268 on Build GRCh38
rs376062621 Chr.3: 10293148-10293148 on Build GRCh38
rs777010012 Chr.3: 10293247-10293247 on Build GRCh38
rs750613439 Chr.3: 10292818-10292818 on Build GRCh38
rs937535881 Chr.3: 10292836-10292836 on Build GRCh38
rs1036094288 Chr.3: 10293238-10293238 on Build GRCh38
rs139684563 Chr.3: 10292862-10292862 on Build GRCh38
rs909397002 Chr.3: 10293189-10293189 on Build GRCh38
rs888737978 Chr.3: 10293256-10293256 on Build GRCh38

View Full Product Details