Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 11272202-11272694 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATG7
Entrez Gene ID
10533
Gene Name
autophagy related 7
Gene Aliases
APG7-LIKE, APG7L, GSA7
UniGene
Hs.38032
Gene Chromosome Location
Chr.3: 11272324-11564704 on Build GRCh38
Transcripts
NM_001136031.2 NM_001144912.1 NM_006395.2 XM_006712931.3 XM_006712932.3 XM_006712933.3 XM_011533277.2 XM_011533278.2 XM_011533279.2 XM_011533280.2 XM_011533281.2 XM_011533282.2 XM_011533283.2 XM_011533284.2 XM_011533285.2 XM_011533286.2 XM_017005543.1 XM_017005544.1 XM_017005546.1 XM_017005547.1 XM_017005548.1 XM_017005549.1 XM_017005550.1 XM_017005551.1 XM_017005552.1

SNP Detail

SNP ID Chromosome Location
rs549513942 Chr.3: 11272393-11272393 on Build GRCh38
rs1054243069 Chr.3: 11272420-11272420 on Build GRCh38
rs2606729 Chr.3: 11272355-11272355 on Build GRCh38
rs925516353 Chr.3: 11272667-11272667 on Build GRCh38
rs910847700 Chr.3: 11272415-11272415 on Build GRCh38
rs760729146 Chr.3: 11272615-11272615 on Build GRCh38
rs773211172 Chr.3: 11272567-11272567 on Build GRCh38
rs980324962 Chr.3: 11272456-11272456 on Build GRCh38
rs377489365 Chr.3: 11272527-11272527 on Build GRCh38
rs769037413 Chr.3: 11272429-11272429 on Build GRCh38
rs915003305 Chr.3: 11272283-11272283 on Build GRCh38
rs1049409353 Chr.3: 11272275-11272275 on Build GRCh38
rs1027059797 Chr.3: 11272398-11272398 on Build GRCh38
rs2122031 Chr.3: 11272457-11272457 on Build GRCh38
rs748964110 Chr.3: 11272366-11272366 on Build GRCh38
rs1030480171 Chr.3: 11272631-11272631 on Build GRCh38
rs935755199 Chr.3: 11272417-11272417 on Build GRCh38
rs1054696611 Chr.3: 11272295-11272295 on Build GRCh38
rs947797589 Chr.3: 11272539-11272539 on Build GRCh38
rs906409361 Chr.3: 11272317-11272317 on Build GRCh38
rs976260788 Chr.3: 11272629-11272629 on Build GRCh38
rs933667764 Chr.3: 11272253-11272253 on Build GRCh38
rs986345863 Chr.3: 11272637-11272637 on Build GRCh38
rs2594971 Chr.3: 11272224-11272224 on Build GRCh38
rs889552099 Chr.3: 11272276-11272276 on Build GRCh38
rs947884094 Chr.3: 11272301-11272301 on Build GRCh38
rs927778822 Chr.3: 11272621-11272621 on Build GRCh38
rs1034317507 Chr.3: 11272513-11272513 on Build GRCh38
rs181919321 Chr.3: 11272534-11272534 on Build GRCh38
rs962275862 Chr.3: 11272362-11272362 on Build GRCh38
rs534538108 Chr.3: 11272500-11272500 on Build GRCh38
rs939256185 Chr.3: 11272322-11272322 on Build GRCh38
rs939205455 Chr.3: 11272627-11272627 on Build GRCh38
rs985140511 Chr.3: 11272407-11272407 on Build GRCh38
rs1034286485 Chr.3: 11272342-11272342 on Build GRCh38
rs1024217201 Chr.3: 11272310-11272310 on Build GRCh38
rs536650425 Chr.3: 11272333-11272333 on Build GRCh38
rs151156601 Chr.3: 11272553-11272553 on Build GRCh38
rs116080632 Chr.3: 11272350-11272350 on Build GRCh38
rs971272351 Chr.3: 11272316-11272316 on Build GRCh38
rs560499991 Chr.3: 11272403-11272403 on Build GRCh38
rs1010475130 Chr.3: 11272309-11272309 on Build GRCh38
rs533497166 Chr.3: 11272241-11272241 on Build GRCh38
rs984854009 Chr.3: 11272351-11272351 on Build GRCh38
rs142065270 Chr.3: 11272302-11272302 on Build GRCh38
rs111373406 Chr.3: 11272454-11272454 on Build GRCh38
rs1001384215 Chr.3: 11272485-11272485 on Build GRCh38
rs567821536 Chr.3: 11272402-11272402 on Build GRCh38
rs768044852 Chr.3: 11272549-11272549 on Build GRCh38
rs932192074 Chr.3: 11272406-11272406 on Build GRCh38
rs935718238 Chr.3: 11272669-11272669 on Build GRCh38
rs773121015 Chr.3: 11272459-11272459 on Build GRCh38
rs558666304 Chr.3: 11272523-11272523 on Build GRCh38
rs560451896 Chr.3: 11272249-11272249 on Build GRCh38
rs35781004 Chr.3: 11272321-11272322 on Build GRCh38
rs999922520 Chr.3: 11272288-11272288 on Build GRCh38
rs112337127 Chr.3: 11272632-11272632 on Build GRCh38
rs549911818 Chr.3: 11272443-11272443 on Build GRCh38
rs965625049 Chr.3: 11272428-11272428 on Build GRCh38

View Full Product Details