Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 12815783-12816282 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAND2
Entrez Gene ID
23066
Gene Name
cullin associated and neddylation dissociated 2 (putative)
Gene Aliases
TIP120B, Tp120b
UniGene
Hs.343664
Gene Chromosome Location
Chr.3: 12796654-12834818 on Build GRCh38
Transcripts
NM_001162499.1 NM_012298.2 XM_011533503.2 XM_011533504.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4463796 c.1306C>T p.L436F Chr.3: 12815873-12815873 on Build GRCh38
6289636 c.1117G>A p.V373I Chr.3: 12815963-12815963 on Build GRCh38
6619671 c.1069G>A p.V357I Chr.3: 12815915-12815915 on Build GRCh38
4888579 c.1044G>T p.L348L Chr.3: 12815890-12815890 on Build GRCh38
7188739 c.1343G>A p.R448Q Chr.3: 12815910-12815910 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs763436605 Chr.3: 12815951-12815951 on Build GRCh38
rs769130360 Chr.3: 12815927-12815927 on Build GRCh38
rs201739910 Chr.3: 12815909-12815909 on Build GRCh38
rs139267616 Chr.3: 12815996-12815996 on Build GRCh38
rs780677712 Chr.3: 12815850-12815850 on Build GRCh38
rs765758141 Chr.3: 12815857-12815857 on Build GRCh38
rs762233872 Chr.3: 12815856-12815856 on Build GRCh38
rs200578477 Chr.3: 12815914-12815914 on Build GRCh38
rs978629639 Chr.3: 12815903-12815903 on Build GRCh38
rs978577268 Chr.3: 12816122-12816122 on Build GRCh38
rs987440175 Chr.3: 12816216-12816216 on Build GRCh38
rs544123329 Chr.3: 12815912-12815912 on Build GRCh38
rs201388614 Chr.3: 12815828-12815828 on Build GRCh38
rs772284886 Chr.3: 12815835-12815835 on Build GRCh38
rs899315122 Chr.3: 12815977-12815977 on Build GRCh38
rs373749682 Chr.3: 12815915-12815915 on Build GRCh38
rs761696281 Chr.3: 12815976-12815976 on Build GRCh38
rs753774394 Chr.3: 12815894-12815894 on Build GRCh38
rs752226246 Chr.3: 12815877-12815877 on Build GRCh38
rs748612767 Chr.3: 12815924-12815924 on Build GRCh38
rs765180511 Chr.3: 12815983-12815983 on Build GRCh38
rs763451738 Chr.3: 12815964-12815964 on Build GRCh38
rs755245571 Chr.3: 12816045-12816045 on Build GRCh38
rs1035419654 Chr.3: 12815854-12815854 on Build GRCh38
rs1035769390 Chr.3: 12816001-12816001 on Build GRCh38
rs970599327 Chr.3: 12815957-12815957 on Build GRCh38
rs372525688 Chr.3: 12815895-12815895 on Build GRCh38
rs747524253 Chr.3: 12815863-12815866 on Build GRCh38
rs2305397 Chr.3: 12815994-12815994 on Build GRCh38
rs760214881 Chr.3: 12815963-12815963 on Build GRCh38
rs1022692991 Chr.3: 12816110-12816110 on Build GRCh38
rs1010747174 Chr.3: 12815892-12815892 on Build GRCh38
rs748250637 Chr.3: 12816253-12816253 on Build GRCh38
rs895514556 Chr.3: 12816079-12816079 on Build GRCh38
rs771721872 Chr.3: 12815954-12815954 on Build GRCh38
rs944495676 Chr.3: 12815852-12815852 on Build GRCh38
rs1047410378 Chr.3: 12815817-12815817 on Build GRCh38
rs764083970 Chr.3: 12815879-12815879 on Build GRCh38
rs781070638 Chr.3: 12816056-12816056 on Build GRCh38
rs930829230 Chr.3: 12815984-12815984 on Build GRCh38
rs775069513 Chr.3: 12815961-12815961 on Build GRCh38
rs768089265 Chr.3: 12816007-12816010 on Build GRCh38
rs574811386 Chr.3: 12815925-12815925 on Build GRCh38
rs769186357 Chr.3: 12815842-12815842 on Build GRCh38
rs778611983 Chr.3: 12815907-12815907 on Build GRCh38
rs773674103 Chr.3: 12815930-12815930 on Build GRCh38
rs537349056 Chr.3: 12816031-12816031 on Build GRCh38
rs960058984 Chr.3: 12815867-12815867 on Build GRCh38
rs376671004 Chr.3: 12815841-12815841 on Build GRCh38
rs1039159564 Chr.3: 12816159-12816159 on Build GRCh38
rs956159686 Chr.3: 12816203-12816203 on Build GRCh38
rs1014714490 Chr.3: 12815918-12815918 on Build GRCh38
rs1039160720 Chr.3: 12815958-12815958 on Build GRCh38
rs375894875 Chr.3: 12816051-12816052 on Build GRCh38
rs753393186 Chr.3: 12815968-12815968 on Build GRCh38
rs1031710232 Chr.3: 12815910-12815910 on Build GRCh38
rs946519054 Chr.3: 12816238-12816238 on Build GRCh38
rs1013077507 Chr.3: 12816081-12816081 on Build GRCh38
rs1031511528 Chr.3: 12816182-12816182 on Build GRCh38
rs755617284 Chr.3: 12815878-12815878 on Build GRCh38
rs571613718 Chr.3: 12815807-12815807 on Build GRCh38
rs973613632 Chr.3: 12815959-12815959 on Build GRCh38
rs968508884 Chr.3: 12816116-12816116 on Build GRCh38
rs751787749 Chr.3: 12816036-12816036 on Build GRCh38
rs921716784 Chr.3: 12816217-12816217 on Build GRCh38
rs773729238 Chr.3: 12815859-12815859 on Build GRCh38
rs775598749 Chr.3: 12815839-12815839 on Build GRCh38
rs1038865823 Chr.3: 12815875-12815875 on Build GRCh38
rs767312912 Chr.3: 12815876-12815876 on Build GRCh38
rs147449397 Chr.3: 12815986-12815986 on Build GRCh38
rs886707906 Chr.3: 12815990-12815990 on Build GRCh38
rs369200093 Chr.3: 12815870-12815870 on Build GRCh38
rs776197816 Chr.3: 12816034-12816035 on Build GRCh38

View Full Product Details