Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14897405-14897913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD5
Entrez Gene ID
152273
Gene Name
FYVE, RhoGEF and PH domain containing 5
Gene Aliases
ZFYVE23
UniGene
Hs.412406
Gene Chromosome Location
Chr.3: 14818962-14934565 on Build GRCh38
Transcripts
NM_001320276.1 NM_152536.3 XM_011533422.1 XM_011533423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5463216 c.2885A>C p.E962A Chr.3: 14897645-14897645 on Build GRCh38
4608989 c.2769G>T p.M923I Chr.3: 14897529-14897529 on Build GRCh38
3915114 c.2143C>T p.H715Y Chr.3: 14897626-14897626 on Build GRCh38
3588606 c.2026G>A p.D676N Chr.3: 14897509-14897509 on Build GRCh38
4592784 c.2821G>A p.E941K Chr.3: 14897581-14897581 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs143755810 Chr.3: 14897841-14897841 on Build GRCh38
rs2307092 Chr.3: 14897581-14897581 on Build GRCh38
rs758533529 Chr.3: 14897528-14897528 on Build GRCh38
rs771539023 Chr.3: 14897578-14897578 on Build GRCh38
rs577336192 Chr.3: 14897847-14897847 on Build GRCh38
rs529361991 Chr.3: 14897884-14897884 on Build GRCh38
rs2307091 Chr.3: 14897582-14897582 on Build GRCh38
rs756410883 Chr.3: 14897687-14897687 on Build GRCh38
rs58244401 Chr.3: 14897830-14897830 on Build GRCh38
rs533550980 Chr.3: 14897544-14897544 on Build GRCh38
rs1003850596 Chr.3: 14897833-14897833 on Build GRCh38
rs193203179 Chr.3: 14897675-14897675 on Build GRCh38
rs981325624 Chr.3: 14897540-14897540 on Build GRCh38
rs770107041 Chr.3: 14897620-14897620 on Build GRCh38
rs141080106 Chr.3: 14897471-14897471 on Build GRCh38
rs369709099 Chr.3: 14897631-14897631 on Build GRCh38
rs2117775 Chr.3: 14897691-14897691 on Build GRCh38
rs2117774 Chr.3: 14897857-14897857 on Build GRCh38
rs377720944 Chr.3: 14897725-14897725 on Build GRCh38
rs192236786 Chr.3: 14897870-14897870 on Build GRCh38
rs757988748 Chr.3: 14897692-14897692 on Build GRCh38
rs990864290 Chr.3: 14897478-14897478 on Build GRCh38
rs374685745 Chr.3: 14897799-14897799 on Build GRCh38
rs577110158 Chr.3: 14897684-14897684 on Build GRCh38
rs925309997 Chr.3: 14897683-14897683 on Build GRCh38
rs374862604 Chr.3: 14897878-14897878 on Build GRCh38
rs1026723428 Chr.3: 14897817-14897817 on Build GRCh38
rs561668642 Chr.3: 14897464-14897464 on Build GRCh38
rs1012642410 Chr.3: 14897887-14897887 on Build GRCh38
rs561636283 Chr.3: 14897730-14897730 on Build GRCh38
rs529529727 Chr.3: 14897718-14897718 on Build GRCh38
rs79598318 Chr.3: 14897751-14897751 on Build GRCh38
rs891327268 Chr.3: 14897518-14897518 on Build GRCh38
rs919109133 Chr.3: 14897704-14897704 on Build GRCh38
rs200072982 Chr.3: 14897634-14897634 on Build GRCh38
rs543332994 Chr.3: 14897461-14897461 on Build GRCh38
rs137983126 Chr.3: 14897619-14897619 on Build GRCh38
rs1001088459 Chr.3: 14897674-14897674 on Build GRCh38
rs77173419 Chr.3: 14897575-14897575 on Build GRCh38
rs768663125 Chr.3: 14897889-14897889 on Build GRCh38
rs902011576 Chr.3: 14897428-14897428 on Build GRCh38
rs763474834 Chr.3: 14897690-14897691 on Build GRCh38
rs183412379 Chr.3: 14897442-14897442 on Build GRCh38
rs748039471 Chr.3: 14897700-14897700 on Build GRCh38
rs997734734 Chr.3: 14897435-14897435 on Build GRCh38
rs766549571 Chr.3: 14897668-14897668 on Build GRCh38
rs1008419889 Chr.3: 14897524-14897524 on Build GRCh38
rs948133281 Chr.3: 14897458-14897458 on Build GRCh38
rs995230401 Chr.3: 14897580-14897580 on Build GRCh38
rs948138176 Chr.3: 14897676-14897676 on Build GRCh38
rs370629804 Chr.3: 14897569-14897569 on Build GRCh38
rs545706772 Chr.3: 14897439-14897439 on Build GRCh38
rs981657072 Chr.3: 14897703-14897703 on Build GRCh38
rs146958378 Chr.3: 14897497-14897497 on Build GRCh38
rs982693679 Chr.3: 14897591-14897591 on Build GRCh38
rs756963147 Chr.3: 14897576-14897576 on Build GRCh38
rs943008965 Chr.3: 14897786-14897786 on Build GRCh38
rs776231624 Chr.3: 14897493-14897493 on Build GRCh38
rs754746108 Chr.3: 14897515-14897515 on Build GRCh38
rs182966141 Chr.3: 14897679-14897679 on Build GRCh38
rs959422153 Chr.3: 14897473-14897473 on Build GRCh38
rs148985468 Chr.3: 14897802-14897802 on Build GRCh38
rs376541365 Chr.3: 14897479-14897479 on Build GRCh38
rs755447648 Chr.3: 14897553-14897553 on Build GRCh38
rs548127230 Chr.3: 14897494-14897494 on Build GRCh38
rs753125761 Chr.3: 14897686-14897686 on Build GRCh38
rs746484154 Chr.3: 14897716-14897716 on Build GRCh38
rs867636693 Chr.3: 14897547-14897547 on Build GRCh38
rs1001024872 Chr.3: 14897861-14897861 on Build GRCh38
rs772017511 Chr.3: 14897463-14897463 on Build GRCh38
rs767037086 Chr.3: 14897543-14897543 on Build GRCh38
rs998082206 Chr.3: 14897611-14897611 on Build GRCh38
rs970859598 Chr.3: 14897698-14897698 on Build GRCh38
rs1035287621 Chr.3: 14897835-14897835 on Build GRCh38
rs1053954194 Chr.3: 14897865-14897865 on Build GRCh38
rs776522779 Chr.3: 14897600-14897600 on Build GRCh38
rs779545692 Chr.3: 14897694-14897694 on Build GRCh38
rs763195626 Chr.3: 14897650-14897650 on Build GRCh38
rs188404853 Chr.3: 14897726-14897726 on Build GRCh38
rs539608717 Chr.3: 14897690-14897690 on Build GRCh38
rs902752482 Chr.3: 14897649-14897649 on Build GRCh38

View Full Product Details