Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 14898660-14899101 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGD5
Entrez Gene ID
152273
Gene Name
FYVE, RhoGEF and PH domain containing 5
Gene Aliases
ZFYVE23
UniGene
Hs.412406
Gene Chromosome Location
Chr.3: 14818962-14934565 on Build GRCh38
Transcripts
NM_001320276.1 NM_152536.3 XM_011533422.1 XM_011533423.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4801362 c.3126C>T p.R1042R Chr.3: 14898798-14898798 on Build GRCh38
5417107 c.2373G>A p.A791A Chr.3: 14898768-14898768 on Build GRCh38
6043622 c.2420T>C p.V807A Chr.3: 14898815-14898815 on Build GRCh38
6022573 c.2356G>T p.G786* Chr.3: 14898751-14898751 on Build GRCh38
1039798 c.2372C>T p.A791V Chr.3: 14898767-14898767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs750191711 Chr.3: 14898765-14898765 on Build GRCh38
rs765569453 Chr.3: 14898711-14898711 on Build GRCh38
rs777686691 Chr.3: 14898784-14898784 on Build GRCh38
rs376308403 Chr.3: 14898860-14898860 on Build GRCh38
rs753342615 Chr.3: 14898836-14898836 on Build GRCh38
rs758570732 Chr.3: 14898767-14898767 on Build GRCh38
rs775179578 Chr.3: 14898801-14898801 on Build GRCh38
rs1015519590 Chr.3: 14898944-14898944 on Build GRCh38
rs1030590903 Chr.3: 14898964-14898964 on Build GRCh38
rs557709381 Chr.3: 14898946-14898946 on Build GRCh38
rs757177669 Chr.3: 14898763-14898763 on Build GRCh38
rs764168603 Chr.3: 14898742-14898742 on Build GRCh38
rs774895170 Chr.3: 14898869-14898885 on Build GRCh38
rs780168216 Chr.3: 14898988-14898988 on Build GRCh38
rs778864696 Chr.3: 14898764-14898764 on Build GRCh38
rs189944242 Chr.3: 14898741-14898741 on Build GRCh38
rs1028361525 Chr.3: 14898685-14898685 on Build GRCh38
rs371489383 Chr.3: 14898936-14898936 on Build GRCh38
rs569963284 Chr.3: 14898730-14898730 on Build GRCh38
rs768715676 Chr.3: 14898777-14898777 on Build GRCh38
rs377247404 Chr.3: 14898691-14898691 on Build GRCh38
rs996973818 Chr.3: 14898913-14898913 on Build GRCh38
rs762229922 Chr.3: 14898693-14898693 on Build GRCh38
rs111242570 Chr.3: 14898941-14898941 on Build GRCh38
rs575495610 Chr.3: 14899036-14899036 on Build GRCh38
rs910167793 Chr.3: 14899065-14899065 on Build GRCh38
rs61757393 Chr.3: 14898768-14898768 on Build GRCh38
rs763332666 Chr.3: 14898788-14898788 on Build GRCh38
rs983037799 Chr.3: 14899064-14899064 on Build GRCh38
rs866530041 Chr.3: 14898851-14898851 on Build GRCh38
rs577582641 Chr.3: 14899061-14899061 on Build GRCh38
rs1030594825 Chr.3: 14898886-14898886 on Build GRCh38
rs748664936 Chr.3: 14898781-14898781 on Build GRCh38
rs765217029 Chr.3: 14898845-14898845 on Build GRCh38
rs200528220 Chr.3: 14898780-14898780 on Build GRCh38
rs763685484 Chr.3: 14898814-14898814 on Build GRCh38
rs999402211 Chr.3: 14898934-14898934 on Build GRCh38
rs759119645 Chr.3: 14898726-14898726 on Build GRCh38
rs996012465 Chr.3: 14898868-14898868 on Build GRCh38
rs372659973 Chr.3: 14898743-14898743 on Build GRCh38
rs369369417 Chr.3: 14898775-14898775 on Build GRCh38
rs890356329 Chr.3: 14898917-14898917 on Build GRCh38
rs796262677 Chr.3: 14898968-14898968 on Build GRCh38
rs760329946 Chr.3: 14898812-14898812 on Build GRCh38
rs745996002 Chr.3: 14898798-14898798 on Build GRCh38
rs917377968 Chr.3: 14898744-14898744 on Build GRCh38
rs928223851 Chr.3: 14898745-14898745 on Build GRCh38
rs372788718 Chr.3: 14898797-14898797 on Build GRCh38
rs1005984423 Chr.3: 14898931-14898931 on Build GRCh38
rs970618732 Chr.3: 14899045-14899045 on Build GRCh38
rs200982279 Chr.3: 14898738-14898738 on Build GRCh38
rs1023365159 Chr.3: 14899018-14899018 on Build GRCh38
rs925714371 Chr.3: 14899063-14899063 on Build GRCh38
rs750233057 Chr.3: 14898847-14898847 on Build GRCh38
rs1047071109 Chr.3: 14898776-14898776 on Build GRCh38
rs182114359 Chr.3: 14899075-14899075 on Build GRCh38
rs767162218 Chr.3: 14898736-14898736 on Build GRCh38
rs761846937 Chr.3: 14898844-14898844 on Build GRCh38
rs780205998 Chr.3: 14898861-14898861 on Build GRCh38
rs751791228 Chr.3: 14898871-14898871 on Build GRCh38
rs969572146 Chr.3: 14899019-14899019 on Build GRCh38
rs773503740 Chr.3: 14898719-14898719 on Build GRCh38
rs982273066 Chr.3: 14899029-14899029 on Build GRCh38

View Full Product Details