Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15227726-15228236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CAPN7
Entrez Gene ID
23473
Gene Name
calpain 7
Gene Aliases
CALPAIN7, PALBH
UniGene
Hs.595234
Gene Chromosome Location
Chr.3: 15206226-15252916 on Build GRCh38
Transcripts
NM_014296.2 XM_011533555.2 XM_011533556.2 XM_011533557.2 XM_017006043.1 XM_017006044.1 XM_017006045.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1040094 c.852G>T p.Q284H Chr.3: 15227965-15227965 on Build GRCh38
6544264 c.734G>T p.W245L Chr.3: 15227847-15227847 on Build GRCh38
5931244 c.811C>T p.P271S Chr.3: 15227924-15227924 on Build GRCh38
1291633 c.780G>A p.K260K Chr.3: 15227893-15227893 on Build GRCh38
5904194 c.776C>T p.S259F Chr.3: 15227889-15227889 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs891222370 Chr.3: 15227807-15227807 on Build GRCh38
rs1016173793 Chr.3: 15227813-15227813 on Build GRCh38
rs906620538 Chr.3: 15227759-15227759 on Build GRCh38
rs745719251 Chr.3: 15227885-15227885 on Build GRCh38
rs775844215 Chr.3: 15227903-15227903 on Build GRCh38
rs779916465 Chr.3: 15228207-15228207 on Build GRCh38
rs778875307 Chr.3: 15228001-15228001 on Build GRCh38
rs1026881505 Chr.3: 15227974-15227974 on Build GRCh38
rs867713702 Chr.3: 15227782-15227782 on Build GRCh38
rs752697560 Chr.3: 15227829-15227830 on Build GRCh38
rs112673046 Chr.3: 15227856-15227856 on Build GRCh38
rs745784159 Chr.3: 15228003-15228003 on Build GRCh38
rs545317180 Chr.3: 15228008-15228008 on Build GRCh38
rs1052866075 Chr.3: 15228186-15228186 on Build GRCh38
rs938433601 Chr.3: 15228045-15228045 on Build GRCh38
rs900430772 Chr.3: 15228151-15228151 on Build GRCh38
rs982085838 Chr.3: 15228063-15228063 on Build GRCh38
rs755718428 Chr.3: 15227817-15227817 on Build GRCh38
rs3752838 Chr.3: 15227927-15227927 on Build GRCh38
rs758856132 Chr.3: 15227949-15227949 on Build GRCh38
rs375040334 Chr.3: 15227997-15227997 on Build GRCh38
rs375050126 Chr.3: 15227850-15227850 on Build GRCh38
rs989578104 Chr.3: 15228004-15228004 on Build GRCh38
rs1006165259 Chr.3: 15227803-15227803 on Build GRCh38
rs777358291 Chr.3: 15227983-15227983 on Build GRCh38
rs201287819 Chr.3: 15228150-15228150 on Build GRCh38
rs774236908 Chr.3: 15227858-15227858 on Build GRCh38
rs76753335 Chr.3: 15228086-15228086 on Build GRCh38
rs963879420 Chr.3: 15227971-15227971 on Build GRCh38
rs907728457 Chr.3: 15228117-15228117 on Build GRCh38
rs771975262 Chr.3: 15228007-15228007 on Build GRCh38
rs765884457 Chr.3: 15227936-15227936 on Build GRCh38
rs34577287 Chr.3: 15228005-15228006 on Build GRCh38
rs368733472 Chr.3: 15228013-15228013 on Build GRCh38
rs202072247 Chr.3: 15227947-15227947 on Build GRCh38
rs756036341 Chr.3: 15228132-15228132 on Build GRCh38
rs368130672 Chr.3: 15227900-15227900 on Build GRCh38
rs761942868 Chr.3: 15227930-15227930 on Build GRCh38
rs1047589005 Chr.3: 15227784-15227784 on Build GRCh38
rs200044345 Chr.3: 15227919-15227919 on Build GRCh38
rs777270524 Chr.3: 15227825-15227825 on Build GRCh38
rs996088566 Chr.3: 15228170-15228170 on Build GRCh38
rs941863172 Chr.3: 15228133-15228133 on Build GRCh38
rs928297595 Chr.3: 15228029-15228029 on Build GRCh38
rs999313041 Chr.3: 15228191-15228191 on Build GRCh38
rs549939411 Chr.3: 15228026-15228026 on Build GRCh38
rs886266638 Chr.3: 15227792-15227792 on Build GRCh38
rs766648836 Chr.3: 15227953-15227953 on Build GRCh38
rs35880488 Chr.3: 15228206-15228207 on Build GRCh38
rs1031194121 Chr.3: 15227993-15227993 on Build GRCh38
rs1051479504 Chr.3: 15228175-15228175 on Build GRCh38
rs751885606 Chr.3: 15227955-15227955 on Build GRCh38
rs879737450 Chr.3: 15227801-15227801 on Build GRCh38
rs890100428 Chr.3: 15228183-15228183 on Build GRCh38
rs371586368 Chr.3: 15227991-15227991 on Build GRCh38
rs781420761 Chr.3: 15228126-15228130 on Build GRCh38
rs930113737 Chr.3: 15228090-15228090 on Build GRCh38
rs1047570207 Chr.3: 15228100-15228100 on Build GRCh38
rs1030379002 Chr.3: 15227901-15227901 on Build GRCh38
rs746767594 Chr.3: 15228010-15228010 on Build GRCh38
rs755737353 Chr.3: 15227956-15227956 on Build GRCh38
rs748740884 Chr.3: 15227832-15227832 on Build GRCh38
rs756069748 Chr.3: 15227873-15227874 on Build GRCh38

View Full Product Details