Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 97077922-97078454 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYD
Entrez Gene ID
1806
Gene Name
dihydropyrimidine dehydrogenase
Gene Aliases
DHP, DHPDHASE, DPD
UniGene
Hs.335034
Gene Chromosome Location
Chr.1: 97077743-97921059 on Build GRCh38
Transcripts
NM_000110.3 XM_005270562.3 XM_017000507.1 XM_017000508.1 XM_017000509.1 XM_017000510.1

SNP Detail

SNP ID Chromosome Location
rs672601269 Chr.1: 97078015-97078015 on Build GRCh38
rs567178473 Chr.1: 97078426-97078426 on Build GRCh38
rs893063528 Chr.1: 97077996-97077996 on Build GRCh38
rs985290642 Chr.1: 97078346-97078346 on Build GRCh38
rs672601271 Chr.1: 97078240-97078240 on Build GRCh38
rs976574994 Chr.1: 97077970-97077970 on Build GRCh38
rs912874652 Chr.1: 97078396-97078396 on Build GRCh38
rs949792299 Chr.1: 97078169-97078169 on Build GRCh38
rs55696854 Chr.1: 97078068-97078068 on Build GRCh38
rs1023870699 Chr.1: 97078153-97078153 on Build GRCh38
rs989138464 Chr.1: 97078390-97078390 on Build GRCh38
rs528999329 Chr.1: 97078408-97078409 on Build GRCh38
rs780504182 Chr.1: 97078030-97078030 on Build GRCh38
rs999524953 Chr.1: 97078080-97078080 on Build GRCh38
rs866711735 Chr.1: 97078253-97078253 on Build GRCh38
rs1035521205 Chr.1: 97078101-97078101 on Build GRCh38
rs1005552853 Chr.1: 97078236-97078236 on Build GRCh38
rs932484793 Chr.1: 97078040-97078040 on Build GRCh38
rs922405307 Chr.1: 97077979-97077979 on Build GRCh38
rs672601270 Chr.1: 97078065-97078065 on Build GRCh38
rs1045909606 Chr.1: 97078177-97078177 on Build GRCh38
rs1041563208 Chr.1: 97078050-97078050 on Build GRCh38
rs755469970 Chr.1: 97078155-97078155 on Build GRCh38
rs17470762 Chr.1: 97078076-97078076 on Build GRCh38
rs935946036 Chr.1: 97078124-97078124 on Build GRCh38
rs1029067403 Chr.1: 97078311-97078311 on Build GRCh38
rs1042481 Chr.1: 97078416-97078416 on Build GRCh38
rs976162155 Chr.1: 97078303-97078303 on Build GRCh38
rs291592 Chr.1: 97078208-97078208 on Build GRCh38
rs1041617229 Chr.1: 97078387-97078387 on Build GRCh38
rs914479701 Chr.1: 97078319-97078319 on Build GRCh38
rs1014886873 Chr.1: 97078261-97078261 on Build GRCh38
rs966457799 Chr.1: 97078300-97078300 on Build GRCh38
rs947206632 Chr.1: 97078045-97078046 on Build GRCh38
rs886046569 Chr.1: 97078124-97078124 on Build GRCh38
rs386633699 Chr.1: 97078196-97078208 on Build GRCh38
rs886046571 Chr.1: 97078390-97078390 on Build GRCh38
rs962983345 Chr.1: 97078166-97078166 on Build GRCh38
rs772715720 Chr.1: 97078389-97078389 on Build GRCh38
rs1042482 Chr.1: 97078403-97078403 on Build GRCh38
rs765882514 Chr.1: 97078420-97078420 on Build GRCh38
rs34856860 Chr.1: 97078420-97078421 on Build GRCh38
rs886046570 Chr.1: 97078311-97078311 on Build GRCh38
rs1052781258 Chr.1: 97078150-97078150 on Build GRCh38
rs137984871 Chr.1: 97078193-97078193 on Build GRCh38
rs75542363 Chr.1: 97078364-97078364 on Build GRCh38
rs535104911 Chr.1: 97078341-97078341 on Build GRCh38
rs291593 Chr.1: 97078196-97078196 on Build GRCh38
rs1001645817 Chr.1: 97078195-97078195 on Build GRCh38
rs925839839 Chr.1: 97078108-97078108 on Build GRCh38

View Full Product Details