Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 97098264-97098801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPYD
Entrez Gene ID
1806
Gene Name
dihydropyrimidine dehydrogenase
Gene Aliases
DHP, DHPDHASE, DPD
UniGene
Hs.335034
Gene Chromosome Location
Chr.1: 97077743-97921059 on Build GRCh38
Transcripts
NM_000110.3 XM_005270562.3 XM_017000507.1 XM_017000508.1 XM_017000509.1 XM_017000510.1 NR_046590.1

Gene Symbol
DPYD-AS1
Entrez Gene ID
100873932
Gene Name
DPYD antisense RNA 1
Gene Aliases
-
UniGene
Hs.682651
Gene Chromosome Location
Chr.1: 97095923-97322955 on Build GRCh38
Transcripts
NM_000110.3 XM_005270562.3 XM_017000507.1 XM_017000508.1 XM_017000509.1 XM_017000510.1 NR_046590.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6649099 c.2731T>C p.C911R Chr.1: 97098524-97098524 on Build GRCh38
913581 c.2630C>A p.P877Q Chr.1: 97098625-97098625 on Build GRCh38
913580 c.2695G>T p.E899* Chr.1: 97098560-97098560 on Build GRCh38
6257598 c.2755C>A p.P919T Chr.1: 97098500-97098500 on Build GRCh38
1126911 c.2683A>G p.I895V Chr.1: 97098572-97098572 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752102711 Chr.1: 97098633-97098633 on Build GRCh38
rs773002572 Chr.1: 97098609-97098609 on Build GRCh38
rs563269168 Chr.1: 97098738-97098738 on Build GRCh38
rs772826416 Chr.1: 97098502-97098502 on Build GRCh38
rs755729055 Chr.1: 97098587-97098587 on Build GRCh38
rs930962520 Chr.1: 97098646-97098646 on Build GRCh38
rs375268978 Chr.1: 97098470-97098470 on Build GRCh38
rs771890504 Chr.1: 97098484-97098484 on Build GRCh38
rs759732624 Chr.1: 97098486-97098486 on Build GRCh38
rs771055121 Chr.1: 97098753-97098753 on Build GRCh38
rs372566634 Chr.1: 97098485-97098485 on Build GRCh38
rs956949059 Chr.1: 97098495-97098495 on Build GRCh38
rs745583005 Chr.1: 97098459-97098459 on Build GRCh38
rs376391343 Chr.1: 97098582-97098582 on Build GRCh38
rs918508856 Chr.1: 97098567-97098567 on Build GRCh38
rs182807909 Chr.1: 97098409-97098409 on Build GRCh38
rs1040929895 Chr.1: 97098703-97098703 on Build GRCh38
rs1008027136 Chr.1: 97098344-97098344 on Build GRCh38
rs199712715 Chr.1: 97098452-97098452 on Build GRCh38
rs113704446 Chr.1: 97098338-97098338 on Build GRCh38
rs886046575 Chr.1: 97098497-97098497 on Build GRCh38
rs749180344 Chr.1: 97098472-97098472 on Build GRCh38
rs768200107 Chr.1: 97098596-97098596 on Build GRCh38
rs771839581 Chr.1: 97098602-97098602 on Build GRCh38
rs762427274 Chr.1: 97098660-97098660 on Build GRCh38
rs764679468 Chr.1: 97098524-97098524 on Build GRCh38
rs866273057 Chr.1: 97098516-97098516 on Build GRCh38
rs983109437 Chr.1: 97098661-97098661 on Build GRCh38
rs577188291 Chr.1: 97098397-97098397 on Build GRCh38
rs1010477474 Chr.1: 97098314-97098314 on Build GRCh38
rs56406418 Chr.1: 97098468-97098468 on Build GRCh38
rs761017799 Chr.1: 97098680-97098680 on Build GRCh38
rs201035051 Chr.1: 97098632-97098632 on Build GRCh38
rs556768807 Chr.1: 97098402-97098402 on Build GRCh38
rs576661222 Chr.1: 97098645-97098645 on Build GRCh38
rs11165777 Chr.1: 97098353-97098353 on Build GRCh38
rs537157979 Chr.1: 97098290-97098290 on Build GRCh38
rs901963323 Chr.1: 97098369-97098369 on Build GRCh38
rs559427764 Chr.1: 97098594-97098594 on Build GRCh38
rs932078269 Chr.1: 97098727-97098727 on Build GRCh38
rs1057516873 Chr.1: 97098575-97098575 on Build GRCh38
rs745451696 Chr.1: 97098663-97098664 on Build GRCh38
rs762911226 Chr.1: 97098623-97098623 on Build GRCh38
rs767928936 Chr.1: 97098672-97098672 on Build GRCh38
rs751998381 Chr.1: 97098444-97098444 on Build GRCh38
rs919596571 Chr.1: 97098621-97098621 on Build GRCh38
rs147545709 Chr.1: 97098599-97098599 on Build GRCh38
rs958829842 Chr.1: 97098437-97098437 on Build GRCh38
rs757695236 Chr.1: 97098548-97098548 on Build GRCh38
rs943930002 Chr.1: 97098696-97098696 on Build GRCh38
rs757750025 Chr.1: 97098446-97098446 on Build GRCh38
rs1057516894 Chr.1: 97098501-97098501 on Build GRCh38
rs1046948873 Chr.1: 97098316-97098316 on Build GRCh38
rs761573540 Chr.1: 97098480-97098480 on Build GRCh38
rs768507059 Chr.1: 97098475-97098475 on Build GRCh38
rs1006621446 Chr.1: 97098726-97098726 on Build GRCh38
rs886952024 Chr.1: 97098330-97098330 on Build GRCh38
rs1045155986 Chr.1: 97098287-97098287 on Build GRCh38
rs752151811 Chr.1: 97098540-97098540 on Build GRCh38
rs368199887 Chr.1: 97098371-97098371 on Build GRCh38
rs369044951 Chr.1: 97098479-97098479 on Build GRCh38
rs759766897 Chr.1: 97098610-97098610 on Build GRCh38
rs55674432 Chr.1: 97098616-97098616 on Build GRCh38
rs771930534 Chr.1: 97098493-97098493 on Build GRCh38
rs557608721 Chr.1: 97098361-97098361 on Build GRCh38
rs1057517230 Chr.1: 97098507-97098507 on Build GRCh38
rs1801267 Chr.1: 97098598-97098598 on Build GRCh38
rs188052243 Chr.1: 97098577-97098577 on Build GRCh38
rs903854232 Chr.1: 97098751-97098751 on Build GRCh38
rs779573574 Chr.1: 97098591-97098591 on Build GRCh38
rs769932607 Chr.1: 97098613-97098613 on Build GRCh38
rs766852110 Chr.1: 97098669-97098670 on Build GRCh38
rs778614771 Chr.1: 97098509-97098509 on Build GRCh38

View Full Product Details