Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100140297-100140796 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TRMT13
Entrez Gene ID
54482
Gene Name
tRNA methyltransferase 13 homolog
Gene Aliases
CCDC76
UniGene
Hs.729095
Gene Chromosome Location
Chr.1: 100133080-100150498 on Build GRCh38
Transcripts
NM_019083.2 NR_135078.1 XM_005270945.3 XM_005270946.2 XM_017001504.1 XM_017001505.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5842861 c.409C>T p.L137F Chr.1: 100140422-100140422 on Build GRCh38
7137020 c.472G>T p.D158Y Chr.1: 100140485-100140485 on Build GRCh38
4444206 c.437C>T p.A146V Chr.1: 100140450-100140450 on Build GRCh38
4019795 c.444C>T p.H148H Chr.1: 100140457-100140457 on Build GRCh38
6280080 c.418C>T p.H140Y Chr.1: 100140431-100140431 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773499082 Chr.1: 100140388-100140388 on Build GRCh38
rs747080442 Chr.1: 100140389-100140389 on Build GRCh38
rs770979011 Chr.1: 100140462-100140462 on Build GRCh38
rs753882742 Chr.1: 100140519-100140519 on Build GRCh38
rs754700215 Chr.1: 100140523-100140523 on Build GRCh38
rs771068741 Chr.1: 100140395-100140395 on Build GRCh38
rs1039219659 Chr.1: 100140647-100140647 on Build GRCh38
rs574550494 Chr.1: 100140674-100140674 on Build GRCh38
rs779664573 Chr.1: 100140359-100140359 on Build GRCh38
rs973640621 Chr.1: 100140723-100140723 on Build GRCh38
rs998675326 Chr.1: 100140609-100140609 on Build GRCh38
rs535127116 Chr.1: 100140680-100140680 on Build GRCh38
rs781402159 Chr.1: 100140578-100140578 on Build GRCh38
rs767309061 Chr.1: 100140510-100140510 on Build GRCh38
rs554962370 Chr.1: 100140694-100140694 on Build GRCh38
rs756794234 Chr.1: 100140432-100140432 on Build GRCh38
rs532713466 Chr.1: 100140466-100140466 on Build GRCh38
rs904377465 Chr.1: 100140513-100140513 on Build GRCh38
rs796199435 Chr.1: 100140568-100140568 on Build GRCh38
rs895867101 Chr.1: 100140657-100140657 on Build GRCh38
rs764557381 Chr.1: 100140477-100140477 on Build GRCh38
rs780045975 Chr.1: 100140451-100140451 on Build GRCh38
rs943708748 Chr.1: 100140423-100140423 on Build GRCh38
rs762576186 Chr.1: 100140587-100140589 on Build GRCh38
rs943492721 Chr.1: 100140645-100140645 on Build GRCh38
rs747274613 Chr.1: 100140461-100140461 on Build GRCh38
rs763386000 Chr.1: 100140412-100140412 on Build GRCh38
rs147168066 Chr.1: 100140460-100140460 on Build GRCh38
rs368205547 Chr.1: 100140408-100140408 on Build GRCh38
rs201744498 Chr.1: 100140544-100140544 on Build GRCh38
rs763219875 Chr.1: 100140474-100140474 on Build GRCh38
rs552515076 Chr.1: 100140472-100140472 on Build GRCh38
rs549989267 Chr.1: 100140434-100140434 on Build GRCh38
rs868129983 Chr.1: 100140411-100140422 on Build GRCh38
rs777094627 Chr.1: 100140401-100140401 on Build GRCh38
rs192753169 Chr.1: 100140695-100140695 on Build GRCh38
rs754315080 Chr.1: 100140547-100140559 on Build GRCh38
rs778821066 Chr.1: 100140530-100140530 on Build GRCh38
rs370753617 Chr.1: 100140457-100140457 on Build GRCh38
rs1032177234 Chr.1: 100140654-100140654 on Build GRCh38
rs1022969913 Chr.1: 100140676-100140676 on Build GRCh38
rs765799603 Chr.1: 100140403-100140403 on Build GRCh38
rs764514713 Chr.1: 100140516-100140526 on Build GRCh38
rs902217017 Chr.1: 100140679-100140679 on Build GRCh38
rs919283663 Chr.1: 100140558-100140558 on Build GRCh38
rs756348411 Chr.1: 100140437-100140437 on Build GRCh38
rs766572079 Chr.1: 100140673-100140673 on Build GRCh38
rs756075541 Chr.1: 100140450-100140450 on Build GRCh38
rs763637129 Chr.1: 100140415-100140415 on Build GRCh38
rs199548690 Chr.1: 100140362-100140362 on Build GRCh38
rs570284946 Chr.1: 100140386-100140386 on Build GRCh38
rs149250065 Chr.1: 100140467-100140467 on Build GRCh38
rs901559838 Chr.1: 100140660-100140660 on Build GRCh38
rs909474101 Chr.1: 100140593-100140593 on Build GRCh38
rs997294062 Chr.1: 100140672-100140672 on Build GRCh38
rs766214911 Chr.1: 100140515-100140515 on Build GRCh38
rs373612434 Chr.1: 100140484-100140484 on Build GRCh38
rs367748763 Chr.1: 100140485-100140485 on Build GRCh38
rs760089360 Chr.1: 100140402-100140402 on Build GRCh38
rs751290098 Chr.1: 100140420-100140420 on Build GRCh38
rs757434500 Chr.1: 100140557-100140557 on Build GRCh38
rs959435814 Chr.1: 100140453-100140453 on Build GRCh38
rs749886415 Chr.1: 100140512-100140512 on Build GRCh38
rs754862301 Chr.1: 100140458-100140458 on Build GRCh38
rs750636958 Chr.1: 100140350-100140350 on Build GRCh38

View Full Product Details