Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100188503-100189005 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBT
Entrez Gene ID
1629
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B
UniGene
Hs.709187
Gene Chromosome Location
Chr.1: 100186922-100249864 on Build GRCh38
Transcripts
NM_001918.3

SNP Detail

SNP ID Chromosome Location
rs577942845 Chr.1: 100188814-100188814 on Build GRCh38
rs190432589 Chr.1: 100188841-100188841 on Build GRCh38
rs977756474 Chr.1: 100188601-100188601 on Build GRCh38
rs901264037 Chr.1: 100188758-100188758 on Build GRCh38
rs938981786 Chr.1: 100188656-100188656 on Build GRCh38
rs1039990503 Chr.1: 100188865-100188865 on Build GRCh38
rs750432218 Chr.1: 100188561-100188561 on Build GRCh38
rs182980145 Chr.1: 100188948-100188948 on Build GRCh38
rs369203638 Chr.1: 100188887-100188887 on Build GRCh38
rs916977862 Chr.1: 100188870-100188870 on Build GRCh38
rs533835450 Chr.1: 100188821-100188821 on Build GRCh38
rs993004255 Chr.1: 100188867-100188867 on Build GRCh38
rs143822588 Chr.1: 100188897-100188897 on Build GRCh38
rs1051132588 Chr.1: 100188824-100188824 on Build GRCh38
rs930039065 Chr.1: 100188817-100188817 on Build GRCh38
rs1050265816 Chr.1: 100188911-100188911 on Build GRCh38
rs1034516244 Chr.1: 100188864-100188864 on Build GRCh38
rs927631296 Chr.1: 100188649-100188649 on Build GRCh38
rs754672374 Chr.1: 100188797-100188797 on Build GRCh38
rs138171272 Chr.1: 100188957-100188957 on Build GRCh38
rs1034698356 Chr.1: 100188970-100188970 on Build GRCh38
rs1010764434 Chr.1: 100188968-100188968 on Build GRCh38
rs966843746 Chr.1: 100188591-100188591 on Build GRCh38
rs557971614 Chr.1: 100188801-100188801 on Build GRCh38
rs756568105 Chr.1: 100188744-100188744 on Build GRCh38
rs944511911 Chr.1: 100188924-100188924 on Build GRCh38
rs941223981 Chr.1: 100188696-100188696 on Build GRCh38
rs185528579 Chr.1: 100188557-100188557 on Build GRCh38
rs990705798 Chr.1: 100188659-100188659 on Build GRCh38
rs927475869 Chr.1: 100188537-100188537 on Build GRCh38
rs766260651 Chr.1: 100188662-100188662 on Build GRCh38
rs1036452327 Chr.1: 100188631-100188631 on Build GRCh38
rs893354618 Chr.1: 100188886-100188886 on Build GRCh38
rs897269544 Chr.1: 100188786-100188786 on Build GRCh38
rs940335253 Chr.1: 100188628-100188628 on Build GRCh38
rs1020725943 Chr.1: 100188807-100188807 on Build GRCh38
rs1006859599 Chr.1: 100188853-100188853 on Build GRCh38
rs535852086 Chr.1: 100188565-100188565 on Build GRCh38
rs917005539 Chr.1: 100188588-100188588 on Build GRCh38
rs569131984 Chr.1: 100188685-100188685 on Build GRCh38
rs957567255 Chr.1: 100188556-100188556 on Build GRCh38
rs555327269 Chr.1: 100188676-100188676 on Build GRCh38
rs888987485 Chr.1: 100188706-100188706 on Build GRCh38
rs968986697 Chr.1: 100188836-100188836 on Build GRCh38
rs1020858381 Chr.1: 100188901-100188901 on Build GRCh38
rs920288699 Chr.1: 100188892-100188892 on Build GRCh38
rs889039306 Chr.1: 100188919-100188919 on Build GRCh38
rs746457018 Chr.1: 100188714-100188714 on Build GRCh38
rs1010212655 Chr.1: 100188764-100188764 on Build GRCh38
rs145958213 Chr.1: 100188573-100188574 on Build GRCh38
rs901238744 Chr.1: 100188955-100188955 on Build GRCh38
rs966519152 Chr.1: 100188908-100188908 on Build GRCh38
rs538185208 Chr.1: 100188718-100188718 on Build GRCh38
rs1050210429 Chr.1: 100188703-100188703 on Build GRCh38

View Full Product Details