Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100190882-100191423 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DBT
Entrez Gene ID
1629
Gene Name
dihydrolipoamide branched chain transacylase E2
Gene Aliases
BCATE2, BCKAD-E2, BCKADE2, BCOADC-E2, E2, E2B
UniGene
Hs.709187
Gene Chromosome Location
Chr.1: 100186922-100249864 on Build GRCh38
Transcripts
NM_001918.3

SNP Detail

SNP ID Chromosome Location
rs766030719 Chr.1: 100191094-100191094 on Build GRCh38
rs920810590 Chr.1: 100191391-100191391 on Build GRCh38
rs1046855256 Chr.1: 100191081-100191081 on Build GRCh38
rs1000509429 Chr.1: 100191243-100191243 on Build GRCh38
rs759070368 Chr.1: 100191255-100191255 on Build GRCh38
rs367578706 Chr.1: 100191332-100191332 on Build GRCh38
rs950874579 Chr.1: 100191393-100191393 on Build GRCh38
rs747087454 Chr.1: 100191341-100191341 on Build GRCh38
rs374637405 Chr.1: 100191362-100191362 on Build GRCh38
rs943709170 Chr.1: 100191164-100191164 on Build GRCh38
rs996775059 Chr.1: 100191258-100191258 on Build GRCh38
rs1042103845 Chr.1: 100191061-100191061 on Build GRCh38
rs11166413 Chr.1: 100190938-100190938 on Build GRCh38
rs960093950 Chr.1: 100191350-100191350 on Build GRCh38
rs973682404 Chr.1: 100191385-100191385 on Build GRCh38
rs1014236603 Chr.1: 100191364-100191364 on Build GRCh38
rs753581201 Chr.1: 100191141-100191142 on Build GRCh38
rs772160709 Chr.1: 100191208-100191208 on Build GRCh38
rs933063340 Chr.1: 100191059-100191059 on Build GRCh38
rs747083961 Chr.1: 100191238-100191238 on Build GRCh38
rs904365505 Chr.1: 100191327-100191327 on Build GRCh38
rs895048969 Chr.1: 100191284-100191284 on Build GRCh38
rs150770445 Chr.1: 100190927-100190927 on Build GRCh38
rs558178963 Chr.1: 100191126-100191126 on Build GRCh38
rs920769590 Chr.1: 100190959-100190959 on Build GRCh38
rs1043328048 Chr.1: 100191321-100191321 on Build GRCh38
rs916185779 Chr.1: 100190910-100190910 on Build GRCh38
rs116806523 Chr.1: 100190908-100190908 on Build GRCh38
rs6701655 Chr.1: 100191105-100191105 on Build GRCh38
rs1039165815 Chr.1: 100191013-100191013 on Build GRCh38
rs12044663 Chr.1: 100191383-100191383 on Build GRCh38
rs544167842 Chr.1: 100191167-100191167 on Build GRCh38
rs943647358 Chr.1: 100190978-100190978 on Build GRCh38
rs1040720679 Chr.1: 100191169-100191169 on Build GRCh38
rs1022956221 Chr.1: 100191378-100191378 on Build GRCh38
rs200067204 Chr.1: 100191153-100191154 on Build GRCh38
rs929425083 Chr.1: 100191018-100191018 on Build GRCh38
rs777946389 Chr.1: 100191330-100191330 on Build GRCh38
rs1026371336 Chr.1: 100191380-100191380 on Build GRCh38
rs768894861 Chr.1: 100190917-100190917 on Build GRCh38
rs370910725 Chr.1: 100191132-100191132 on Build GRCh38

View Full Product Details