Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100873479-100873978 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL2
Entrez Gene ID
2135
Gene Name
exostosin like glycosyltransferase 2
Gene Aliases
EXTR2
UniGene
Hs.357637
Gene Chromosome Location
Chr.1: 100872372-100895411 on Build GRCh38
Transcripts
NM_001033025.2 NM_001261440.1 NM_001261441.1 NM_001261442.1 NM_001439.3 NR_048570.1 XM_005270621.1 XM_011540994.2 XM_011540995.2 XM_011540996.2 XM_017000651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
414218 c.983G>C p.R328T Chr.1: 100873952-100873952 on Build GRCh38
109109 c.989T>A p.I330K Chr.1: 100873946-100873946 on Build GRCh38
5637326 c.983G>A p.R328K Chr.1: 100873952-100873952 on Build GRCh38
1491654 c.991T>A p.*331K Chr.1: 100873944-100873944 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs547403385 Chr.1: 100873593-100873593 on Build GRCh38
rs145730346 Chr.1: 100873946-100873946 on Build GRCh38
rs536077825 Chr.1: 100873616-100873616 on Build GRCh38
rs181186368 Chr.1: 100873800-100873800 on Build GRCh38
rs749170439 Chr.1: 100873640-100873640 on Build GRCh38
rs768375439 Chr.1: 100873752-100873752 on Build GRCh38
rs757701745 Chr.1: 100873920-100873920 on Build GRCh38
rs987715785 Chr.1: 100873830-100873830 on Build GRCh38
rs188814460 Chr.1: 100873751-100873751 on Build GRCh38
rs754429497 Chr.1: 100873919-100873922 on Build GRCh38
rs774333579 Chr.1: 100873880-100873880 on Build GRCh38
rs1035368359 Chr.1: 100873528-100873528 on Build GRCh38
rs186189164 Chr.1: 100873869-100873869 on Build GRCh38
rs749139678 Chr.1: 100873700-100873700 on Build GRCh38
rs372520087 Chr.1: 100873944-100873944 on Build GRCh38
rs753596576 Chr.1: 100873938-100873938 on Build GRCh38
rs556276130 Chr.1: 100873629-100873629 on Build GRCh38
rs779557476 Chr.1: 100873934-100873934 on Build GRCh38
rs769582461 Chr.1: 100873512-100873512 on Build GRCh38
rs768165809 Chr.1: 100873919-100873919 on Build GRCh38
rs887055162 Chr.1: 100873536-100873536 on Build GRCh38
rs1016757504 Chr.1: 100873667-100873667 on Build GRCh38
rs754632254 Chr.1: 100873939-100873939 on Build GRCh38
rs761808317 Chr.1: 100873906-100873906 on Build GRCh38
rs756547353 Chr.1: 100873929-100873929 on Build GRCh38
rs912110177 Chr.1: 100873868-100873869 on Build GRCh38
rs779329534 Chr.1: 100873934-100873939 on Build GRCh38
rs554076097 Chr.1: 100873836-100873836 on Build GRCh38
rs762360131 Chr.1: 100873911-100873911 on Build GRCh38
rs750719633 Chr.1: 100873928-100873928 on Build GRCh38
rs975135196 Chr.1: 100873722-100873722 on Build GRCh38
rs757288526 Chr.1: 100873798-100873798 on Build GRCh38
rs565807175 Chr.1: 100873924-100873924 on Build GRCh38
rs1003988010 Chr.1: 100873544-100873544 on Build GRCh38
rs569658966 Chr.1: 100873672-100873672 on Build GRCh38
rs567517871 Chr.1: 100873579-100873579 on Build GRCh38
rs1027952580 Chr.1: 100873738-100873738 on Build GRCh38
rs944640715 Chr.1: 100873804-100873804 on Build GRCh38

View Full Product Details