Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100878318-100878862 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL2
Entrez Gene ID
2135
Gene Name
exostosin like glycosyltransferase 2
Gene Aliases
EXTR2
UniGene
Hs.357637
Gene Chromosome Location
Chr.1: 100872372-100895411 on Build GRCh38
Transcripts
NM_001033025.2 NM_001261440.1 NM_001261441.1 NM_001261442.1 NM_001439.3 NR_048570.1 XM_005270621.1 XM_011540994.2 XM_011540995.2 XM_011540996.2 XM_017000651.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
160859 c.22G>C p.G8R Chr.1: 100878431-100878431 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs531095141 Chr.1: 100878757-100878757 on Build GRCh38
rs1027087958 Chr.1: 100878432-100878432 on Build GRCh38
rs1046683624 Chr.1: 100878651-100878651 on Build GRCh38
rs992093151 Chr.1: 100878354-100878354 on Build GRCh38
rs1016061032 Chr.1: 100878759-100878759 on Build GRCh38
rs539535395 Chr.1: 100878509-100878509 on Build GRCh38
rs965410477 Chr.1: 100878776-100878776 on Build GRCh38
rs548579384 Chr.1: 100878416-100878416 on Build GRCh38
rs879173240 Chr.1: 100878494-100878494 on Build GRCh38
rs373536553 Chr.1: 100878564-100878564 on Build GRCh38
rs760602921 Chr.1: 100878498-100878499 on Build GRCh38
rs774924776 Chr.1: 100878835-100878835 on Build GRCh38
rs901592142 Chr.1: 100878570-100878570 on Build GRCh38
rs371625428 Chr.1: 100878641-100878641 on Build GRCh38
rs913599632 Chr.1: 100878638-100878638 on Build GRCh38
rs368994013 Chr.1: 100878385-100878385 on Build GRCh38
rs1026244348 Chr.1: 100878775-100878775 on Build GRCh38
rs79194676 Chr.1: 100878502-100878502 on Build GRCh38
rs531026925 Chr.1: 100878468-100878468 on Build GRCh38
rs925103657 Chr.1: 100878567-100878567 on Build GRCh38
rs555386683 Chr.1: 100878721-100878721 on Build GRCh38
rs905419620 Chr.1: 100878663-100878663 on Build GRCh38
rs368365204 Chr.1: 100878562-100878562 on Build GRCh38
rs562342166 Chr.1: 100878418-100878418 on Build GRCh38
rs575465297 Chr.1: 100878826-100878826 on Build GRCh38
rs181802464 Chr.1: 100878364-100878364 on Build GRCh38
rs998571370 Chr.1: 100878825-100878825 on Build GRCh38
rs888616965 Chr.1: 100878750-100878750 on Build GRCh38
rs1002445194 Chr.1: 100878664-100878664 on Build GRCh38
rs759750697 Chr.1: 100878408-100878408 on Build GRCh38
rs752174364 Chr.1: 100878442-100878442 on Build GRCh38
rs41287274 Chr.1: 100878613-100878613 on Build GRCh38
rs142035650 Chr.1: 100878362-100878362 on Build GRCh38
rs923152780 Chr.1: 100878523-100878523 on Build GRCh38
rs763172677 Chr.1: 100878427-100878427 on Build GRCh38
rs185162752 Chr.1: 100878473-100878473 on Build GRCh38
rs971486813 Chr.1: 100878387-100878387 on Build GRCh38
rs771375829 Chr.1: 100878482-100878482 on Build GRCh38
rs1013076627 Chr.1: 100878716-100878716 on Build GRCh38
rs909014154 Chr.1: 100878525-100878525 on Build GRCh38
rs764681637 Chr.1: 100878436-100878436 on Build GRCh38
rs1048619556 Chr.1: 100878737-100878737 on Build GRCh38
rs893410034 Chr.1: 100878689-100878689 on Build GRCh38
rs1052611092 Chr.1: 100878624-100878624 on Build GRCh38
rs547017960 Chr.1: 100878528-100878528 on Build GRCh38

View Full Product Details