Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 100888730-100889065 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EXTL2
Entrez Gene ID
2135
Gene Name
exostosin like glycosyltransferase 2
Gene Aliases
EXTR2
UniGene
Hs.357637
Gene Chromosome Location
Chr.1: 100872372-100895411 on Build GRCh38
Transcripts
NM_001033025.2 NM_001261440.1 NM_001261441.1 NM_001261442.1 NM_001439.3 NR_048570.1 XM_005270621.1 XM_011540994.2 XM_011540995.2 XM_011540996.2 XM_017000651.1

SNP Detail

SNP ID Chromosome Location
rs548753335 Chr.1: 100888979-100888979 on Build GRCh38
rs565523516 Chr.1: 100888767-100888767 on Build GRCh38
rs1040920461 Chr.1: 100888973-100888973 on Build GRCh38
rs373028355 Chr.1: 100888770-100888770 on Build GRCh38
rs373536267 Chr.1: 100888847-100888847 on Build GRCh38
rs996604049 Chr.1: 100888993-100888993 on Build GRCh38
rs12747459 Chr.1: 100888923-100888923 on Build GRCh38
rs953827967 Chr.1: 100888792-100888792 on Build GRCh38
rs768548911 Chr.1: 100888844-100888844 on Build GRCh38
rs376269400 Chr.1: 100888820-100888820 on Build GRCh38
rs375873273 Chr.1: 100888887-100888891 on Build GRCh38
rs977688773 Chr.1: 100888804-100888804 on Build GRCh38
rs916055823 Chr.1: 100888873-100888873 on Build GRCh38
rs767940159 Chr.1: 100888765-100888765 on Build GRCh38
rs77649301 Chr.1: 100888830-100888830 on Build GRCh38
rs541265076 Chr.1: 100889038-100889038 on Build GRCh38
rs766003550 Chr.1: 100888778-100888778 on Build GRCh38
rs976289240 Chr.1: 100888838-100888838 on Build GRCh38
rs768784866 Chr.1: 100888757-100888757 on Build GRCh38
rs545047459 Chr.1: 100888876-100888876 on Build GRCh38
rs374861571 Chr.1: 100888760-100888760 on Build GRCh38
rs929278584 Chr.1: 100888917-100888917 on Build GRCh38
rs781142215 Chr.1: 100888920-100888920 on Build GRCh38
rs1048177592 Chr.1: 100888919-100888919 on Build GRCh38
rs914187042 Chr.1: 100888839-100888839 on Build GRCh38
rs567030478 Chr.1: 100888994-100888994 on Build GRCh38
rs200957907 Chr.1: 100888768-100888768 on Build GRCh38
rs764587983 Chr.1: 100888777-100888778 on Build GRCh38
rs968336407 Chr.1: 100888803-100888803 on Build GRCh38
rs76722689 Chr.1: 100888808-100888808 on Build GRCh38
rs111669428 Chr.1: 100888855-100888855 on Build GRCh38
rs556632131 Chr.1: 100888822-100888822 on Build GRCh38
rs998551356 Chr.1: 100889023-100889023 on Build GRCh38
rs569540230 Chr.1: 100888908-100888908 on Build GRCh38
rs904203293 Chr.1: 100889011-100889011 on Build GRCh38
rs554399525 Chr.1: 100888758-100888758 on Build GRCh38
rs920613288 Chr.1: 100888913-100888913 on Build GRCh38

View Full Product Details