Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15427425-15427951 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EAF1
Entrez Gene ID
85403
Gene Name
ELL associated factor 1
Gene Aliases
-
UniGene
Hs.716733
Gene Chromosome Location
Chr.3: 15427557-15442613 on Build GRCh38
Transcripts
NM_033083.6 XM_005265518.1 NM_001301790.1 NM_001301791.1 NM_001301792.1 NM_152396.3 XM_005264867.3 XM_005264870.4 XM_006712972.3 XM_011533357.2 XM_017005718.1 XM_017005719.1 XM_017005720.1 XM_017005721.1 XM_017005722.1 XM_017005723.1 XM_017005724.1

Gene Symbol
METTL6
Entrez Gene ID
131965
Gene Name
methyltransferase like 6
Gene Aliases
-
UniGene
Hs.149487
Gene Chromosome Location
Chr.3: 15381275-15427580 on Build GRCh38
Transcripts
NM_033083.6 XM_005265518.1 NM_001301790.1 NM_001301791.1 NM_001301792.1 NM_152396.3 XM_005264867.3 XM_005264870.4 XM_006712972.3 XM_011533357.2 XM_017005718.1 XM_017005719.1 XM_017005720.1 XM_017005721.1 XM_017005722.1 XM_017005723.1 XM_017005724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1204852 c.13G>A p.A5T Chr.3: 15427792-15427792 on Build GRCh38
445837 c.69G>A p.E23E Chr.3: 15427848-15427848 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1000349960 Chr.3: 15427608-15427608 on Build GRCh38
rs921903485 Chr.3: 15427679-15427679 on Build GRCh38
rs369264153 Chr.3: 15427916-15427916 on Build GRCh38
rs41284029 Chr.3: 15427876-15427876 on Build GRCh38
rs796703458 Chr.3: 15427540-15427540 on Build GRCh38
rs759842708 Chr.3: 15427749-15427749 on Build GRCh38
rs554297741 Chr.3: 15427661-15427661 on Build GRCh38
rs1000062227 Chr.3: 15427819-15427819 on Build GRCh38
rs139779980 Chr.3: 15427475-15427475 on Build GRCh38
rs367822381 Chr.3: 15427521-15427521 on Build GRCh38
rs889099199 Chr.3: 15427558-15427558 on Build GRCh38
rs548505049 Chr.3: 15427629-15427629 on Build GRCh38
rs747519706 Chr.3: 15427777-15427777 on Build GRCh38
rs1010756524 Chr.3: 15427821-15427821 on Build GRCh38
rs550106781 Chr.3: 15427600-15427600 on Build GRCh38
rs781405243 Chr.3: 15427808-15427808 on Build GRCh38
rs568592679 Chr.3: 15427658-15427658 on Build GRCh38
rs144383348 Chr.3: 15427498-15427498 on Build GRCh38
rs570893627 Chr.3: 15427687-15427687 on Build GRCh38
rs377132324 Chr.3: 15427497-15427497 on Build GRCh38
rs552476954 Chr.3: 15427584-15427584 on Build GRCh38
rs372062063 Chr.3: 15427751-15427751 on Build GRCh38
rs1034125193 Chr.3: 15427614-15427614 on Build GRCh38
rs370022222 Chr.3: 15427790-15427790 on Build GRCh38
rs775341002 Chr.3: 15427901-15427901 on Build GRCh38
rs761196948 Chr.3: 15427509-15427509 on Build GRCh38
rs1040334601 Chr.3: 15427744-15427744 on Build GRCh38
rs764049403 Chr.3: 15427576-15427576 on Build GRCh38
rs77636884 Chr.3: 15427860-15427860 on Build GRCh38
rs539822287 Chr.3: 15427711-15427711 on Build GRCh38
rs575466609 Chr.3: 15427791-15427791 on Build GRCh38
rs554473608 Chr.3: 15427873-15427873 on Build GRCh38
rs972272054 Chr.3: 15427897-15427897 on Build GRCh38
rs952413115 Chr.3: 15427917-15427917 on Build GRCh38
rs767213809 Chr.3: 15427631-15427631 on Build GRCh38
rs887551073 Chr.3: 15427560-15427560 on Build GRCh38
rs762752600 Chr.3: 15427877-15427877 on Build GRCh38
rs986609530 Chr.3: 15427695-15427695 on Build GRCh38
rs559766250 Chr.3: 15427527-15427527 on Build GRCh38
rs887460335 Chr.3: 15427478-15427478 on Build GRCh38
rs1024453239 Chr.3: 15427587-15427587 on Build GRCh38
rs993469476 Chr.3: 15427505-15427505 on Build GRCh38
rs376417139 Chr.3: 15427754-15427754 on Build GRCh38
rs1040648609 Chr.3: 15427462-15427463 on Build GRCh38
rs531963162 Chr.3: 15427577-15427577 on Build GRCh38
rs764077366 Chr.3: 15427908-15427908 on Build GRCh38
rs910610505 Chr.3: 15427724-15427724 on Build GRCh38
rs755228974 Chr.3: 15427741-15427741 on Build GRCh38
rs142360478 Chr.3: 15427914-15427914 on Build GRCh38
rs2290536 Chr.3: 15427494-15427494 on Build GRCh38
rs1020094108 Chr.3: 15427606-15427606 on Build GRCh38
rs980295541 Chr.3: 15427601-15427601 on Build GRCh38
rs769876525 Chr.3: 15427833-15427833 on Build GRCh38
rs1041694440 Chr.3: 15427796-15427796 on Build GRCh38
rs1036449833 Chr.3: 15427516-15427516 on Build GRCh38
rs1048786626 Chr.3: 15427545-15427545 on Build GRCh38
rs528665241 Chr.3: 15427585-15427585 on Build GRCh38
rs369500475 Chr.3: 15427581-15427581 on Build GRCh38
rs750301549 Chr.3: 15427643-15427643 on Build GRCh38
rs1020145125 Chr.3: 15427489-15427489 on Build GRCh38
rs548787128 Chr.3: 15427729-15427729 on Build GRCh38
rs931800620 Chr.3: 15427703-15427703 on Build GRCh38
rs925914840 Chr.3: 15427504-15427504 on Build GRCh38
rs760602660 Chr.3: 15427902-15427902 on Build GRCh38
rs142692553 Chr.3: 15427741-15427741 on Build GRCh38
rs900239790 Chr.3: 15427474-15427474 on Build GRCh38
rs534337132 Chr.3: 15427660-15427660 on Build GRCh38
rs530408990 Chr.3: 15427515-15427515 on Build GRCh38
rs958601728 Chr.3: 15427618-15427618 on Build GRCh38
rs115952434 Chr.3: 15427483-15427483 on Build GRCh38
rs904339006 Chr.3: 15427816-15427816 on Build GRCh38
rs978542025 Chr.3: 15427495-15427495 on Build GRCh38
rs950752454 Chr.3: 15427518-15427518 on Build GRCh38
rs146615412 Chr.3: 15427607-15427607 on Build GRCh38
rs892477686 Chr.3: 15427789-15427789 on Build GRCh38
rs966429544 Chr.3: 15427848-15427848 on Build GRCh38
rs748304883 Chr.3: 15427820-15427820 on Build GRCh38
rs976419413 Chr.3: 15427650-15427650 on Build GRCh38
rs764506619 Chr.3: 15427712-15427713 on Build GRCh38
rs569470477 Chr.3: 15427586-15427586 on Build GRCh38
rs930851861 Chr.3: 15427683-15427683 on Build GRCh38
rs1015039438 Chr.3: 15427578-15427578 on Build GRCh38
rs569693223 Chr.3: 15427735-15427735 on Build GRCh38
rs1010157962 Chr.3: 15427597-15427597 on Build GRCh38
rs972324246 Chr.3: 15427656-15427656 on Build GRCh38
rs1040169376 Chr.3: 15427522-15427522 on Build GRCh38
rs957479601 Chr.3: 15427609-15427609 on Build GRCh38
rs199920203 Chr.3: 15427763-15427763 on Build GRCh38
rs913143347 Chr.3: 15427512-15427512 on Build GRCh38
rs961727953 Chr.3: 15427892-15427892 on Build GRCh38
rs992484576 Chr.3: 15427619-15427619 on Build GRCh38
rs372055507 Chr.3: 15427715-15427715 on Build GRCh38
rs757012244 Chr.3: 15427589-15427589 on Build GRCh38
rs539896651 Chr.3: 15427501-15427501 on Build GRCh38
rs780508737 Chr.3: 15427895-15427896 on Build GRCh38
rs962236122 Chr.3: 15427647-15427647 on Build GRCh38
rs1050574931 Chr.3: 15427546-15427546 on Build GRCh38
rs1027254816 Chr.3: 15427476-15427476 on Build GRCh38

View Full Product Details