Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 15453649-15454181 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
COLQ
Entrez Gene ID
8292
Gene Name
collagen like tail subunit of asymmetric acetylcholinesterase
Gene Aliases
CMS5, EAD
UniGene
Hs.146735
Gene Chromosome Location
Chr.3: 15450133-15521751 on Build GRCh38
Transcripts
NM_005677.3 NM_080538.2 NM_080539.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4815044 c.1196A>C p.H399P Chr.3: 15453901-15453901 on Build GRCh38
3589018 c.1257C>T p.T419T Chr.3: 15453840-15453840 on Build GRCh38
3589019 c.1257C>T p.T419T Chr.3: 15453840-15453840 on Build GRCh38
4674129 c.1268+2T>C p.? Chr.3: 15453827-15453827 on Build GRCh38
6096759 c.1243G>A p.E415K Chr.3: 15453884-15453884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs142726065 Chr.3: 15454016-15454016 on Build GRCh38
rs954024724 Chr.3: 15454124-15454126 on Build GRCh38
rs756084974 Chr.3: 15453832-15453832 on Build GRCh38
rs764064240 Chr.3: 15453916-15453916 on Build GRCh38
rs776084775 Chr.3: 15454105-15454105 on Build GRCh38
rs779790449 Chr.3: 15453975-15453975 on Build GRCh38
rs75838637 Chr.3: 15453976-15453976 on Build GRCh38
rs1011603836 Chr.3: 15453980-15453980 on Build GRCh38
rs575113580 Chr.3: 15454002-15454002 on Build GRCh38
rs745502271 Chr.3: 15453849-15453849 on Build GRCh38
rs1030167814 Chr.3: 15454116-15454116 on Build GRCh38
rs771866680 Chr.3: 15453915-15453915 on Build GRCh38
rs776664665 Chr.3: 15453789-15453789 on Build GRCh38
rs758190106 Chr.3: 15453944-15453946 on Build GRCh38
rs983827998 Chr.3: 15453923-15453923 on Build GRCh38
rs755400333 Chr.3: 15453705-15453705 on Build GRCh38
rs781616949 Chr.3: 15453978-15453978 on Build GRCh38
rs753690228 Chr.3: 15453853-15453853 on Build GRCh38
rs769114264 Chr.3: 15453872-15453872 on Build GRCh38
rs141721093 Chr.3: 15453845-15453845 on Build GRCh38
rs745843788 Chr.3: 15453803-15453806 on Build GRCh38
rs3773459 Chr.3: 15454137-15454137 on Build GRCh38
rs769772442 Chr.3: 15453890-15453890 on Build GRCh38
rs536042715 Chr.3: 15453850-15453850 on Build GRCh38
rs121908923 Chr.3: 15453838-15453838 on Build GRCh38
rs752498913 Chr.3: 15453907-15453907 on Build GRCh38
rs1025361623 Chr.3: 15453898-15453898 on Build GRCh38
rs372048848 Chr.3: 15454108-15454108 on Build GRCh38
rs764641330 Chr.3: 15453946-15453946 on Build GRCh38
rs550616958 Chr.3: 15453841-15453841 on Build GRCh38
rs909917035 Chr.3: 15454133-15454133 on Build GRCh38
rs753990880 Chr.3: 15453671-15453671 on Build GRCh38
rs757958993 Chr.3: 15453966-15453966 on Build GRCh38
rs552643954 Chr.3: 15453727-15453727 on Build GRCh38
rs111472409 Chr.3: 15453703-15453703 on Build GRCh38
rs773394999 Chr.3: 15453894-15453894 on Build GRCh38
rs116629575 Chr.3: 15453678-15453678 on Build GRCh38
rs749999496 Chr.3: 15453948-15453948 on Build GRCh38
rs761916663 Chr.3: 15453790-15453790 on Build GRCh38
rs201104986 Chr.3: 15453779-15453779 on Build GRCh38
rs139574075 Chr.3: 15453899-15453899 on Build GRCh38
rs760567044 Chr.3: 15453915-15453915 on Build GRCh38
rs1021112406 Chr.3: 15453984-15453984 on Build GRCh38
rs753348773 Chr.3: 15453839-15453839 on Build GRCh38
rs946213053 Chr.3: 15454109-15454109 on Build GRCh38
rs1077827 Chr.3: 15453794-15453794 on Build GRCh38
rs532610252 Chr.3: 15453682-15453682 on Build GRCh38
rs750628758 Chr.3: 15453799-15453799 on Build GRCh38
rs185829251 Chr.3: 15453846-15453846 on Build GRCh38
rs374059685 Chr.3: 15453824-15453824 on Build GRCh38
rs780011630 Chr.3: 15453936-15453936 on Build GRCh38
rs3773458 Chr.3: 15454147-15454147 on Build GRCh38
rs985410979 Chr.3: 15454130-15454130 on Build GRCh38
rs964361808 Chr.3: 15454012-15454012 on Build GRCh38
rs36125649 Chr.3: 15453812-15453813 on Build GRCh38
rs375272767 Chr.3: 15453931-15453931 on Build GRCh38
rs777147864 Chr.3: 15453873-15453873 on Build GRCh38
rs746655707 Chr.3: 15454033-15454033 on Build GRCh38
rs371351288 Chr.3: 15453822-15453822 on Build GRCh38
rs770615065 Chr.3: 15454038-15454038 on Build GRCh38
rs558548890 Chr.3: 15453977-15453977 on Build GRCh38
rs753830857 Chr.3: 15453918-15453918 on Build GRCh38
rs55866379 Chr.3: 15453879-15453879 on Build GRCh38
rs560658151 Chr.3: 15454153-15454153 on Build GRCh38
rs140670616 Chr.3: 15453922-15453922 on Build GRCh38
rs763299098 Chr.3: 15453800-15453800 on Build GRCh38
rs71308146 Chr.3: 15453795-15453795 on Build GRCh38
rs759121012 Chr.3: 15454119-15454119 on Build GRCh38
rs3773457 Chr.3: 15454152-15454152 on Build GRCh38
rs769202297 Chr.3: 15453780-15453780 on Build GRCh38

View Full Product Details