Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 19905434-19905955 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EFHB
Entrez Gene ID
151651
Gene Name
EF-hand domain family member B
Gene Aliases
CFAP21
UniGene
Hs.733920
Gene Chromosome Location
Chr.3: 19879472-19947036 on Build GRCh38
Transcripts
NM_144715.3 XM_005264889.1 XM_011533382.1 XM_011533383.2 XM_011533385.2 XM_017005741.1 XM_017005742.1 XM_017005743.1 XM_017005744.1 XM_017005745.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1566834 c.1417A>T p.M473L Chr.3: 19905621-19905621 on Build GRCh38
1043259 c.1390T>C p.S464P Chr.3: 19905648-19905648 on Build GRCh38
1043262 c.1339G>T p.V447L Chr.3: 19905699-19905699 on Build GRCh38
7229274 c.1418+7T>G p.? Chr.3: 19905613-19905613 on Build GRCh38
3915675 c.1376G>A p.R459Q Chr.3: 19905662-19905662 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1030094622 Chr.3: 19905864-19905864 on Build GRCh38
rs768838478 Chr.3: 19905672-19905672 on Build GRCh38
rs775421555 Chr.3: 19905667-19905667 on Build GRCh38
rs760754526 Chr.3: 19905596-19905596 on Build GRCh38
rs764360070 Chr.3: 19905603-19905603 on Build GRCh38
rs926322044 Chr.3: 19905892-19905892 on Build GRCh38
rs539999953 Chr.3: 19905525-19905525 on Build GRCh38
rs988095425 Chr.3: 19905556-19905556 on Build GRCh38
rs747185043 Chr.3: 19905668-19905668 on Build GRCh38
rs770058952 Chr.3: 19905796-19905796 on Build GRCh38
rs763239774 Chr.3: 19905822-19905822 on Build GRCh38
rs879779375 Chr.3: 19905476-19905476 on Build GRCh38
rs757647822 Chr.3: 19905663-19905663 on Build GRCh38
rs528567885 Chr.3: 19905878-19905878 on Build GRCh38
rs754943518 Chr.3: 19905637-19905637 on Build GRCh38
rs762049673 Chr.3: 19905696-19905696 on Build GRCh38
rs573725901 Chr.3: 19905462-19905462 on Build GRCh38
rs375643086 Chr.3: 19905789-19905789 on Build GRCh38
rs762782890 Chr.3: 19905612-19905612 on Build GRCh38
rs766878836 Chr.3: 19905638-19905638 on Build GRCh38
rs757258386 Chr.3: 19905921-19905921 on Build GRCh38
rs532694286 Chr.3: 19905790-19905790 on Build GRCh38
rs371075148 Chr.3: 19905735-19905735 on Build GRCh38
rs768678839 Chr.3: 19905779-19905779 on Build GRCh38
rs746506274 Chr.3: 19905756-19905756 on Build GRCh38
rs763720167 Chr.3: 19905716-19905716 on Build GRCh38
rs753504094 Chr.3: 19905724-19905724 on Build GRCh38
rs913531723 Chr.3: 19905558-19905558 on Build GRCh38
rs759377531 Chr.3: 19905707-19905707 on Build GRCh38
rs201511973 Chr.3: 19905660-19905660 on Build GRCh38
rs771534517 Chr.3: 19905570-19905570 on Build GRCh38
rs377473787 Chr.3: 19905698-19905698 on Build GRCh38
rs1056356879 Chr.3: 19905662-19905662 on Build GRCh38
rs755715054 Chr.3: 19905643-19905643 on Build GRCh38
rs948174235 Chr.3: 19905479-19905479 on Build GRCh38
rs751408991 Chr.3: 19905632-19905632 on Build GRCh38
rs751679135 Chr.3: 19905902-19905902 on Build GRCh38
rs752117290 Chr.3: 19905711-19905711 on Build GRCh38
rs777606861 Chr.3: 19905658-19905658 on Build GRCh38
rs911756326 Chr.3: 19905523-19905523 on Build GRCh38
rs2929377 Chr.3: 19905804-19905804 on Build GRCh38
rs796986606 Chr.3: 19905559-19905559 on Build GRCh38
rs760019193 Chr.3: 19905666-19905666 on Build GRCh38
rs775647860 Chr.3: 19905592-19905592 on Build GRCh38
rs757957447 Chr.3: 19905578-19905598 on Build GRCh38
rs768138337 Chr.3: 19905490-19905490 on Build GRCh38
rs759415846 Chr.3: 19905797-19905797 on Build GRCh38
rs747572256 Chr.3: 19905509-19905509 on Build GRCh38
rs776781361 Chr.3: 19905677-19905677 on Build GRCh38
rs986349176 Chr.3: 19905512-19905512 on Build GRCh38
rs954919755 Chr.3: 19905615-19905615 on Build GRCh38
rs187844318 Chr.3: 19905544-19905544 on Build GRCh38
rs1015543183 Chr.3: 19905915-19905916 on Build GRCh38
rs756976123 Chr.3: 19905729-19905729 on Build GRCh38
rs963501088 Chr.3: 19905930-19905930 on Build GRCh38
rs192668366 Chr.3: 19905897-19905897 on Build GRCh38
rs114559064 Chr.3: 19905565-19905565 on Build GRCh38
rs2931394 Chr.3: 19905500-19905500 on Build GRCh38
rs780654338 Chr.3: 19905748-19905748 on Build GRCh38
rs965908922 Chr.3: 19905782-19905782 on Build GRCh38
rs772990199 Chr.3: 19905611-19905611 on Build GRCh38
rs146363233 Chr.3: 19905721-19905721 on Build GRCh38
rs544501678 Chr.3: 19905489-19905489 on Build GRCh38
rs566303955 Chr.3: 19905816-19905816 on Build GRCh38
rs148913630 Chr.3: 19905480-19905481 on Build GRCh38
rs895149620 Chr.3: 19905478-19905478 on Build GRCh38
rs774205612 Chr.3: 19905704-19905704 on Build GRCh38
rs747490154 Chr.3: 19905775-19905775 on Build GRCh38
rs938538232 Chr.3: 19905752-19905752 on Build GRCh38
rs142412525 Chr.3: 19905734-19905734 on Build GRCh38
rs66526230 Chr.3: 19905613-19905613 on Build GRCh38
rs775049447 Chr.3: 19905582-19905582 on Build GRCh38
rs758829170 Chr.3: 19905746-19905746 on Build GRCh38
rs746324633 Chr.3: 19905573-19905574 on Build GRCh38
rs779469960 Chr.3: 19905617-19905618 on Build GRCh38
rs777255302 Chr.3: 19905664-19905664 on Build GRCh38
rs898111938 Chr.3: 19905697-19905697 on Build GRCh38
rs531632963 Chr.3: 19905714-19905714 on Build GRCh38
rs560592013 Chr.3: 19905587-19905587 on Build GRCh38
rs767557048 Chr.3: 19905709-19905709 on Build GRCh38
rs146671119 Chr.3: 19905641-19905641 on Build GRCh38
rs766093282 Chr.3: 19905702-19905702 on Build GRCh38
rs752554055 Chr.3: 19905709-19905709 on Build GRCh38
rs755825347 Chr.3: 19905706-19905706 on Build GRCh38
rs1015190460 Chr.3: 19905691-19905691 on Build GRCh38
rs964364734 Chr.3: 19905591-19905591 on Build GRCh38

View Full Product Details