Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 28324364-28324868 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AZI2
Entrez Gene ID
64343
Gene Name
5-azacytidine induced 2
Gene Aliases
AZ2, NAP1, TILP
UniGene
Hs.706676
Gene Chromosome Location
Chr.3: 28322353-28349129 on Build GRCh38
Transcripts
NM_001271650.1 NM_022461.4 XM_005265392.2 XM_011534009.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4653680 c.772G>T p.A258S Chr.3: 28324449-28324449 on Build GRCh38
5756256 c.826A>C p.M276L Chr.3: 28324395-28324395 on Build GRCh38
6609177 c.809G>A p.S270N Chr.3: 28324412-28324412 on Build GRCh38
1422283 c.832T>G p.F278V Chr.3: 28324389-28324389 on Build GRCh38
6268245 c.810C>A p.S270R Chr.3: 28324411-28324411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749672598 Chr.3: 28324449-28324449 on Build GRCh38
rs529303922 Chr.3: 28324766-28324766 on Build GRCh38
rs775767513 Chr.3: 28324669-28324669 on Build GRCh38
rs906316414 Chr.3: 28324830-28324830 on Build GRCh38
rs34113092 Chr.3: 28324395-28324396 on Build GRCh38
rs954312117 Chr.3: 28324577-28324577 on Build GRCh38
rs998449701 Chr.3: 28324540-28324540 on Build GRCh38
rs971008807 Chr.3: 28324681-28324683 on Build GRCh38
rs766146944 Chr.3: 28324404-28324404 on Build GRCh38
rs576017309 Chr.3: 28324495-28324495 on Build GRCh38
rs373443811 Chr.3: 28324483-28324483 on Build GRCh38
rs763711823 Chr.3: 28324402-28324403 on Build GRCh38
rs75035657 Chr.3: 28324619-28324619 on Build GRCh38
rs201784193 Chr.3: 28324438-28324438 on Build GRCh38
rs757646681 Chr.3: 28324442-28324442 on Build GRCh38
rs1032988943 Chr.3: 28324595-28324595 on Build GRCh38
rs1003891547 Chr.3: 28324512-28324512 on Build GRCh38
rs567741514 Chr.3: 28324441-28324441 on Build GRCh38
rs369262666 Chr.3: 28324469-28324469 on Build GRCh38
rs762488876 Chr.3: 28324394-28324394 on Build GRCh38
rs370962571 Chr.3: 28324704-28324704 on Build GRCh38
rs772925245 Chr.3: 28324393-28324393 on Build GRCh38
rs901313419 Chr.3: 28324525-28324525 on Build GRCh38
rs560820862 Chr.3: 28324738-28324738 on Build GRCh38
rs35179283 Chr.3: 28324506-28324506 on Build GRCh38
rs760748005 Chr.3: 28324806-28324806 on Build GRCh38
rs754717597 Chr.3: 28324388-28324388 on Build GRCh38
rs779033447 Chr.3: 28324464-28324464 on Build GRCh38
rs35779568 Chr.3: 28324447-28324447 on Build GRCh38
rs34458963 Chr.3: 28324416-28324416 on Build GRCh38
rs773709190 Chr.3: 28324505-28324505 on Build GRCh38
rs546597949 Chr.3: 28324421-28324421 on Build GRCh38
rs191458065 Chr.3: 28324656-28324656 on Build GRCh38
rs201895936 Chr.3: 28324840-28324840 on Build GRCh38
rs1029030866 Chr.3: 28324542-28324542 on Build GRCh38
rs531092464 Chr.3: 28324795-28324796 on Build GRCh38
rs999682325 Chr.3: 28324833-28324833 on Build GRCh38
rs912649003 Chr.3: 28324587-28324587 on Build GRCh38
rs199951188 Chr.3: 28324472-28324472 on Build GRCh38
rs981962338 Chr.3: 28324693-28324693 on Build GRCh38
rs1048065379 Chr.3: 28324759-28324759 on Build GRCh38
rs35055681 Chr.3: 28324770-28324770 on Build GRCh38
rs772272115 Chr.3: 28324590-28324590 on Build GRCh38
rs1046514926 Chr.3: 28324761-28324761 on Build GRCh38
rs1037069360 Chr.3: 28324603-28324603 on Build GRCh38
rs375459126 Chr.3: 28324452-28324452 on Build GRCh38
rs1012373443 Chr.3: 28324742-28324742 on Build GRCh38
rs35793201 Chr.3: 28324796-28324796 on Build GRCh38
rs1048048575 Chr.3: 28324471-28324471 on Build GRCh38
rs562082194 Chr.3: 28324514-28324514 on Build GRCh38
rs35465423 Chr.3: 28324465-28324465 on Build GRCh38
rs757277376 Chr.3: 28324459-28324459 on Build GRCh38
rs931604969 Chr.3: 28324621-28324621 on Build GRCh38
rs545192948 Chr.3: 28324578-28324578 on Build GRCh38
rs377409936 Chr.3: 28324466-28324466 on Build GRCh38
rs779589060 Chr.3: 28324493-28324493 on Build GRCh38
rs186663592 Chr.3: 28324601-28324601 on Build GRCh38
rs35565231 Chr.3: 28324495-28324496 on Build GRCh38
rs35669786 Chr.3: 28324745-28324745 on Build GRCh38
rs937921871 Chr.3: 28324755-28324757 on Build GRCh38
rs767970338 Chr.3: 28324434-28324434 on Build GRCh38

View Full Product Details