Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32525956-32526462 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DYNC1LI1
Entrez Gene ID
51143
Gene Name
dynein cytoplasmic 1 light intermediate chain 1
Gene Aliases
DLC-A, DNCLI1, LIC1
UniGene
Hs.529495
Gene Chromosome Location
Chr.3: 32525971-32570874 on Build GRCh38
Transcripts
NM_016141.3

SNP Detail

SNP ID Chromosome Location
rs970559439 Chr.3: 32526056-32526056 on Build GRCh38
rs943095614 Chr.3: 32526017-32526017 on Build GRCh38
rs891710686 Chr.3: 32526343-32526343 on Build GRCh38
rs963221702 Chr.3: 32526199-32526199 on Build GRCh38
rs1012596251 Chr.3: 32526045-32526045 on Build GRCh38
rs866485548 Chr.3: 32526310-32526310 on Build GRCh38
rs925670800 Chr.3: 32526296-32526296 on Build GRCh38
rs905997580 Chr.3: 32526138-32526138 on Build GRCh38
rs906490399 Chr.3: 32526242-32526242 on Build GRCh38
rs904345775 Chr.3: 32526022-32526022 on Build GRCh38
rs541050393 Chr.3: 32526282-32526282 on Build GRCh38
rs182818570 Chr.3: 32526294-32526294 on Build GRCh38
rs974810147 Chr.3: 32526299-32526299 on Build GRCh38
rs935848242 Chr.3: 32526307-32526307 on Build GRCh38
rs1024220445 Chr.3: 32526098-32526098 on Build GRCh38
rs972896644 Chr.3: 32526201-32526201 on Build GRCh38
rs77147332 Chr.3: 32526270-32526270 on Build GRCh38
rs965075523 Chr.3: 32526297-32526300 on Build GRCh38
rs116363983 Chr.3: 32526079-32526079 on Build GRCh38
rs773409350 Chr.3: 32526277-32526277 on Build GRCh38
rs76466472 Chr.3: 32526054-32526054 on Build GRCh38
rs950813602 Chr.3: 32526077-32526077 on Build GRCh38
rs1025505674 Chr.3: 32526257-32526257 on Build GRCh38
rs866394701 Chr.3: 32526078-32526080 on Build GRCh38
rs60896336 Chr.3: 32526080-32526081 on Build GRCh38
rs987579358 Chr.3: 32526243-32526243 on Build GRCh38
rs553600006 Chr.3: 32526267-32526267 on Build GRCh38
rs537052735 Chr.3: 32526101-32526101 on Build GRCh38
rs558043870 Chr.3: 32526079-32526080 on Build GRCh38
rs10662666 Chr.3: 32526062-32526063 on Build GRCh38
rs567907980 Chr.3: 32526080-32526080 on Build GRCh38
rs905860310 Chr.3: 32526417-32526417 on Build GRCh38
rs768830156 Chr.3: 32526150-32526150 on Build GRCh38
rs972524550 Chr.3: 32526075-32526076 on Build GRCh38
rs755755225 Chr.3: 32526064-32526065 on Build GRCh38
rs933572204 Chr.3: 32526106-32526108 on Build GRCh38
rs796926783 Chr.3: 32526063-32526064 on Build GRCh38
rs1038918803 Chr.3: 32526020-32526020 on Build GRCh38
rs1001567336 Chr.3: 32526146-32526146 on Build GRCh38
rs994079295 Chr.3: 32526246-32526246 on Build GRCh38
rs1025481328 Chr.3: 32526077-32526078 on Build GRCh38
rs1053518820 Chr.3: 32526189-32526189 on Build GRCh38
rs200314394 Chr.3: 32526095-32526096 on Build GRCh38
rs936442779 Chr.3: 32526177-32526177 on Build GRCh38
rs557806991 Chr.3: 32526315-32526315 on Build GRCh38
rs1026323516 Chr.3: 32526227-32526227 on Build GRCh38
rs1053467195 Chr.3: 32526060-32526060 on Build GRCh38
rs1014679796 Chr.3: 32526084-32526084 on Build GRCh38
rs78533317 Chr.3: 32526371-32526371 on Build GRCh38
rs1045744753 Chr.3: 32526397-32526397 on Build GRCh38
rs568114317 Chr.3: 32525983-32525983 on Build GRCh38
rs1032453425 Chr.3: 32526032-32526034 on Build GRCh38
rs975405744 Chr.3: 32526095-32526095 on Build GRCh38
rs935958176 Chr.3: 32526041-32526041 on Build GRCh38
rs1011881452 Chr.3: 32526229-32526229 on Build GRCh38
rs1052925858 Chr.3: 32526318-32526318 on Build GRCh38
rs115678103 Chr.3: 32526235-32526235 on Build GRCh38
rs761063425 Chr.3: 32526314-32526314 on Build GRCh38

View Full Product Details