Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32703841-32704343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNOT10
Entrez Gene ID
25904
Gene Name
CCR4-NOT transcription complex subunit 10
Gene Aliases
-
UniGene
Hs.444851
Gene Chromosome Location
Chr.3: 32685145-32773875 on Build GRCh38
Transcripts
NM_001256741.1 NM_001256742.1 NM_015442.2 NR_046352.1 XM_006713084.3 XM_006713085.3 XM_011533566.2 XM_017006109.1 XM_017006110.1 XM_017006111.1 XM_017006112.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6097484 c.297+1G>T p.? Chr.3: 32703963-32703963 on Build GRCh38
1043658 c.37A>G p.K13E Chr.3: 32703882-32703882 on Build GRCh38
7182156 c.51A>G p.T17T Chr.3: 32703896-32703896 on Build GRCh38
5528834 c.59C>T p.S20F Chr.3: 32703904-32703904 on Build GRCh38
5528835 c.239C>T p.S80F Chr.3: 32703904-32703904 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs901305806 Chr.3: 32704045-32704045 on Build GRCh38
rs542693660 Chr.3: 32704047-32704047 on Build GRCh38
rs983882884 Chr.3: 32704082-32704082 on Build GRCh38
rs777377542 Chr.3: 32703904-32703904 on Build GRCh38
rs200470162 Chr.3: 32704129-32704129 on Build GRCh38
rs1028740214 Chr.3: 32704064-32704064 on Build GRCh38
rs907929870 Chr.3: 32704089-32704089 on Build GRCh38
rs561414373 Chr.3: 32704069-32704069 on Build GRCh38
rs932795696 Chr.3: 32704310-32704310 on Build GRCh38
rs772429666 Chr.3: 32703935-32703935 on Build GRCh38
rs368778952 Chr.3: 32703916-32703916 on Build GRCh38
rs374466072 Chr.3: 32703886-32703886 on Build GRCh38
rs6766872 Chr.3: 32704304-32704304 on Build GRCh38
rs767447727 Chr.3: 32704005-32704005 on Build GRCh38
rs916033802 Chr.3: 32703944-32703944 on Build GRCh38
rs756985576 Chr.3: 32703917-32703917 on Build GRCh38
rs1041298686 Chr.3: 32704298-32704298 on Build GRCh38
rs1001732035 Chr.3: 32704315-32704315 on Build GRCh38
rs372155147 Chr.3: 32703943-32703943 on Build GRCh38
rs959895821 Chr.3: 32704165-32704165 on Build GRCh38
rs763213881 Chr.3: 32703989-32703989 on Build GRCh38
rs751867812 Chr.3: 32703972-32703972 on Build GRCh38
rs908509930 Chr.3: 32704261-32704261 on Build GRCh38
rs138163550 Chr.3: 32704140-32704140 on Build GRCh38
rs1011230223 Chr.3: 32704111-32704111 on Build GRCh38
rs1021761976 Chr.3: 32704299-32704299 on Build GRCh38
rs755958646 Chr.3: 32703899-32703899 on Build GRCh38
rs748152507 Chr.3: 32703979-32703979 on Build GRCh38
rs148956635 Chr.3: 32703877-32703877 on Build GRCh38
rs931857927 Chr.3: 32704131-32704131 on Build GRCh38
rs980352642 Chr.3: 32704026-32704026 on Build GRCh38
rs942123864 Chr.3: 32704110-32704110 on Build GRCh38
rs991314335 Chr.3: 32704218-32704218 on Build GRCh38
rs776809482 Chr.3: 32703978-32703978 on Build GRCh38
rs764404030 Chr.3: 32703872-32703872 on Build GRCh38
rs761232256 Chr.3: 32703956-32703956 on Build GRCh38
rs540296411 Chr.3: 32703947-32703947 on Build GRCh38
rs371171751 Chr.3: 32703921-32703921 on Build GRCh38
rs200738944 Chr.3: 32703974-32703974 on Build GRCh38
rs893526621 Chr.3: 32704276-32704276 on Build GRCh38
rs775923795 Chr.3: 32703907-32703915 on Build GRCh38
rs915982887 Chr.3: 32704225-32704225 on Build GRCh38
rs748947629 Chr.3: 32704305-32704305 on Build GRCh38
rs201152453 Chr.3: 32703962-32703962 on Build GRCh38
rs746298388 Chr.3: 32703932-32703932 on Build GRCh38
rs977332333 Chr.3: 32704130-32704130 on Build GRCh38
rs112206246 Chr.3: 32704121-32704121 on Build GRCh38
rs1041704791 Chr.3: 32704024-32704024 on Build GRCh38
rs750963293 Chr.3: 32704211-32704211 on Build GRCh38
rs563487079 Chr.3: 32703874-32703874 on Build GRCh38
rs756509702 Chr.3: 32704026-32704033 on Build GRCh38
rs757387739 Chr.3: 32703982-32703982 on Build GRCh38
rs76808846 Chr.3: 32703983-32703983 on Build GRCh38
rs931420987 Chr.3: 32704233-32704233 on Build GRCh38
rs940021641 Chr.3: 32704285-32704285 on Build GRCh38
rs984186822 Chr.3: 32704256-32704256 on Build GRCh38

View Full Product Details