Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32951489-32951993 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR4
Entrez Gene ID
1233
Gene Name
C-C motif chemokine receptor 4
Gene Aliases
CC-CKR-4, CD194, CKR4, CMKBR4, ChemR13, HGCN:14099, K5-5
UniGene
Hs.184926
Gene Chromosome Location
Chr.3: 32951555-32955312 on Build GRCh38
Transcripts
NM_005508.4 XM_017005687.1

SNP Detail

SNP ID Chromosome Location
rs1043872776 Chr.3: 32951520-32951520 on Build GRCh38
rs376708122 Chr.3: 32951766-32951766 on Build GRCh38
rs898344580 Chr.3: 32951819-32951819 on Build GRCh38
rs563972381 Chr.3: 32951732-32951732 on Build GRCh38
rs1024544192 Chr.3: 32951765-32951765 on Build GRCh38
rs866455287 Chr.3: 32951615-32951615 on Build GRCh38
rs891471870 Chr.3: 32951607-32951607 on Build GRCh38
rs77890524 Chr.3: 32951875-32951876 on Build GRCh38
rs913825852 Chr.3: 32951860-32951860 on Build GRCh38
rs183147260 Chr.3: 32951878-32951878 on Build GRCh38
rs747326687 Chr.3: 32951522-32951522 on Build GRCh38
rs866628244 Chr.3: 32951944-32951944 on Build GRCh38
rs925460184 Chr.3: 32951718-32951718 on Build GRCh38
rs1045801851 Chr.3: 32951738-32951738 on Build GRCh38
rs891519715 Chr.3: 32951728-32951728 on Build GRCh38
rs1013959027 Chr.3: 32951737-32951737 on Build GRCh38
rs1043924713 Chr.3: 32951713-32951713 on Build GRCh38
rs993989377 Chr.3: 32951821-32951821 on Build GRCh38
rs952175615 Chr.3: 32951781-32951781 on Build GRCh38
rs955430746 Chr.3: 32951667-32951667 on Build GRCh38
rs972463560 Chr.3: 32951597-32951597 on Build GRCh38
rs535198674 Chr.3: 32951639-32951639 on Build GRCh38
rs553324182 Chr.3: 32951656-32951656 on Build GRCh38
rs970376522 Chr.3: 32951613-32951613 on Build GRCh38
rs1043749335 Chr.3: 32951673-32951673 on Build GRCh38
rs187241746 Chr.3: 32951611-32951611 on Build GRCh38
rs533489555 Chr.3: 32951643-32951643 on Build GRCh38
rs1014322172 Chr.3: 32951608-32951608 on Build GRCh38
rs1052777406 Chr.3: 32951606-32951606 on Build GRCh38
rs1018701975 Chr.3: 32951934-32951934 on Build GRCh38
rs892380395 Chr.3: 32951655-32951655 on Build GRCh38
rs192508671 Chr.3: 32951753-32951753 on Build GRCh38
rs979664277 Chr.3: 32951967-32951967 on Build GRCh38
rs142023895 Chr.3: 32951727-32951727 on Build GRCh38
rs987189280 Chr.3: 32951694-32951694 on Build GRCh38
rs1016837777 Chr.3: 32951794-32951794 on Build GRCh38
rs760600665 Chr.3: 32951572-32951572 on Build GRCh38
rs903611966 Chr.3: 32951708-32951708 on Build GRCh38
rs1028034554 Chr.3: 32951780-32951780 on Build GRCh38
rs920728754 Chr.3: 32951606-32951606 on Build GRCh38
rs975911889 Chr.3: 32951853-32951853 on Build GRCh38
rs748903545 Chr.3: 32951850-32951850 on Build GRCh38
rs962497486 Chr.3: 32951629-32951629 on Build GRCh38
rs781423136 Chr.3: 32951750-32951750 on Build GRCh38
rs955642064 Chr.3: 32951885-32951885 on Build GRCh38
rs781421722 Chr.3: 32951900-32951900 on Build GRCh38
rs1025751615 Chr.3: 32951832-32951832 on Build GRCh38
rs1001455593 Chr.3: 32951758-32951758 on Build GRCh38
rs555430404 Chr.3: 32951590-32951590 on Build GRCh38
rs578225539 Chr.3: 32951702-32951702 on Build GRCh38
rs942949150 Chr.3: 32951703-32951703 on Build GRCh38
rs962045826 Chr.3: 32951594-32951594 on Build GRCh38
rs1016785406 Chr.3: 32951625-32951625 on Build GRCh38
rs958962012 Chr.3: 32951524-32951524 on Build GRCh38
rs993515381 Chr.3: 32951772-32951772 on Build GRCh38
rs71903231 Chr.3: 32951608-32951608 on Build GRCh38
rs575667344 Chr.3: 32951771-32951771 on Build GRCh38
rs935470885 Chr.3: 32951724-32951724 on Build GRCh38
rs764970197 Chr.3: 32951741-32951741 on Build GRCh38

View Full Product Details