Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 32954485-32954997 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCR4
Entrez Gene ID
1233
Gene Name
C-C motif chemokine receptor 4
Gene Aliases
CC-CKR-4, CD194, CKR4, CMKBR4, ChemR13, HGCN:14099, K5-5
UniGene
Hs.184926
Gene Chromosome Location
Chr.3: 32951555-32955312 on Build GRCh38
Transcripts
NM_005508.4 XM_017005687.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5344454 c.1065T>C p.D355D Chr.3: 32954487-32954487 on Build GRCh38
4957947 c.1071T>C p.H357H Chr.3: 32954493-32954493 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs542846538 Chr.3: 32954584-32954584 on Build GRCh38
rs951512746 Chr.3: 32954755-32954757 on Build GRCh38
rs532415931 Chr.3: 32954932-32954932 on Build GRCh38
rs139563771 Chr.3: 32954691-32954691 on Build GRCh38
rs967018949 Chr.3: 32954951-32954951 on Build GRCh38
rs1021265701 Chr.3: 32954640-32954640 on Build GRCh38
rs990870151 Chr.3: 32954723-32954723 on Build GRCh38
rs188417194 Chr.3: 32954881-32954881 on Build GRCh38
rs41321952 Chr.3: 32954931-32954931 on Build GRCh38
rs879205515 Chr.3: 32954939-32954939 on Build GRCh38
rs368309911 Chr.3: 32954795-32954795 on Build GRCh38
rs369230735 Chr.3: 32954828-32954828 on Build GRCh38
rs908106970 Chr.3: 32954787-32954787 on Build GRCh38
rs1051168096 Chr.3: 32954516-32954516 on Build GRCh38
rs373086385 Chr.3: 32954539-32954539 on Build GRCh38
rs371345875 Chr.3: 32954863-32954863 on Build GRCh38
rs569360863 Chr.3: 32954933-32954933 on Build GRCh38
rs959448877 Chr.3: 32954722-32954722 on Build GRCh38
rs557455406 Chr.3: 32954583-32954584 on Build GRCh38
rs777721930 Chr.3: 32954809-32954809 on Build GRCh38
rs957666853 Chr.3: 32954582-32954582 on Build GRCh38
rs149753854 Chr.3: 32954721-32954721 on Build GRCh38
rs1020897856 Chr.3: 32954936-32954936 on Build GRCh38
rs550654769 Chr.3: 32954716-32954716 on Build GRCh38
rs552472728 Chr.3: 32954930-32954930 on Build GRCh38
rs774882827 Chr.3: 32954543-32954543 on Build GRCh38
rs752991634 Chr.3: 32954569-32954569 on Build GRCh38
rs1048001077 Chr.3: 32954778-32954778 on Build GRCh38
rs144635444 Chr.3: 32954760-32954760 on Build GRCh38
rs1034861978 Chr.3: 32954973-32954973 on Build GRCh38
rs532149106 Chr.3: 32954708-32954708 on Build GRCh38
rs530229714 Chr.3: 32954748-32954748 on Build GRCh38
rs776312559 Chr.3: 32954524-32954524 on Build GRCh38
rs374130488 Chr.3: 32954511-32954511 on Build GRCh38
rs1023883001 Chr.3: 32954692-32954692 on Build GRCh38
rs1003504908 Chr.3: 32954710-32954710 on Build GRCh38
rs996559209 Chr.3: 32954859-32954859 on Build GRCh38
rs374947742 Chr.3: 32954630-32954630 on Build GRCh38
rs758075435 Chr.3: 32954964-32954964 on Build GRCh38
rs186322317 Chr.3: 32954805-32954805 on Build GRCh38
rs750113475 Chr.3: 32954786-32954786 on Build GRCh38
rs998988596 Chr.3: 32954592-32954592 on Build GRCh38
rs575302301 Chr.3: 32954526-32954526 on Build GRCh38
rs747783621 Chr.3: 32954525-32954525 on Build GRCh38
rs773569364 Chr.3: 32954527-32954527 on Build GRCh38
rs561020992 Chr.3: 32954596-32954596 on Build GRCh38
rs975241307 Chr.3: 32954880-32954880 on Build GRCh38
rs150574084 Chr.3: 32954642-32954642 on Build GRCh38
rs761298486 Chr.3: 32954563-32954563 on Build GRCh38
rs1011024123 Chr.3: 32954672-32954672 on Build GRCh38
rs557532417 Chr.3: 32954558-32954558 on Build GRCh38
rs900639747 Chr.3: 32954852-32954852 on Build GRCh38
rs770867629 Chr.3: 32954671-32954671 on Build GRCh38
rs1040940123 Chr.3: 32954841-32954841 on Build GRCh38
rs1003425078 Chr.3: 32954956-32954956 on Build GRCh38
rs1054193471 Chr.3: 32954616-32954616 on Build GRCh38
rs529277420 Chr.3: 32954811-32954811 on Build GRCh38

View Full Product Details