Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33496167-33496670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASP2
Entrez Gene ID
23122
Gene Name
cytoplasmic linker associated protein 2
Gene Aliases
-
UniGene
Hs.108614
Gene Chromosome Location
Chr.3: 33496245-33718356 on Build GRCh38
Transcripts
NM_001207044.1 NM_015097.2 XM_006713040.1 XM_006713041.1 XM_006713042.1 XM_006713048.1 XM_006713049.1 XM_006713050.1 XM_006713052.1 XM_006713053.1 XM_011533515.1 XM_017005947.1 XM_017005948.1 XM_017005949.1 XM_017005950.1 XM_017005951.1 XM_017005952.1 XM_017005953.1 XM_017005954.1 XM_017005955.1 XM_017005956.1 XM_017005957.1 XM_017005958.1 XM_017005959.1 XM_017005960.1 XM_017005961.1 XM_017005962.1 XM_017005963.1 XM_017005964.1 XM_017005965.1 XM_017005966.1 XM_017005967.1 XM_017005968.1 XM_017005969.1 XM_017005970.1 XM_017005971.1 XM_017005972.1 XM_017005973.1 XM_017005974.1 XM_017005975.1 XM_017005976.1 XM_017005977.1 XM_017005978.1 XM_017005979.1 XM_017005980.1 XM_017005981.1 XM_017005982.1 XM_017005983.1 XM_017005984.1 XM_017005985.1 XM_017005986.1

SNP Detail

SNP ID Chromosome Location
rs4679035 Chr.3: 33496221-33496221 on Build GRCh38
rs1038372474 Chr.3: 33496250-33496250 on Build GRCh38
rs568594459 Chr.3: 33496434-33496434 on Build GRCh38
rs913498291 Chr.3: 33496461-33496461 on Build GRCh38
rs1008742643 Chr.3: 33496341-33496341 on Build GRCh38
rs901807464 Chr.3: 33496608-33496608 on Build GRCh38
rs778501352 Chr.3: 33496475-33496476 on Build GRCh38
rs866528427 Chr.3: 33496626-33496626 on Build GRCh38
rs974400043 Chr.3: 33496432-33496432 on Build GRCh38
rs1009516377 Chr.3: 33496615-33496615 on Build GRCh38
rs547107701 Chr.3: 33496531-33496531 on Build GRCh38
rs1046203558 Chr.3: 33496214-33496214 on Build GRCh38
rs994740882 Chr.3: 33496281-33496281 on Build GRCh38
rs899078767 Chr.3: 33496279-33496279 on Build GRCh38
rs1001903268 Chr.3: 33496233-33496233 on Build GRCh38
rs889023836 Chr.3: 33496280-33496280 on Build GRCh38
rs949700572 Chr.3: 33496456-33496456 on Build GRCh38
rs968572260 Chr.3: 33496401-33496401 on Build GRCh38
rs181036567 Chr.3: 33496599-33496599 on Build GRCh38
rs1040270392 Chr.3: 33496582-33496582 on Build GRCh38
rs930416188 Chr.3: 33496488-33496488 on Build GRCh38
rs900238513 Chr.3: 33496642-33496642 on Build GRCh38
rs1048867446 Chr.3: 33496517-33496517 on Build GRCh38
rs1009106418 Chr.3: 33496617-33496617 on Build GRCh38
rs189825663 Chr.3: 33496507-33496507 on Build GRCh38
rs1026083364 Chr.3: 33496284-33496284 on Build GRCh38
rs144417703 Chr.3: 33496491-33496491 on Build GRCh38
rs981636983 Chr.3: 33496489-33496489 on Build GRCh38
rs80104935 Chr.3: 33496409-33496409 on Build GRCh38
rs929995194 Chr.3: 33496596-33496596 on Build GRCh38
rs148431469 Chr.3: 33496593-33496593 on Build GRCh38
rs950229536 Chr.3: 33496487-33496487 on Build GRCh38
rs926808872 Chr.3: 33496412-33496412 on Build GRCh38
rs537523984 Chr.3: 33496444-33496444 on Build GRCh38
rs1009568575 Chr.3: 33496388-33496388 on Build GRCh38
rs56906831 Chr.3: 33496449-33496449 on Build GRCh38
rs898525615 Chr.3: 33496567-33496567 on Build GRCh38
rs755251872 Chr.3: 33496468-33496470 on Build GRCh38
rs531079741 Chr.3: 33496326-33496326 on Build GRCh38
rs10764 Chr.3: 33496538-33496538 on Build GRCh38
rs1020993209 Chr.3: 33496399-33496399 on Build GRCh38
rs1052812 Chr.3: 33496549-33496549 on Build GRCh38
rs886364072 Chr.3: 33496607-33496607 on Build GRCh38
rs950580926 Chr.3: 33496333-33496333 on Build GRCh38
rs557944978 Chr.3: 33496555-33496555 on Build GRCh38
rs563640531 Chr.3: 33496309-33496309 on Build GRCh38
rs1047606882 Chr.3: 33496597-33496597 on Build GRCh38
rs778810985 Chr.3: 33496395-33496395 on Build GRCh38
rs369346163 Chr.3: 33496286-33496286 on Build GRCh38
rs1019075298 Chr.3: 33496349-33496349 on Build GRCh38
rs11717495 Chr.3: 33496368-33496368 on Build GRCh38
rs950263887 Chr.3: 33496195-33496195 on Build GRCh38
rs966030319 Chr.3: 33496332-33496332 on Build GRCh38

View Full Product Details