Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33497246-33497756 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASP2
Entrez Gene ID
23122
Gene Name
cytoplasmic linker associated protein 2
Gene Aliases
-
UniGene
Hs.108614
Gene Chromosome Location
Chr.3: 33496245-33718356 on Build GRCh38
Transcripts
NM_001207044.1 NM_015097.2 XM_006713040.1 XM_006713041.1 XM_006713042.1 XM_006713048.1 XM_006713049.1 XM_006713050.1 XM_006713052.1 XM_006713053.1 XM_011533515.1 XM_017005947.1 XM_017005948.1 XM_017005949.1 XM_017005950.1 XM_017005951.1 XM_017005952.1 XM_017005953.1 XM_017005954.1 XM_017005955.1 XM_017005956.1 XM_017005957.1 XM_017005958.1 XM_017005959.1 XM_017005960.1 XM_017005961.1 XM_017005962.1 XM_017005963.1 XM_017005964.1 XM_017005965.1 XM_017005966.1 XM_017005967.1 XM_017005968.1 XM_017005969.1 XM_017005970.1 XM_017005971.1 XM_017005972.1 XM_017005973.1 XM_017005974.1 XM_017005975.1 XM_017005976.1 XM_017005977.1 XM_017005978.1 XM_017005979.1 XM_017005980.1 XM_017005981.1 XM_017005982.1 XM_017005983.1 XM_017005984.1 XM_017005985.1 XM_017005986.1

SNP Detail

SNP ID Chromosome Location
rs945886285 Chr.3: 33497650-33497650 on Build GRCh38
rs569729610 Chr.3: 33497571-33497571 on Build GRCh38
rs755400689 Chr.3: 33497509-33497509 on Build GRCh38
rs892885391 Chr.3: 33497375-33497375 on Build GRCh38
rs921846181 Chr.3: 33497565-33497565 on Build GRCh38
rs561375397 Chr.3: 33497432-33497432 on Build GRCh38
rs1055425493 Chr.3: 33497714-33497714 on Build GRCh38
rs1046526226 Chr.3: 33497514-33497514 on Build GRCh38
rs1006287219 Chr.3: 33497274-33497274 on Build GRCh38
rs916603605 Chr.3: 33497459-33497459 on Build GRCh38
rs543876183 Chr.3: 33497662-33497662 on Build GRCh38
rs549398496 Chr.3: 33497278-33497278 on Build GRCh38
rs958948994 Chr.3: 33497469-33497469 on Build GRCh38
rs958173157 Chr.3: 33497716-33497716 on Build GRCh38
rs931921969 Chr.3: 33497605-33497605 on Build GRCh38
rs925166168 Chr.3: 33497703-33497703 on Build GRCh38
rs541453579 Chr.3: 33497272-33497272 on Build GRCh38
rs931764969 Chr.3: 33497314-33497314 on Build GRCh38
rs866186709 Chr.3: 33497325-33497325 on Build GRCh38
rs938321248 Chr.3: 33497686-33497686 on Build GRCh38
rs1048584578 Chr.3: 33497553-33497553 on Build GRCh38
rs1041757711 Chr.3: 33497405-33497405 on Build GRCh38
rs1027140365 Chr.3: 33497495-33497495 on Build GRCh38
rs143936549 Chr.3: 33497438-33497438 on Build GRCh38
rs377189598 Chr.3: 33497334-33497338 on Build GRCh38
rs906954679 Chr.3: 33497681-33497681 on Build GRCh38
rs546968920 Chr.3: 33497460-33497460 on Build GRCh38
rs923072483 Chr.3: 33497358-33497358 on Build GRCh38
rs1017657680 Chr.3: 33497598-33497598 on Build GRCh38
rs551622519 Chr.3: 33497536-33497536 on Build GRCh38
rs1054330748 Chr.3: 33497324-33497324 on Build GRCh38
rs978319278 Chr.3: 33497700-33497700 on Build GRCh38
rs965376478 Chr.3: 33497552-33497552 on Build GRCh38
rs975824201 Chr.3: 33497558-33497558 on Build GRCh38
rs548021377 Chr.3: 33497585-33497585 on Build GRCh38
rs749186332 Chr.3: 33497533-33497533 on Build GRCh38
rs780211313 Chr.3: 33497510-33497510 on Build GRCh38
rs1010015114 Chr.3: 33497381-33497381 on Build GRCh38
rs1054275349 Chr.3: 33497630-33497630 on Build GRCh38
rs977988161 Chr.3: 33497370-33497370 on Build GRCh38
rs888677277 Chr.3: 33497554-33497554 on Build GRCh38
rs966469576 Chr.3: 33497699-33497699 on Build GRCh38
rs994735641 Chr.3: 33497549-33497549 on Build GRCh38
rs9311015 Chr.3: 33497467-33497467 on Build GRCh38
rs866392408 Chr.3: 33497730-33497730 on Build GRCh38
rs1030032965 Chr.3: 33497655-33497655 on Build GRCh38
rs906842265 Chr.3: 33497411-33497411 on Build GRCh38
rs966516270 Chr.3: 33497368-33497368 on Build GRCh38
rs539099496 Chr.3: 33497490-33497490 on Build GRCh38
rs570964473 Chr.3: 33497640-33497640 on Build GRCh38
rs768352840 Chr.3: 33497638-33497638 on Build GRCh38
rs975816271 Chr.3: 33497327-33497327 on Build GRCh38
rs995628087 Chr.3: 33497477-33497477 on Build GRCh38
rs997180935 Chr.3: 33497652-33497652 on Build GRCh38
rs964404603 Chr.3: 33497282-33497282 on Build GRCh38
rs190370584 Chr.3: 33497718-33497718 on Build GRCh38

View Full Product Details