Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 33516842-33517355 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLASP2
Entrez Gene ID
23122
Gene Name
cytoplasmic linker associated protein 2
Gene Aliases
-
UniGene
Hs.108614
Gene Chromosome Location
Chr.3: 33496245-33718356 on Build GRCh38
Transcripts
NM_001207044.1 NM_015097.2 XM_006713040.1 XM_006713041.1 XM_006713042.1 XM_006713048.1 XM_006713049.1 XM_006713050.1 XM_006713052.1 XM_006713053.1 XM_011533515.1 XM_017005947.1 XM_017005948.1 XM_017005949.1 XM_017005950.1 XM_017005951.1 XM_017005952.1 XM_017005953.1 XM_017005954.1 XM_017005955.1 XM_017005956.1 XM_017005957.1 XM_017005958.1 XM_017005959.1 XM_017005960.1 XM_017005961.1 XM_017005962.1 XM_017005963.1 XM_017005964.1 XM_017005965.1 XM_017005966.1 XM_017005967.1 XM_017005968.1 XM_017005969.1 XM_017005970.1 XM_017005971.1 XM_017005972.1 XM_017005973.1 XM_017005974.1 XM_017005975.1 XM_017005976.1 XM_017005977.1 XM_017005978.1 XM_017005979.1 XM_017005980.1 XM_017005981.1 XM_017005982.1 XM_017005983.1 XM_017005984.1 XM_017005985.1 XM_017005986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7236721 c.3272C>G p.S1091C Chr.3: 33517051-33517051 on Build GRCh38
4149857 c.3961G>T p.E1321* Chr.3: 33517001-33517001 on Build GRCh38
7133293 c.3850G>C p.E1284Q Chr.3: 33517112-33517112 on Build GRCh38
6097473 c.3134A>T p.H1045L Chr.3: 33517159-33517159 on Build GRCh38
7133292 c.3871G>C p.E1291Q Chr.3: 33517112-33517112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756833053 Chr.3: 33516945-33516949 on Build GRCh38
rs540149273 Chr.3: 33516929-33516929 on Build GRCh38
rs755915215 Chr.3: 33517068-33517068 on Build GRCh38
rs568243876 Chr.3: 33517248-33517248 on Build GRCh38
rs748075075 Chr.3: 33517048-33517048 on Build GRCh38
rs998334159 Chr.3: 33516906-33516906 on Build GRCh38
rs751091919 Chr.3: 33517292-33517293 on Build GRCh38
rs990984974 Chr.3: 33517086-33517086 on Build GRCh38
rs776038384 Chr.3: 33517104-33517104 on Build GRCh38
rs758473046 Chr.3: 33517189-33517189 on Build GRCh38
rs1006538681 Chr.3: 33516876-33516876 on Build GRCh38
rs1034868744 Chr.3: 33517074-33517074 on Build GRCh38
rs1357541 Chr.3: 33516915-33516915 on Build GRCh38
rs758462310 Chr.3: 33517026-33517026 on Build GRCh38
rs536516510 Chr.3: 33517329-33517329 on Build GRCh38
rs778564120 Chr.3: 33517187-33517188 on Build GRCh38
rs879240853 Chr.3: 33516890-33516890 on Build GRCh38
rs902960569 Chr.3: 33517306-33517306 on Build GRCh38
rs1042310619 Chr.3: 33517331-33517331 on Build GRCh38
rs77504651 Chr.3: 33516905-33516905 on Build GRCh38
rs200939212 Chr.3: 33517193-33517193 on Build GRCh38
rs115908242 Chr.3: 33516869-33516869 on Build GRCh38
rs570763044 Chr.3: 33517219-33517219 on Build GRCh38
rs930768824 Chr.3: 33517159-33517159 on Build GRCh38
rs1041832553 Chr.3: 33517301-33517301 on Build GRCh38
rs748907521 Chr.3: 33517076-33517076 on Build GRCh38
rs771551971 Chr.3: 33517099-33517099 on Build GRCh38
rs908164925 Chr.3: 33517165-33517165 on Build GRCh38
rs765427933 Chr.3: 33517178-33517178 on Build GRCh38
rs765231638 Chr.3: 33517008-33517008 on Build GRCh38
rs115622470 Chr.3: 33517234-33517234 on Build GRCh38
rs1054984937 Chr.3: 33517300-33517300 on Build GRCh38
rs1010819205 Chr.3: 33517037-33517037 on Build GRCh38
rs752629081 Chr.3: 33517198-33517198 on Build GRCh38
rs943607582 Chr.3: 33517202-33517202 on Build GRCh38
rs761099869 Chr.3: 33516968-33516968 on Build GRCh38
rs191181518 Chr.3: 33517229-33517229 on Build GRCh38
rs560504448 Chr.3: 33517108-33517108 on Build GRCh38
rs777450082 Chr.3: 33517072-33517072 on Build GRCh38
rs910768453 Chr.3: 33517202-33517203 on Build GRCh38
rs183373873 Chr.3: 33517081-33517081 on Build GRCh38
rs756036091 Chr.3: 33517214-33517214 on Build GRCh38
rs745358591 Chr.3: 33517094-33517094 on Build GRCh38
rs376402092 Chr.3: 33517151-33517151 on Build GRCh38
rs761137275 Chr.3: 33517107-33517107 on Build GRCh38
rs781181989 Chr.3: 33517047-33517047 on Build GRCh38
rs562583413 Chr.3: 33516996-33516996 on Build GRCh38
rs753202843 Chr.3: 33516965-33516965 on Build GRCh38
rs536065071 Chr.3: 33516892-33516892 on Build GRCh38
rs749755563 Chr.3: 33516941-33516941 on Build GRCh38
rs1000000562 Chr.3: 33516916-33516916 on Build GRCh38
rs944911199 Chr.3: 33516919-33516919 on Build GRCh38
rs548789620 Chr.3: 33517087-33517087 on Build GRCh38
rs185914650 Chr.3: 33516937-33516937 on Build GRCh38
rs7635999 Chr.3: 33517316-33517316 on Build GRCh38
rs574558157 Chr.3: 33516875-33516875 on Build GRCh38
rs181043424 Chr.3: 33516935-33516935 on Build GRCh38
rs1357542 Chr.3: 33516867-33516867 on Build GRCh38
rs933693462 Chr.3: 33517296-33517296 on Build GRCh38
rs764410511 Chr.3: 33516973-33516973 on Build GRCh38
rs760055712 Chr.3: 33516938-33516938 on Build GRCh38
rs750706744 Chr.3: 33517180-33517180 on Build GRCh38
rs966520046 Chr.3: 33517071-33517071 on Build GRCh38
rs200758119 Chr.3: 33516997-33516997 on Build GRCh38
rs944860392 Chr.3: 33517177-33517177 on Build GRCh38
rs750512970 Chr.3: 33517018-33517018 on Build GRCh38
rs900621626 Chr.3: 33516957-33516957 on Build GRCh38
rs187121803 Chr.3: 33517131-33517131 on Build GRCh38
rs190374140 Chr.3: 33516962-33516962 on Build GRCh38

View Full Product Details