Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37416957-37417503 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C3orf35
Entrez Gene ID
339883
Gene Name
chromosome 3 open reading frame 35
Gene Aliases
APRG1
UniGene
Hs.475945
Gene Chromosome Location
Chr.3: 37379111-37435497 on Build GRCh38
Transcripts
NM_001302831.1 NM_001302832.1 NM_178339.2 NM_178342.2 NM_178344.2 NR_126512.1 NR_126513.1 NR_126514.1 XM_017006291.1 XM_017006292.1 XM_017006293.1 XM_017006294.1 XM_017006295.1 XM_017006296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3427543 c.52G>T p.E18* Chr.3: 37417318-37417318 on Build GRCh38
5399186 c.146G>A p.R49K Chr.3: 37417412-37417412 on Build GRCh38
39361 c.85G>A p.A29T Chr.3: 37417351-37417351 on Build GRCh38
3660509 c.226+7G>T p.? Chr.3: 37417499-37417499 on Build GRCh38
5544576 c.150G>A p.R50R Chr.3: 37417416-37417416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904649599 Chr.3: 37417093-37417093 on Build GRCh38
rs150550724 Chr.3: 37417435-37417435 on Build GRCh38
rs767188987 Chr.3: 37417240-37417240 on Build GRCh38
rs541780396 Chr.3: 37417311-37417311 on Build GRCh38
rs772871496 Chr.3: 37417432-37417432 on Build GRCh38
rs567353801 Chr.3: 37417423-37417423 on Build GRCh38
rs953138605 Chr.3: 37417154-37417154 on Build GRCh38
rs772073350 Chr.3: 37417414-37417414 on Build GRCh38
rs776485911 Chr.3: 37417427-37417427 on Build GRCh38
rs774104173 Chr.3: 37417231-37417231 on Build GRCh38
rs914409771 Chr.3: 37417167-37417167 on Build GRCh38
rs766947824 Chr.3: 37417438-37417438 on Build GRCh38
rs564301662 Chr.3: 37417328-37417328 on Build GRCh38
rs767525736 Chr.3: 37417422-37417443 on Build GRCh38
rs986403175 Chr.3: 37417156-37417156 on Build GRCh38
rs1056448043 Chr.3: 37417283-37417283 on Build GRCh38
rs575367688 Chr.3: 37417258-37417258 on Build GRCh38
rs770888801 Chr.3: 37417225-37417225 on Build GRCh38
rs199710176 Chr.3: 37417105-37417105 on Build GRCh38
rs193097162 Chr.3: 37417355-37417355 on Build GRCh38
rs1026910608 Chr.3: 37417275-37417275 on Build GRCh38
rs750793904 Chr.3: 37417279-37417279 on Build GRCh38
rs923945454 Chr.3: 37417063-37417063 on Build GRCh38
rs141747430 Chr.3: 37417350-37417350 on Build GRCh38
rs372151800 Chr.3: 37417413-37417413 on Build GRCh38
rs760000269 Chr.3: 37417449-37417449 on Build GRCh38
rs367948667 Chr.3: 37417367-37417367 on Build GRCh38
rs747658245 Chr.3: 37417430-37417430 on Build GRCh38
rs549042345 Chr.3: 37417374-37417374 on Build GRCh38
rs935297751 Chr.3: 37417065-37417065 on Build GRCh38
rs977200293 Chr.3: 37417473-37417473 on Build GRCh38
rs777830157 Chr.3: 37417412-37417412 on Build GRCh38
rs1012851249 Chr.3: 37417088-37417088 on Build GRCh38
rs560116850 Chr.3: 37417039-37417039 on Build GRCh38
rs753418348 Chr.3: 37417260-37417260 on Build GRCh38
rs375545769 Chr.3: 37417303-37417303 on Build GRCh38
rs896542814 Chr.3: 37417069-37417069 on Build GRCh38
rs368233774 Chr.3: 37417306-37417306 on Build GRCh38
rs767719240 Chr.3: 37417474-37417474 on Build GRCh38
rs545158033 Chr.3: 37417110-37417110 on Build GRCh38
rs746863872 Chr.3: 37417417-37417417 on Build GRCh38
rs760199532 Chr.3: 37417345-37417345 on Build GRCh38
rs556781768 Chr.3: 37417140-37417140 on Build GRCh38
rs752758059 Chr.3: 37417386-37417386 on Build GRCh38
rs75417768 Chr.3: 37417360-37417360 on Build GRCh38
rs780318918 Chr.3: 37417284-37417284 on Build GRCh38
rs577780833 Chr.3: 37417096-37417096 on Build GRCh38
rs879035733 Chr.3: 37417041-37417041 on Build GRCh38
rs528742914 Chr.3: 37417441-37417441 on Build GRCh38
rs961714634 Chr.3: 37417131-37417131 on Build GRCh38
rs756249863 Chr.3: 37417398-37417398 on Build GRCh38
rs754894408 Chr.3: 37417394-37417395 on Build GRCh38
rs751983688 Chr.3: 37417364-37417364 on Build GRCh38
rs769363063 Chr.3: 37417431-37417431 on Build GRCh38
rs1029990288 Chr.3: 37417372-37417372 on Build GRCh38
rs936504585 Chr.3: 37417101-37417101 on Build GRCh38
rs995010875 Chr.3: 37417448-37417448 on Build GRCh38
rs1052110504 Chr.3: 37417421-37417421 on Build GRCh38
rs745654766 Chr.3: 37417321-37417321 on Build GRCh38
rs758890556 Chr.3: 37417282-37417282 on Build GRCh38
rs890126702 Chr.3: 37417150-37417150 on Build GRCh38
rs1031570548 Chr.3: 37417480-37417480 on Build GRCh38
rs200577382 Chr.3: 37417334-37417334 on Build GRCh38
rs775372232 Chr.3: 37417331-37417331 on Build GRCh38
rs770835896 Chr.3: 37417310-37417310 on Build GRCh38
rs555383089 Chr.3: 37417038-37417038 on Build GRCh38
rs759378329 Chr.3: 37417235-37417235 on Build GRCh38
rs779676773 Chr.3: 37417415-37417415 on Build GRCh38
rs113724034 Chr.3: 37417267-37417267 on Build GRCh38
rs760376070 Chr.3: 37417251-37417251 on Build GRCh38
rs968482646 Chr.3: 37417214-37417214 on Build GRCh38
rs199499167 Chr.3: 37417354-37417354 on Build GRCh38
rs751675294 Chr.3: 37417293-37417293 on Build GRCh38
rs73826522 Chr.3: 37417066-37417066 on Build GRCh38
rs374602897 Chr.3: 37417048-37417048 on Build GRCh38
rs537058663 Chr.3: 37416992-37416992 on Build GRCh38
rs1043745510 Chr.3: 37417049-37417049 on Build GRCh38
rs968534009 Chr.3: 37417468-37417468 on Build GRCh38
rs1053603290 Chr.3: 37417395-37417395 on Build GRCh38
rs763529207 Chr.3: 37417357-37417357 on Build GRCh38
rs775075723 Chr.3: 37417246-37417246 on Build GRCh38
rs752187519 Chr.3: 37417145-37417145 on Build GRCh38
rs953347895 Chr.3: 37417032-37417032 on Build GRCh38
rs985881001 Chr.3: 37417024-37417024 on Build GRCh38
rs888902873 Chr.3: 37417387-37417387 on Build GRCh38
rs567460139 Chr.3: 37417351-37417351 on Build GRCh38
rs186633646 Chr.3: 37417276-37417276 on Build GRCh38
rs527707604 Chr.3: 37417113-37417113 on Build GRCh38
rs1017755044 Chr.3: 37417026-37417026 on Build GRCh38
rs985959170 Chr.3: 37417033-37417033 on Build GRCh38
rs553218895 Chr.3: 37417091-37417091 on Build GRCh38
rs774554578 Chr.3: 37417442-37417442 on Build GRCh38

View Full Product Details