Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 37982665-37983172 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTDSPL
Entrez Gene ID
10217
Gene Name
CTD small phosphatase like
Gene Aliases
C3orf8, HYA22, PSR1, RBSP3, SCP3
UniGene
Hs.475963
Gene Chromosome Location
Chr.3: 37862152-37984469 on Build GRCh38
Transcripts
NM_001008392.1 NM_005808.2 XM_017005519.1 XM_017005520.1

SNP Detail

SNP ID Chromosome Location
rs1036433635 Chr.3: 37982897-37982897 on Build GRCh38
rs1042172047 Chr.3: 37983126-37983126 on Build GRCh38
rs985448277 Chr.3: 37982849-37982849 on Build GRCh38
rs756477392 Chr.3: 37982691-37982691 on Build GRCh38
rs999346518 Chr.3: 37982965-37982966 on Build GRCh38
rs940932391 Chr.3: 37982908-37982908 on Build GRCh38
rs773149996 Chr.3: 37982884-37982884 on Build GRCh38
rs11129775 Chr.3: 37982917-37982917 on Build GRCh38
rs780451295 Chr.3: 37982835-37982835 on Build GRCh38
rs1035345182 Chr.3: 37982767-37982767 on Build GRCh38
rs953619601 Chr.3: 37982817-37982817 on Build GRCh38
rs908062274 Chr.3: 37982876-37982876 on Build GRCh38
rs982517087 Chr.3: 37983068-37983068 on Build GRCh38
rs375209790 Chr.3: 37982930-37982930 on Build GRCh38
rs894397093 Chr.3: 37983148-37983148 on Build GRCh38
rs952448336 Chr.3: 37983057-37983057 on Build GRCh38
rs1026649121 Chr.3: 37983036-37983036 on Build GRCh38
rs928728108 Chr.3: 37982902-37982902 on Build GRCh38
rs567053284 Chr.3: 37982833-37982833 on Build GRCh38
rs746801560 Chr.3: 37983098-37983098 on Build GRCh38
rs75142232 Chr.3: 37983092-37983092 on Build GRCh38
rs1003231389 Chr.3: 37982765-37982765 on Build GRCh38
rs947435021 Chr.3: 37983125-37983125 on Build GRCh38
rs11709029 Chr.3: 37982929-37982929 on Build GRCh38
rs903039735 Chr.3: 37982980-37982980 on Build GRCh38
rs115135888 Chr.3: 37983031-37983031 on Build GRCh38
rs1041539121 Chr.3: 37982960-37982960 on Build GRCh38
rs187387844 Chr.3: 37982803-37982803 on Build GRCh38
rs542982572 Chr.3: 37983107-37983107 on Build GRCh38
rs35294369 Chr.3: 37982760-37982760 on Build GRCh38
rs958624899 Chr.3: 37983005-37983005 on Build GRCh38
rs984923729 Chr.3: 37982864-37982864 on Build GRCh38
rs79505957 Chr.3: 37983134-37983134 on Build GRCh38
rs1056970237 Chr.3: 37983139-37983139 on Build GRCh38
rs923559168 Chr.3: 37982922-37982922 on Build GRCh38
rs932260729 Chr.3: 37983101-37983101 on Build GRCh38
rs879629599 Chr.3: 37982925-37982925 on Build GRCh38
rs919644820 Chr.3: 37982808-37982808 on Build GRCh38
rs903281823 Chr.3: 37983133-37983133 on Build GRCh38
rs962491293 Chr.3: 37983079-37983079 on Build GRCh38
rs749764982 Chr.3: 37982848-37982848 on Build GRCh38
rs940851791 Chr.3: 37982865-37982865 on Build GRCh38
rs766989431 Chr.3: 37983119-37983119 on Build GRCh38
rs77923385 Chr.3: 37983001-37983001 on Build GRCh38
rs1035988968 Chr.3: 37983002-37983002 on Build GRCh38
rs534367073 Chr.3: 37982861-37982861 on Build GRCh38
rs985011632 Chr.3: 37983093-37983093 on Build GRCh38
rs901626456 Chr.3: 37983028-37983028 on Build GRCh38
rs958593387 Chr.3: 37982782-37982782 on Build GRCh38
rs1050409374 Chr.3: 37983117-37983117 on Build GRCh38
rs1043645241 Chr.3: 37982940-37982940 on Build GRCh38
rs558012435 Chr.3: 37983076-37983076 on Build GRCh38
rs747645056 Chr.3: 37982946-37982946 on Build GRCh38
rs893222532 Chr.3: 37982944-37982944 on Build GRCh38
rs1055833920 Chr.3: 37982923-37982923 on Build GRCh38

View Full Product Details