Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38059650-38060151 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEC1
Entrez Gene ID
9940
Gene Name
deleted in lung and esophageal cancer 1
Gene Aliases
CFAP81, DLC-1, DLC1, F56
UniGene
Hs.714499
Gene Chromosome Location
Chr.3: 38038595-38122741 on Build GRCh38
Transcripts
NM_001321153.1 NM_007335.3 NM_007337.3 XM_006713438.3 XM_006713439.3 XM_006713440.3 XM_011534317.2 XM_011534318.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1422667 c.613C>A p.H205N Chr.3: 38059792-38059792 on Build GRCh38
4584299 c.654G>A p.P218P Chr.3: 38059833-38059833 on Build GRCh38
5590540 c.623C>T p.S208F Chr.3: 38059802-38059802 on Build GRCh38
6644606 c.637T>C p.Y213H Chr.3: 38059816-38059816 on Build GRCh38
1566798 c.643G>A p.D215N Chr.3: 38059822-38059822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1054679008 Chr.3: 38059730-38059730 on Build GRCh38
rs1019239131 Chr.3: 38060036-38060036 on Build GRCh38
rs76690040 Chr.3: 38059873-38059873 on Build GRCh38
rs776755272 Chr.3: 38060013-38060013 on Build GRCh38
rs964922048 Chr.3: 38060074-38060074 on Build GRCh38
rs913313501 Chr.3: 38060108-38060108 on Build GRCh38
rs777329992 Chr.3: 38059692-38059693 on Build GRCh38
rs776961217 Chr.3: 38059846-38059846 on Build GRCh38
rs1008536187 Chr.3: 38060014-38060014 on Build GRCh38
rs372838936 Chr.3: 38060041-38060041 on Build GRCh38
rs907121140 Chr.3: 38059988-38059988 on Build GRCh38
rs898261762 Chr.3: 38060015-38060015 on Build GRCh38
rs35347349 Chr.3: 38059846-38059846 on Build GRCh38
rs749238302 Chr.3: 38059773-38059773 on Build GRCh38
rs112900948 Chr.3: 38059959-38059963 on Build GRCh38
rs765278885 Chr.3: 38059788-38059788 on Build GRCh38
rs380566 Chr.3: 38059833-38059833 on Build GRCh38
rs758915315 Chr.3: 38059824-38059824 on Build GRCh38
rs376309726 Chr.3: 38059802-38059802 on Build GRCh38
rs757153925 Chr.3: 38059809-38059809 on Build GRCh38
rs149190717 Chr.3: 38059822-38059822 on Build GRCh38
rs752481817 Chr.3: 38059800-38059800 on Build GRCh38
rs755854100 Chr.3: 38059801-38059801 on Build GRCh38
rs773212985 Chr.3: 38059856-38059856 on Build GRCh38
rs935879735 Chr.3: 38059674-38059674 on Build GRCh38
rs891731297 Chr.3: 38059734-38059734 on Build GRCh38
rs34012183 Chr.3: 38059754-38059754 on Build GRCh38
rs776717392 Chr.3: 38059707-38059707 on Build GRCh38
rs767306644 Chr.3: 38059799-38059799 on Build GRCh38
rs547071525 Chr.3: 38059798-38059798 on Build GRCh38
rs911455205 Chr.3: 38060035-38060035 on Build GRCh38
rs748946666 Chr.3: 38059789-38059791 on Build GRCh38
rs771024725 Chr.3: 38060001-38060001 on Build GRCh38
rs779607820 Chr.3: 38059762-38059762 on Build GRCh38
rs962763087 Chr.3: 38059791-38059791 on Build GRCh38
rs767255188 Chr.3: 38059736-38059736 on Build GRCh38
rs576030226 Chr.3: 38059679-38059679 on Build GRCh38
rs763743229 Chr.3: 38059883-38059883 on Build GRCh38
rs776714626 Chr.3: 38059783-38059783 on Build GRCh38
rs921900711 Chr.3: 38060070-38060070 on Build GRCh38
rs778527202 Chr.3: 38059821-38059821 on Build GRCh38
rs759797688 Chr.3: 38060044-38060044 on Build GRCh38
rs753558054 Chr.3: 38059891-38059891 on Build GRCh38
rs451863 Chr.3: 38059831-38059831 on Build GRCh38
rs182898193 Chr.3: 38059784-38059784 on Build GRCh38
rs753407076 Chr.3: 38059804-38059804 on Build GRCh38
rs990600630 Chr.3: 38060095-38060095 on Build GRCh38
rs996356646 Chr.3: 38060079-38060079 on Build GRCh38
rs775929039 Chr.3: 38059765-38059765 on Build GRCh38
rs930690990 Chr.3: 38059808-38059808 on Build GRCh38
rs761787138 Chr.3: 38059709-38059709 on Build GRCh38
rs76834284 Chr.3: 38059874-38059874 on Build GRCh38
rs760910598 Chr.3: 38059693-38059693 on Build GRCh38
rs780414386 Chr.3: 38059826-38059826 on Build GRCh38
rs945969735 Chr.3: 38059947-38059947 on Build GRCh38
rs11708345 Chr.3: 38059931-38059931 on Build GRCh38
rs775341611 Chr.3: 38059763-38059763 on Build GRCh38
rs756845387 Chr.3: 38059811-38059811 on Build GRCh38
rs142348713 Chr.3: 38060046-38060046 on Build GRCh38
rs769895875 Chr.3: 38059853-38059853 on Build GRCh38
rs533245447 Chr.3: 38059852-38059852 on Build GRCh38
rs754820344 Chr.3: 38059722-38059722 on Build GRCh38
rs777275073 Chr.3: 38059752-38059752 on Build GRCh38
rs751467584 Chr.3: 38059714-38059714 on Build GRCh38
rs375926980 Chr.3: 38059832-38059832 on Build GRCh38
rs764083181 Chr.3: 38059695-38059695 on Build GRCh38
rs759486330 Chr.3: 38059858-38059858 on Build GRCh38
rs936062060 Chr.3: 38059890-38059890 on Build GRCh38
rs76603930 Chr.3: 38059757-38059757 on Build GRCh38
rs766371729 Chr.3: 38059712-38059712 on Build GRCh38
rs752326062 Chr.3: 38059742-38059742 on Build GRCh38
rs1027828635 Chr.3: 38060031-38060031 on Build GRCh38
rs974761443 Chr.3: 38060062-38060062 on Build GRCh38
rs760698866 Chr.3: 38059974-38059974 on Build GRCh38
rs187491596 Chr.3: 38060040-38060040 on Build GRCh38
rs995633698 Chr.3: 38060020-38060020 on Build GRCh38
rs957485513 Chr.3: 38060083-38060083 on Build GRCh38
rs755728434 Chr.3: 38059749-38059749 on Build GRCh38
rs891590185 Chr.3: 38059943-38059943 on Build GRCh38
rs950820276 Chr.3: 38060033-38060033 on Build GRCh38
rs770439676 Chr.3: 38059806-38059808 on Build GRCh38
rs768982207 Chr.3: 38059774-38059774 on Build GRCh38
rs774298544 Chr.3: 38059790-38059790 on Build GRCh38
rs562302374 Chr.3: 38059908-38059908 on Build GRCh38
rs973620002 Chr.3: 38059813-38059813 on Build GRCh38
rs796669659 Chr.3: 38059873-38059874 on Build GRCh38

View Full Product Details