Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 38482120-38482627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACVR2B
Entrez Gene ID
93
Gene Name
activin A receptor type 2B
Gene Aliases
ACTRIIB, ActR-IIB, HTX4
UniGene
Hs.174273
Gene Chromosome Location
Chr.3: 38454299-38493142 on Build GRCh38
Transcripts
NM_001106.3 XM_005265583.3 XM_017007514.1 XM_017007515.1 XM_017007516.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6528854 c.1227G>C p.E409D Chr.3: 38482443-38482443 on Build GRCh38
1422750 c.1240T>G p.F414V Chr.3: 38482456-38482456 on Build GRCh38
350864 c.1105G>T p.V369L Chr.3: 38482228-38482228 on Build GRCh38
1422748 c.1140C>A p.A380A Chr.3: 38482263-38482263 on Build GRCh38
4117275 c.1148G>A p.R383H Chr.3: 38482271-38482271 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs760458790 Chr.3: 38482347-38482347 on Build GRCh38
rs961153672 Chr.3: 38482228-38482228 on Build GRCh38
rs762644398 Chr.3: 38482566-38482566 on Build GRCh38
rs780481900 Chr.3: 38482390-38482390 on Build GRCh38
rs999144243 Chr.3: 38482534-38482534 on Build GRCh38
rs904463166 Chr.3: 38482517-38482517 on Build GRCh38
rs373893017 Chr.3: 38482152-38482152 on Build GRCh38
rs749622519 Chr.3: 38482603-38482603 on Build GRCh38
rs115155428 Chr.3: 38482193-38482193 on Build GRCh38
rs770169018 Chr.3: 38482406-38482406 on Build GRCh38
rs751584141 Chr.3: 38482191-38482191 on Build GRCh38
rs773512733 Chr.3: 38482408-38482408 on Build GRCh38
rs781509169 Chr.3: 38482449-38482449 on Build GRCh38
rs373852166 Chr.3: 38482558-38482558 on Build GRCh38
rs749494825 Chr.3: 38482479-38482479 on Build GRCh38
rs532011138 Chr.3: 38482418-38482418 on Build GRCh38
rs776494725 Chr.3: 38482512-38482512 on Build GRCh38
rs749439811 Chr.3: 38482410-38482410 on Build GRCh38
rs778374625 Chr.3: 38482245-38482245 on Build GRCh38
rs775292498 Chr.3: 38482364-38482364 on Build GRCh38
rs781594097 Chr.3: 38482404-38482404 on Build GRCh38
rs1038397919 Chr.3: 38482575-38482575 on Build GRCh38
rs749940757 Chr.3: 38482376-38482376 on Build GRCh38
rs755527007 Chr.3: 38482444-38482444 on Build GRCh38
rs762802231 Chr.3: 38482396-38482396 on Build GRCh38
rs753483029 Chr.3: 38482355-38482355 on Build GRCh38
rs755884935 Chr.3: 38482212-38482212 on Build GRCh38
rs374865845 Chr.3: 38482182-38482182 on Build GRCh38
rs9876823 Chr.3: 38482567-38482567 on Build GRCh38
rs763922415 Chr.3: 38482348-38482348 on Build GRCh38
rs758494015 Chr.3: 38482165-38482165 on Build GRCh38
rs772891201 Chr.3: 38482565-38482565 on Build GRCh38
rs763083195 Chr.3: 38482335-38482335 on Build GRCh38
rs765010818 Chr.3: 38482560-38482560 on Build GRCh38
rs778210512 Chr.3: 38482477-38482477 on Build GRCh38
rs1025852963 Chr.3: 38482206-38482206 on Build GRCh38
rs988287106 Chr.3: 38482468-38482468 on Build GRCh38
rs188465635 Chr.3: 38482580-38482580 on Build GRCh38
rs77316668 Chr.3: 38482595-38482595 on Build GRCh38
rs140285560 Chr.3: 38482503-38482503 on Build GRCh38
rs763888963 Chr.3: 38482427-38482427 on Build GRCh38
rs748470062 Chr.3: 38482405-38482405 on Build GRCh38
rs376817774 Chr.3: 38482147-38482147 on Build GRCh38
rs201145228 Chr.3: 38482312-38482312 on Build GRCh38
rs752442546 Chr.3: 38482210-38482210 on Build GRCh38
rs745896399 Chr.3: 38482497-38482497 on Build GRCh38
rs1022712387 Chr.3: 38482211-38482211 on Build GRCh38
rs761653104 Chr.3: 38482559-38482559 on Build GRCh38
rs1023854579 Chr.3: 38482267-38482267 on Build GRCh38
rs778499283 Chr.3: 38482369-38482369 on Build GRCh38
rs368044147 Chr.3: 38482169-38482169 on Build GRCh38
rs146067304 Chr.3: 38482485-38482485 on Build GRCh38
rs767371487 Chr.3: 38482336-38482336 on Build GRCh38
rs770969114 Chr.3: 38482484-38482484 on Build GRCh38
rs969747733 Chr.3: 38482224-38482224 on Build GRCh38
rs747347266 Chr.3: 38482273-38482273 on Build GRCh38
rs761061144 Chr.3: 38482160-38482160 on Build GRCh38
rs201625314 Chr.3: 38482393-38482393 on Build GRCh38
rs762092222 Chr.3: 38482325-38482325 on Build GRCh38
rs772598999 Chr.3: 38482258-38482258 on Build GRCh38
rs552934349 Chr.3: 38482385-38482385 on Build GRCh38
rs773445192 Chr.3: 38482333-38482333 on Build GRCh38
rs759064243 Chr.3: 38482161-38482161 on Build GRCh38
rs199839373 Chr.3: 38482201-38482201 on Build GRCh38
rs745334215 Chr.3: 38482251-38482251 on Build GRCh38
rs979120415 Chr.3: 38482391-38482391 on Build GRCh38
rs772476257 Chr.3: 38482151-38482151 on Build GRCh38
rs149695736 Chr.3: 38482263-38482263 on Build GRCh38
rs753528065 Chr.3: 38482238-38482238 on Build GRCh38
rs765402817 Chr.3: 38482184-38482184 on Build GRCh38
rs756925890 Chr.3: 38482361-38482361 on Build GRCh38
rs752052061 Chr.3: 38482440-38482440 on Build GRCh38
rs532752371 Chr.3: 38482344-38482344 on Build GRCh38
rs13093134 Chr.3: 38482500-38482500 on Build GRCh38
rs151252568 Chr.3: 38482594-38482594 on Build GRCh38
rs2126533 Chr.3: 38482303-38482303 on Build GRCh38
rs769910670 Chr.3: 38482503-38482505 on Build GRCh38
rs750062087 Chr.3: 38482435-38482435 on Build GRCh38
rs768941474 Chr.3: 38482397-38482397 on Build GRCh38
rs748524971 Chr.3: 38482467-38482467 on Build GRCh38
rs752669674 Chr.3: 38482340-38482340 on Build GRCh38
rs147650411 Chr.3: 38482442-38482442 on Build GRCh38
rs372522416 Chr.3: 38482332-38482332 on Build GRCh38
rs777350236 Chr.3: 38482233-38482233 on Build GRCh38
rs775333679 Chr.3: 38482419-38482419 on Build GRCh38
rs959824588 Chr.3: 38482185-38482185 on Build GRCh38
rs778263926 Chr.3: 38482597-38482597 on Build GRCh38
rs759519802 Chr.3: 38482192-38482192 on Build GRCh38
rs368450103 Chr.3: 38482314-38482314 on Build GRCh38
rs145005293 Chr.3: 38482434-38482434 on Build GRCh38
rs375131694 Chr.3: 38482470-38482470 on Build GRCh38
rs530383572 Chr.3: 38482381-38482381 on Build GRCh38
rs756801382 Chr.3: 38482243-38482243 on Build GRCh38
rs192767800 Chr.3: 38482179-38482179 on Build GRCh38
rs142291277 Chr.3: 38482591-38482591 on Build GRCh38
rs760639941 Chr.3: 38482425-38482425 on Build GRCh38

View Full Product Details