Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39142539-39143031 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP1
Entrez Gene ID
64651
Gene Name
cysteine and serine rich nuclear protein 1
Gene Aliases
AXUD1, CSRNP-1, FAM130B, TAIP-3, URAX1
UniGene
Hs.370950
Gene Chromosome Location
Chr.3: 39141851-39154602 on Build GRCh38
Transcripts
NM_001320559.1 NM_001320560.1 NM_033027.3 XM_017007048.1 XM_017007049.1

SNP Detail

SNP ID Chromosome Location
rs892049619 Chr.3: 39142774-39142774 on Build GRCh38
rs901813753 Chr.3: 39142754-39142754 on Build GRCh38
rs867774180 Chr.3: 39142939-39142939 on Build GRCh38
rs933325324 Chr.3: 39142755-39142755 on Build GRCh38
rs917015414 Chr.3: 39142821-39142821 on Build GRCh38
rs758294419 Chr.3: 39142762-39142762 on Build GRCh38
rs1053555636 Chr.3: 39142756-39142756 on Build GRCh38
rs746841129 Chr.3: 39143007-39143007 on Build GRCh38
rs975012764 Chr.3: 39142981-39142981 on Build GRCh38
rs866831466 Chr.3: 39142704-39142704 on Build GRCh38
rs892541972 Chr.3: 39142703-39142703 on Build GRCh38
rs77677193 Chr.3: 39142712-39142712 on Build GRCh38
rs1053778142 Chr.3: 39142858-39142858 on Build GRCh38
rs936645897 Chr.3: 39142618-39142618 on Build GRCh38
rs567417718 Chr.3: 39142658-39142658 on Build GRCh38
rs1002336754 Chr.3: 39142804-39142804 on Build GRCh38
rs573912089 Chr.3: 39142823-39142824 on Build GRCh38
rs992456472 Chr.3: 39142594-39142594 on Build GRCh38
rs704960 Chr.3: 39142862-39142862 on Build GRCh38
rs909082095 Chr.3: 39142717-39142717 on Build GRCh38
rs538680167 Chr.3: 39142802-39142802 on Build GRCh38
rs980937244 Chr.3: 39142824-39142824 on Build GRCh38
rs779781679 Chr.3: 39143006-39143006 on Build GRCh38
rs141303054 Chr.3: 39142819-39142819 on Build GRCh38
rs1054234057 Chr.3: 39142691-39142691 on Build GRCh38
rs752731298 Chr.3: 39142652-39142652 on Build GRCh38
rs926858708 Chr.3: 39142838-39142838 on Build GRCh38
rs114897817 Chr.3: 39142978-39142978 on Build GRCh38
rs573233408 Chr.3: 39142780-39142780 on Build GRCh38
rs150772341 Chr.3: 39142979-39142979 on Build GRCh38
rs1030143951 Chr.3: 39142711-39142711 on Build GRCh38
rs1025973172 Chr.3: 39142887-39142887 on Build GRCh38
rs145039917 Chr.3: 39142857-39142857 on Build GRCh38
rs536051065 Chr.3: 39142697-39142697 on Build GRCh38
rs553480530 Chr.3: 39142727-39142727 on Build GRCh38
rs373892326 Chr.3: 39142772-39142772 on Build GRCh38
rs973086247 Chr.3: 39142768-39142768 on Build GRCh38
rs753894764 Chr.3: 39142862-39142862 on Build GRCh38
rs575552025 Chr.3: 39142853-39142853 on Build GRCh38
rs1021768424 Chr.3: 39142783-39142783 on Build GRCh38
rs960766177 Chr.3: 39142832-39142832 on Build GRCh38
rs943421484 Chr.3: 39142726-39142726 on Build GRCh38
rs950626501 Chr.3: 39142941-39142941 on Build GRCh38
rs185077222 Chr.3: 39142889-39142889 on Build GRCh38
rs533874019 Chr.3: 39142770-39142770 on Build GRCh38
rs890261240 Chr.3: 39142736-39142736 on Build GRCh38
rs992965165 Chr.3: 39142807-39142807 on Build GRCh38
rs905450848 Chr.3: 39142576-39142576 on Build GRCh38
rs868797872 Chr.3: 39142824-39142824 on Build GRCh38
rs984679253 Chr.3: 39142949-39142949 on Build GRCh38
rs755209802 Chr.3: 39142894-39142897 on Build GRCh38
rs1033446678 Chr.3: 39142831-39142831 on Build GRCh38

View Full Product Details