Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39142918-39143428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CSRNP1
Entrez Gene ID
64651
Gene Name
cysteine and serine rich nuclear protein 1
Gene Aliases
AXUD1, CSRNP-1, FAM130B, TAIP-3, URAX1
UniGene
Hs.370950
Gene Chromosome Location
Chr.3: 39141851-39154602 on Build GRCh38
Transcripts
NM_001320559.1 NM_001320560.1 NM_033027.3 XM_017007048.1 XM_017007049.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6342700 c.1458C>A p.S486R Chr.3: 39143367-39143367 on Build GRCh38
3593793 c.1614C>T p.P538P Chr.3: 39143211-39143211 on Build GRCh38
3593791 c.1665C>T p.S555S Chr.3: 39143160-39143160 on Build GRCh38
3593792 c.1634C>T p.P545L Chr.3: 39143191-39143191 on Build GRCh38
4909428 c.1449G>C p.V483V Chr.3: 39143376-39143376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053118513 Chr.3: 39143051-39143051 on Build GRCh38
rs762320018 Chr.3: 39143073-39143073 on Build GRCh38
rs368077080 Chr.3: 39143093-39143093 on Build GRCh38
rs774697337 Chr.3: 39143347-39143347 on Build GRCh38
rs752013464 Chr.3: 39143231-39143233 on Build GRCh38
rs371134715 Chr.3: 39143375-39143375 on Build GRCh38
rs746841129 Chr.3: 39143007-39143007 on Build GRCh38
rs769006580 Chr.3: 39143163-39143163 on Build GRCh38
rs913212988 Chr.3: 39143276-39143276 on Build GRCh38
rs780866317 Chr.3: 39143123-39143123 on Build GRCh38
rs975012764 Chr.3: 39142981-39142981 on Build GRCh38
rs377458175 Chr.3: 39143012-39143012 on Build GRCh38
rs369147979 Chr.3: 39143124-39143124 on Build GRCh38
rs267599815 Chr.3: 39143169-39143169 on Build GRCh38
rs201765293 Chr.3: 39143204-39143204 on Build GRCh38
rs772447030 Chr.3: 39143060-39143060 on Build GRCh38
rs1024021437 Chr.3: 39143128-39143128 on Build GRCh38
rs775624061 Chr.3: 39143061-39143061 on Build GRCh38
rs933387373 Chr.3: 39143234-39143234 on Build GRCh38
rs780139369 Chr.3: 39143385-39143385 on Build GRCh38
rs200165962 Chr.3: 39143025-39143025 on Build GRCh38
rs762266789 Chr.3: 39143170-39143170 on Build GRCh38
rs768790408 Chr.3: 39143337-39143337 on Build GRCh38
rs750974555 Chr.3: 39143199-39143199 on Build GRCh38
rs34628941 Chr.3: 39143211-39143211 on Build GRCh38
rs748487431 Chr.3: 39143354-39143354 on Build GRCh38
rs781003783 Chr.3: 39143221-39143221 on Build GRCh38
rs779781679 Chr.3: 39143006-39143006 on Build GRCh38
rs577008244 Chr.3: 39143144-39143144 on Build GRCh38
rs377548334 Chr.3: 39143328-39143328 on Build GRCh38
rs142734319 Chr.3: 39143082-39143082 on Build GRCh38
rs757146156 Chr.3: 39143383-39143383 on Build GRCh38
rs556447889 Chr.3: 39143237-39143237 on Build GRCh38
rs947381174 Chr.3: 39143320-39143320 on Build GRCh38
rs375503400 Chr.3: 39143021-39143021 on Build GRCh38
rs768820347 Chr.3: 39143281-39143281 on Build GRCh38
rs752177833 Chr.3: 39143331-39143331 on Build GRCh38
rs1051755277 Chr.3: 39143187-39143187 on Build GRCh38
rs773768262 Chr.3: 39143362-39143362 on Build GRCh38
rs112383054 Chr.3: 39143135-39143135 on Build GRCh38
rs765751680 Chr.3: 39143196-39143196 on Build GRCh38
rs763468626 Chr.3: 39143361-39143361 on Build GRCh38
rs780011728 Chr.3: 39143279-39143279 on Build GRCh38
rs114897817 Chr.3: 39142978-39142978 on Build GRCh38
rs374512400 Chr.3: 39143166-39143166 on Build GRCh38
rs1013929707 Chr.3: 39143101-39143101 on Build GRCh38
rs766093888 Chr.3: 39143037-39143038 on Build GRCh38
rs372854348 Chr.3: 39143133-39143133 on Build GRCh38
rs150772341 Chr.3: 39142979-39142979 on Build GRCh38
rs764432988 Chr.3: 39143062-39143062 on Build GRCh38
rs200230516 Chr.3: 39143035-39143035 on Build GRCh38
rs754593845 Chr.3: 39143214-39143214 on Build GRCh38
rs764039494 Chr.3: 39143380-39143380 on Build GRCh38
rs868231312 Chr.3: 39143141-39143141 on Build GRCh38
rs762414194 Chr.3: 39143200-39143200 on Build GRCh38
rs759182414 Chr.3: 39143368-39143368 on Build GRCh38
rs372252051 Chr.3: 39143229-39143229 on Build GRCh38
rs147162469 Chr.3: 39143393-39143393 on Build GRCh38
rs566969447 Chr.3: 39143259-39143259 on Build GRCh38
rs111692560 Chr.3: 39143019-39143019 on Build GRCh38
rs1016123717 Chr.3: 39143188-39143188 on Build GRCh38
rs538715292 Chr.3: 39143273-39143273 on Build GRCh38
rs781238568 Chr.3: 39143397-39143397 on Build GRCh38
rs990113401 Chr.3: 39143161-39143161 on Build GRCh38
rs1001697783 Chr.3: 39143109-39143109 on Build GRCh38
rs752439836 Chr.3: 39143378-39143378 on Build GRCh38
rs112560069 Chr.3: 39143190-39143190 on Build GRCh38
rs202204996 Chr.3: 39143080-39143080 on Build GRCh38
rs199566978 Chr.3: 39143353-39143353 on Build GRCh38
rs180980340 Chr.3: 39143145-39143145 on Build GRCh38
rs1013924222 Chr.3: 39143401-39143402 on Build GRCh38
rs950626501 Chr.3: 39142941-39142941 on Build GRCh38
rs984645145 Chr.3: 39143148-39143148 on Build GRCh38
rs145148130 Chr.3: 39143359-39143359 on Build GRCh38
rs764658535 Chr.3: 39143078-39143078 on Build GRCh38
rs759794460 Chr.3: 39143046-39143046 on Build GRCh38
rs558531976 Chr.3: 39143310-39143310 on Build GRCh38
rs201069132 Chr.3: 39143147-39143147 on Build GRCh38
rs751206841 Chr.3: 39143084-39143084 on Build GRCh38
rs755470248 Chr.3: 39143253-39143255 on Build GRCh38
rs755131354 Chr.3: 39143395-39143395 on Build GRCh38
rs984679253 Chr.3: 39142949-39142949 on Build GRCh38
rs139132297 Chr.3: 39143139-39143139 on Build GRCh38
rs747186948 Chr.3: 39143317-39143317 on Build GRCh38
rs144549039 Chr.3: 39143403-39143403 on Build GRCh38
rs767034761 Chr.3: 39143321-39143323 on Build GRCh38
rs756024543 Chr.3: 39143242-39143242 on Build GRCh38
rs375668820 Chr.3: 39143136-39143136 on Build GRCh38

View Full Product Details