Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39263389-39263905 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CR1
Entrez Gene ID
1524
Gene Name
C-X3-C motif chemokine receptor 1
Gene Aliases
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
UniGene
Hs.78913
Gene Chromosome Location
Chr.3: 39263494-39281735 on Build GRCh38
Transcripts
NM_001171171.1 NM_001171172.1 NM_001171174.1 NM_001337.3

SNP Detail

SNP ID Chromosome Location
rs866717917 Chr.3: 39263615-39263615 on Build GRCh38
rs1049356066 Chr.3: 39263636-39263636 on Build GRCh38
rs888682836 Chr.3: 39263537-39263537 on Build GRCh38
rs973516486 Chr.3: 39263857-39263857 on Build GRCh38
rs967928325 Chr.3: 39263791-39263791 on Build GRCh38
rs895209391 Chr.3: 39263796-39263796 on Build GRCh38
rs1000668013 Chr.3: 39263764-39263764 on Build GRCh38
rs919635105 Chr.3: 39263598-39263598 on Build GRCh38
rs780735224 Chr.3: 39263437-39263437 on Build GRCh38
rs547265938 Chr.3: 39263642-39263642 on Build GRCh38
rs1015554986 Chr.3: 39263845-39263845 on Build GRCh38
rs769191499 Chr.3: 39263552-39263552 on Build GRCh38
rs917944831 Chr.3: 39263416-39263416 on Build GRCh38
rs778674470 Chr.3: 39263612-39263612 on Build GRCh38
rs1012353021 Chr.3: 39263802-39263802 on Build GRCh38
rs749620726 Chr.3: 39263449-39263449 on Build GRCh38
rs537065702 Chr.3: 39263850-39263850 on Build GRCh38
rs902336777 Chr.3: 39263750-39263750 on Build GRCh38
rs756510818 Chr.3: 39263427-39263427 on Build GRCh38
rs890814487 Chr.3: 39263649-39263649 on Build GRCh38
rs17038663 Chr.3: 39263813-39263813 on Build GRCh38
rs942225239 Chr.3: 39263484-39263484 on Build GRCh38
rs532696294 Chr.3: 39263768-39263768 on Build GRCh38
rs959227865 Chr.3: 39263703-39263703 on Build GRCh38
rs41481047 Chr.3: 39263825-39263825 on Build GRCh38
rs185018390 Chr.3: 39263526-39263526 on Build GRCh38
rs983719982 Chr.3: 39263839-39263839 on Build GRCh38
rs1032179920 Chr.3: 39263769-39263769 on Build GRCh38
rs757211660 Chr.3: 39263748-39263748 on Build GRCh38
rs973549086 Chr.3: 39263549-39263549 on Build GRCh38
rs747818724 Chr.3: 39263700-39263700 on Build GRCh38
rs1003591610 Chr.3: 39263667-39263667 on Build GRCh38
rs978168962 Chr.3: 39263798-39263798 on Build GRCh38
rs56022635 Chr.3: 39263830-39263831 on Build GRCh38
rs921672014 Chr.3: 39263866-39263866 on Build GRCh38
rs901379555 Chr.3: 39263466-39263466 on Build GRCh38
rs996959832 Chr.3: 39263477-39263477 on Build GRCh38
rs189825753 Chr.3: 39263856-39263856 on Build GRCh38
rs560571059 Chr.3: 39263659-39263659 on Build GRCh38
rs1049997730 Chr.3: 39263512-39263512 on Build GRCh38
rs922114700 Chr.3: 39263865-39263865 on Build GRCh38
rs772536046 Chr.3: 39263855-39263855 on Build GRCh38

View Full Product Details