Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 39264520-39265020 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CX3CR1
Entrez Gene ID
1524
Gene Name
C-X3-C motif chemokine receptor 1
Gene Aliases
CCRL1, CMKBRL1, CMKDR1, GPR13, GPRV28, V28
UniGene
Hs.78913
Gene Chromosome Location
Chr.3: 39263494-39281735 on Build GRCh38
Transcripts
NM_001171171.1 NM_001171172.1 NM_001171174.1 NM_001337.3

SNP Detail

SNP ID Chromosome Location
rs116614287 Chr.3: 39264661-39264661 on Build GRCh38
rs997296978 Chr.3: 39264984-39264985 on Build GRCh38
rs191423773 Chr.3: 39264954-39264954 on Build GRCh38
rs1050358123 Chr.3: 39264758-39264758 on Build GRCh38
rs142231266 Chr.3: 39264656-39264656 on Build GRCh38
rs1050389095 Chr.3: 39264580-39264580 on Build GRCh38
rs889344636 Chr.3: 39264601-39264601 on Build GRCh38
rs563117219 Chr.3: 39264826-39264826 on Build GRCh38
rs918710646 Chr.3: 39264870-39264870 on Build GRCh38
rs56369061 Chr.3: 39264611-39264612 on Build GRCh38
rs889145051 Chr.3: 39264783-39264783 on Build GRCh38
rs546600393 Chr.3: 39264965-39264965 on Build GRCh38
rs546593642 Chr.3: 39264551-39264551 on Build GRCh38
rs554596689 Chr.3: 39264970-39264970 on Build GRCh38
rs916819355 Chr.3: 39264910-39264910 on Build GRCh38
rs967597240 Chr.3: 39264639-39264639 on Build GRCh38
rs566068551 Chr.3: 39264610-39264625 on Build GRCh38
rs71534229 Chr.3: 39264613-39264613 on Build GRCh38
rs746457800 Chr.3: 39264708-39264708 on Build GRCh38
rs1030792404 Chr.3: 39264644-39264644 on Build GRCh38
rs969695000 Chr.3: 39264704-39264704 on Build GRCh38
rs777731814 Chr.3: 39264904-39264912 on Build GRCh38
rs1015992665 Chr.3: 39264856-39264856 on Build GRCh38
rs1010688357 Chr.3: 39264942-39264942 on Build GRCh38
rs953083965 Chr.3: 39264804-39264804 on Build GRCh38
rs1034910260 Chr.3: 39264974-39264974 on Build GRCh38
rs960902207 Chr.3: 39264884-39264884 on Build GRCh38
rs1028583680 Chr.3: 39264567-39264567 on Build GRCh38
rs544331650 Chr.3: 39264859-39264859 on Build GRCh38
rs529678827 Chr.3: 39264963-39264963 on Build GRCh38
rs868670307 Chr.3: 39264612-39264612 on Build GRCh38
rs538532239 Chr.3: 39264606-39264606 on Build GRCh38
rs984409313 Chr.3: 39264850-39264850 on Build GRCh38
rs1033194964 Chr.3: 39264876-39264876 on Build GRCh38
rs41391746 Chr.3: 39264756-39264756 on Build GRCh38
rs927783949 Chr.3: 39264721-39264721 on Build GRCh38
rs568700404 Chr.3: 39264616-39264616 on Build GRCh38
rs920641133 Chr.3: 39264753-39264753 on Build GRCh38
rs903344562 Chr.3: 39264860-39264860 on Build GRCh38
rs777912242 Chr.3: 39264575-39264575 on Build GRCh38
rs145988232 Chr.3: 39264833-39264833 on Build GRCh38
rs11710546 Chr.3: 39264728-39264728 on Build GRCh38
rs973294842 Chr.3: 39264621-39264621 on Build GRCh38
rs147817479 Chr.3: 39264990-39264990 on Build GRCh38
rs972315560 Chr.3: 39264750-39264750 on Build GRCh38
rs999001098 Chr.3: 39264873-39264873 on Build GRCh38
rs999386505 Chr.3: 39264714-39264714 on Build GRCh38
rs566325123 Chr.3: 39264966-39264966 on Build GRCh38
rs918857459 Chr.3: 39264638-39264638 on Build GRCh38
rs536266802 Chr.3: 39264940-39264940 on Build GRCh38
rs879630059 Chr.3: 39264932-39264932 on Build GRCh38
rs55783502 Chr.3: 39264797-39264797 on Build GRCh38
rs901715487 Chr.3: 39264977-39264977 on Build GRCh38
rs76874165 Chr.3: 39264643-39264643 on Build GRCh38
rs984545243 Chr.3: 39264588-39264588 on Build GRCh38
rs550681562 Chr.3: 39264683-39264683 on Build GRCh38
rs948057921 Chr.3: 39264939-39264939 on Build GRCh38
rs540588581 Chr.3: 39264705-39264705 on Build GRCh38
rs775091952 Chr.3: 39264886-39264886 on Build GRCh38
rs376778365 Chr.3: 39264908-39264908 on Build GRCh38
rs566731450 Chr.3: 39264602-39264602 on Build GRCh38

View Full Product Details