Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40310753-40311250 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1B
Entrez Gene ID
10289
Gene Name
eukaryotic translation initiation factor 1B
Gene Aliases
GC20
UniGene
Hs.315230
Gene Chromosome Location
Chr.3: 40309682-40312424 on Build GRCh38
Transcripts
NM_005875.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4680525 c.165_166insA p.K58fs*13 Chr.3: 40311026-40311027 on Build GRCh38
6097718 c.177T>A p.L59L Chr.3: 40311038-40311038 on Build GRCh38
5994780 c.176T>C p.L59P Chr.3: 40311037-40311037 on Build GRCh38
302121 c.73G>A p.A25T Chr.3: 40310934-40310934 on Build GRCh38
3775010 c.180G>A p.V60V Chr.3: 40311041-40311041 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755279770 Chr.3: 40310851-40310851 on Build GRCh38
rs748733173 Chr.3: 40310872-40310872 on Build GRCh38
rs765804227 Chr.3: 40311114-40311114 on Build GRCh38
rs929153687 Chr.3: 40311187-40311187 on Build GRCh38
rs780808042 Chr.3: 40311090-40311090 on Build GRCh38
rs750376606 Chr.3: 40310786-40310786 on Build GRCh38
rs35523102 Chr.3: 40310865-40310865 on Build GRCh38
rs773882061 Chr.3: 40310876-40310876 on Build GRCh38
rs535009130 Chr.3: 40310865-40310867 on Build GRCh38
rs750924490 Chr.3: 40310864-40310865 on Build GRCh38
rs112803565 Chr.3: 40310891-40310891 on Build GRCh38
rs770476330 Chr.3: 40310873-40310873 on Build GRCh38
rs549592226 Chr.3: 40310791-40310791 on Build GRCh38
rs1039159474 Chr.3: 40311222-40311222 on Build GRCh38
rs563941264 Chr.3: 40310836-40310836 on Build GRCh38
rs778562519 Chr.3: 40310927-40310927 on Build GRCh38
rs117372497 Chr.3: 40310838-40310838 on Build GRCh38
rs760588561 Chr.3: 40310959-40310959 on Build GRCh38
rs747879100 Chr.3: 40311089-40311089 on Build GRCh38
rs567560393 Chr.3: 40310866-40310866 on Build GRCh38
rs919147855 Chr.3: 40311158-40311158 on Build GRCh38
rs754691160 Chr.3: 40310874-40310875 on Build GRCh38
rs553290310 Chr.3: 40310866-40310867 on Build GRCh38
rs771727032 Chr.3: 40310897-40310897 on Build GRCh38
rs751708497 Chr.3: 40311062-40311062 on Build GRCh38
rs1027569798 Chr.3: 40310798-40310801 on Build GRCh38
rs927464483 Chr.3: 40310895-40310895 on Build GRCh38
rs371724665 Chr.3: 40311014-40311014 on Build GRCh38
rs752045013 Chr.3: 40310840-40310840 on Build GRCh38
rs984603351 Chr.3: 40311204-40311204 on Build GRCh38
rs1454491 Chr.3: 40311156-40311156 on Build GRCh38
rs778410314 Chr.3: 40310869-40310869 on Build GRCh38
rs776682893 Chr.3: 40310972-40310972 on Build GRCh38
rs79109389 Chr.3: 40311154-40311154 on Build GRCh38
rs566037132 Chr.3: 40310880-40310880 on Build GRCh38
rs908994283 Chr.3: 40311209-40311209 on Build GRCh38
rs1007660351 Chr.3: 40310800-40310800 on Build GRCh38
rs775081959 Chr.3: 40310933-40310933 on Build GRCh38
rs1454490 Chr.3: 40311149-40311149 on Build GRCh38
rs779667944 Chr.3: 40311188-40311188 on Build GRCh38
rs865939294 Chr.3: 40310922-40310922 on Build GRCh38
rs199629126 Chr.3: 40310861-40310861 on Build GRCh38
rs748556500 Chr.3: 40310858-40310858 on Build GRCh38
rs577972908 Chr.3: 40310867-40310867 on Build GRCh38
rs759772022 Chr.3: 40311067-40311067 on Build GRCh38
rs1038693883 Chr.3: 40310826-40310826 on Build GRCh38
rs781646925 Chr.3: 40310852-40310852 on Build GRCh38
rs767378110 Chr.3: 40310782-40310782 on Build GRCh38
rs762286126 Chr.3: 40310865-40310866 on Build GRCh38
rs1038817680 Chr.3: 40311185-40311185 on Build GRCh38
rs763097539 Chr.3: 40310990-40310990 on Build GRCh38
rs887520388 Chr.3: 40310794-40310794 on Build GRCh38
rs551375221 Chr.3: 40311078-40311078 on Build GRCh38
rs751040257 Chr.3: 40310865-40310868 on Build GRCh38
rs756102941 Chr.3: 40310837-40310837 on Build GRCh38
rs114327418 Chr.3: 40310966-40310966 on Build GRCh38
rs778649528 Chr.3: 40311210-40311210 on Build GRCh38
rs989237947 Chr.3: 40311111-40311113 on Build GRCh38

View Full Product Details