Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40312241-40312752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF1B
Entrez Gene ID
10289
Gene Name
eukaryotic translation initiation factor 1B
Gene Aliases
GC20
UniGene
Hs.315230
Gene Chromosome Location
Chr.3: 40309682-40312424 on Build GRCh38
Transcripts
NM_005875.2

SNP Detail

SNP ID Chromosome Location
rs1018692527 Chr.3: 40312341-40312341 on Build GRCh38
rs868256014 Chr.3: 40312272-40312272 on Build GRCh38
rs1040106583 Chr.3: 40312711-40312711 on Build GRCh38
rs3836541 Chr.3: 40312345-40312346 on Build GRCh38
rs978817833 Chr.3: 40312444-40312444 on Build GRCh38
rs968444566 Chr.3: 40312361-40312361 on Build GRCh38
rs76420744 Chr.3: 40312308-40312308 on Build GRCh38
rs965790938 Chr.3: 40312388-40312388 on Build GRCh38
rs36080461 Chr.3: 40312428-40312429 on Build GRCh38
rs974366890 Chr.3: 40312433-40312433 on Build GRCh38
rs1041962799 Chr.3: 40312648-40312648 on Build GRCh38
rs534267387 Chr.3: 40312668-40312668 on Build GRCh38
rs3836542 Chr.3: 40312492-40312513 on Build GRCh38
rs747395671 Chr.3: 40312358-40312358 on Build GRCh38
rs928692413 Chr.3: 40312488-40312488 on Build GRCh38
rs752409436 Chr.3: 40312699-40312699 on Build GRCh38
rs11374095 Chr.3: 40312300-40312301 on Build GRCh38
rs893147167 Chr.3: 40312613-40312613 on Build GRCh38
rs1057365614 Chr.3: 40312484-40312484 on Build GRCh38
rs183153885 Chr.3: 40312514-40312514 on Build GRCh38
rs912957161 Chr.3: 40312356-40312356 on Build GRCh38
rs397698852 Chr.3: 40312307-40312308 on Build GRCh38
rs145200238 Chr.3: 40312465-40312465 on Build GRCh38
rs988539034 Chr.3: 40312355-40312355 on Build GRCh38
rs147610987 Chr.3: 40312643-40312643 on Build GRCh38
rs368026725 Chr.3: 40312649-40312651 on Build GRCh38
rs760574211 Chr.3: 40312487-40312487 on Build GRCh38
rs1020830596 Chr.3: 40312269-40312269 on Build GRCh38
rs903470150 Chr.3: 40312691-40312691 on Build GRCh38
rs954506761 Chr.3: 40312330-40312330 on Build GRCh38
rs937580287 Chr.3: 40312459-40312459 on Build GRCh38
rs199932639 Chr.3: 40312300-40312300 on Build GRCh38
rs951939946 Chr.3: 40312447-40312447 on Build GRCh38
rs917422648 Chr.3: 40312507-40312507 on Build GRCh38
rs182623965 Chr.3: 40312458-40312458 on Build GRCh38
rs561869511 Chr.3: 40312647-40312649 on Build GRCh38
rs984732968 Chr.3: 40312456-40312456 on Build GRCh38
rs1008376235 Chr.3: 40312697-40312701 on Build GRCh38
rs764727103 Chr.3: 40312554-40312554 on Build GRCh38
rs878988278 Chr.3: 40312284-40312284 on Build GRCh38
rs6804545 Chr.3: 40312549-40312549 on Build GRCh38
rs556877048 Chr.3: 40312360-40312360 on Build GRCh38
rs996060112 Chr.3: 40312321-40312321 on Build GRCh38
rs1030129670 Chr.3: 40312329-40312329 on Build GRCh38
rs559549361 Chr.3: 40312483-40312483 on Build GRCh38

View Full Product Details