Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 40414591-40415099 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ENTPD3
Entrez Gene ID
956
Gene Name
ectonucleoside triphosphate diphosphohydrolase 3
Gene Aliases
CD39L3, HB6, NTPDase-3
UniGene
Hs.441145
Gene Chromosome Location
Chr.3: 40387156-40428626 on Build GRCh38
Transcripts
NM_001248.3 NM_001291960.1 NM_001291961.1 XM_011534266.2 NR_040100.1

Gene Symbol
ENTPD3-AS1
Entrez Gene ID
285266
Gene Name
ENTPD3 antisense RNA 1
Gene Aliases
-
UniGene
Hs.619663
Gene Chromosome Location
Chr.3: 40390951-40453308 on Build GRCh38
Transcripts
NM_001248.3 NM_001291960.1 NM_001291961.1 XM_011534266.2 NR_040100.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4117519 c.597+2T>C p.? Chr.3: 40414842-40414842 on Build GRCh38
3593939 c.557G>A p.G186E Chr.3: 40414800-40414800 on Build GRCh38
5797736 c.463G>A p.E155K Chr.3: 40414706-40414706 on Build GRCh38
4840279 c.590T>G p.F197C Chr.3: 40414833-40414833 on Build GRCh38
1692628 c.595G>A p.E199K Chr.3: 40414838-40414838 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs956733236 Chr.3: 40414987-40414987 on Build GRCh38
rs751126145 Chr.3: 40414817-40414820 on Build GRCh38
rs775624176 Chr.3: 40414822-40414822 on Build GRCh38
rs922649250 Chr.3: 40414701-40414701 on Build GRCh38
rs767639264 Chr.3: 40414684-40414684 on Build GRCh38
rs760917433 Chr.3: 40414823-40414823 on Build GRCh38
rs199532859 Chr.3: 40414819-40414819 on Build GRCh38
rs759746823 Chr.3: 40414663-40414663 on Build GRCh38
rs749902353 Chr.3: 40414744-40414744 on Build GRCh38
rs758062140 Chr.3: 40414746-40414746 on Build GRCh38
rs779627045 Chr.3: 40414756-40414756 on Build GRCh38
rs771097069 Chr.3: 40414643-40414643 on Build GRCh38
rs762416626 Chr.3: 40414856-40414856 on Build GRCh38
rs754704592 Chr.3: 40414886-40414886 on Build GRCh38
rs754302619 Chr.3: 40414720-40414720 on Build GRCh38
rs995288243 Chr.3: 40414888-40414888 on Build GRCh38
rs912966385 Chr.3: 40414635-40414635 on Build GRCh38
rs772325075 Chr.3: 40414820-40414820 on Build GRCh38
rs764552323 Chr.3: 40414713-40414713 on Build GRCh38
rs781080367 Chr.3: 40414887-40414887 on Build GRCh38
rs75633564 Chr.3: 40414629-40414629 on Build GRCh38
rs545374968 Chr.3: 40414892-40414892 on Build GRCh38
rs531053650 Chr.3: 40414925-40414925 on Build GRCh38
rs761086925 Chr.3: 40414706-40414706 on Build GRCh38
rs915349129 Chr.3: 40415004-40415004 on Build GRCh38
rs145439935 Chr.3: 40414918-40414918 on Build GRCh38
rs199612621 Chr.3: 40414761-40414761 on Build GRCh38
rs147275720 Chr.3: 40414930-40414930 on Build GRCh38
rs945755853 Chr.3: 40414648-40414648 on Build GRCh38
rs749351976 Chr.3: 40414632-40414632 on Build GRCh38
rs776992266 Chr.3: 40414835-40414835 on Build GRCh38
rs977166440 Chr.3: 40414659-40414659 on Build GRCh38
rs765726571 Chr.3: 40414847-40414847 on Build GRCh38
rs993234652 Chr.3: 40414873-40414873 on Build GRCh38
rs897551461 Chr.3: 40414850-40414850 on Build GRCh38
rs750054107 Chr.3: 40414913-40414913 on Build GRCh38
rs530080028 Chr.3: 40414974-40414974 on Build GRCh38
rs372974975 Chr.3: 40414735-40414735 on Build GRCh38
rs763411270 Chr.3: 40414731-40414731 on Build GRCh38
rs1027430954 Chr.3: 40414890-40414890 on Build GRCh38
rs749933824 Chr.3: 40414852-40414852 on Build GRCh38
rs968234491 Chr.3: 40415051-40415051 on Build GRCh38
rs1007840216 Chr.3: 40414922-40414922 on Build GRCh38
rs775919662 Chr.3: 40414687-40414687 on Build GRCh38
rs774320301 Chr.3: 40414656-40414656 on Build GRCh38
rs766319813 Chr.3: 40414933-40414933 on Build GRCh38
rs573202419 Chr.3: 40414883-40414883 on Build GRCh38
rs748052846 Chr.3: 40414802-40414802 on Build GRCh38
rs376278763 Chr.3: 40414843-40414843 on Build GRCh38
rs545154743 Chr.3: 40414904-40414904 on Build GRCh38
rs1014047821 Chr.3: 40414939-40414939 on Build GRCh38
rs754864092 Chr.3: 40414787-40414787 on Build GRCh38
rs150819230 Chr.3: 40414622-40414622 on Build GRCh38
rs371849573 Chr.3: 40414804-40414804 on Build GRCh38
rs561243228 Chr.3: 40414963-40414963 on Build GRCh38
rs766864928 Chr.3: 40414782-40414784 on Build GRCh38
rs781168719 Chr.3: 40414800-40414800 on Build GRCh38
rs1039595963 Chr.3: 40414885-40414885 on Build GRCh38
rs559559462 Chr.3: 40415000-40415000 on Build GRCh38
rs751219677 Chr.3: 40414884-40414884 on Build GRCh38
rs774423742 Chr.3: 40414815-40414815 on Build GRCh38
rs764392874 Chr.3: 40414824-40414824 on Build GRCh38
rs545683407 Chr.3: 40414995-40414995 on Build GRCh38
rs139278593 Chr.3: 40414816-40414816 on Build GRCh38
rs935436464 Chr.3: 40414731-40414731 on Build GRCh38
rs755743394 Chr.3: 40414962-40414962 on Build GRCh38
rs144846398 Chr.3: 40414980-40414980 on Build GRCh38
rs775252921 Chr.3: 40414797-40414797 on Build GRCh38
rs764744475 Chr.3: 40414741-40414741 on Build GRCh38
rs771656116 Chr.3: 40414719-40414719 on Build GRCh38
rs573560311 Chr.3: 40414631-40414631 on Build GRCh38
rs746775333 Chr.3: 40414779-40414779 on Build GRCh38
rs980807420 Chr.3: 40414968-40414968 on Build GRCh38
rs79713000 Chr.3: 40414832-40414832 on Build GRCh38
rs914031728 Chr.3: 40415062-40415062 on Build GRCh38
rs938993770 Chr.3: 40414839-40414839 on Build GRCh38
rs1056162441 Chr.3: 40414840-40414840 on Build GRCh38

View Full Product Details