Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 42874330-42874818 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP8B1
Entrez Gene ID
1582
Gene Name
cytochrome P450 family 8 subfamily B member 1
Gene Aliases
CP8B, CYP12
UniGene
Hs.447793
Gene Chromosome Location
Chr.3: 42872192-42876141 on Build GRCh38
Transcripts
NM_004391.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1423151 c.1202C>T p.T401I Chr.3: 42874615-42874615 on Build GRCh38
4117665 c.1274A>C p.K425T Chr.3: 42874543-42874543 on Build GRCh38
1202907 c.1487G>A p.R496H Chr.3: 42874330-42874330 on Build GRCh38
3066341 c.1308C>T p.G436G Chr.3: 42874509-42874509 on Build GRCh38
3916062 c.1278C>T p.I426I Chr.3: 42874539-42874539 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs759570260 Chr.3: 42874790-42874790 on Build GRCh38
rs756447307 Chr.3: 42874724-42874724 on Build GRCh38
rs772337032 Chr.3: 42874452-42874452 on Build GRCh38
rs746803518 Chr.3: 42874771-42874771 on Build GRCh38
rs771857190 Chr.3: 42874765-42874765 on Build GRCh38
rs770517385 Chr.3: 42874568-42874568 on Build GRCh38
rs61747186 Chr.3: 42874746-42874746 on Build GRCh38
rs761246497 Chr.3: 42874406-42874406 on Build GRCh38
rs776582312 Chr.3: 42874677-42874677 on Build GRCh38
rs761638023 Chr.3: 42874683-42874683 on Build GRCh38
rs973036069 Chr.3: 42874441-42874441 on Build GRCh38
rs770523960 Chr.3: 42874648-42874648 on Build GRCh38
rs748484847 Chr.3: 42874625-42874625 on Build GRCh38
rs757363149 Chr.3: 42874448-42874448 on Build GRCh38
rs528095572 Chr.3: 42874772-42874772 on Build GRCh38
rs868826615 Chr.3: 42874542-42874542 on Build GRCh38
rs771834978 Chr.3: 42874656-42874656 on Build GRCh38
rs200069231 Chr.3: 42874654-42874654 on Build GRCh38
rs148221181 Chr.3: 42874523-42874523 on Build GRCh38
rs151059871 Chr.3: 42874443-42874443 on Build GRCh38
rs760453758 Chr.3: 42874672-42874672 on Build GRCh38
rs368940524 Chr.3: 42874597-42874597 on Build GRCh38
rs761111073 Chr.3: 42874372-42874372 on Build GRCh38
rs1050194322 Chr.3: 42874474-42874474 on Build GRCh38
rs953058812 Chr.3: 42874687-42874687 on Build GRCh38
rs150329640 Chr.3: 42874791-42874791 on Build GRCh38
rs764572567 Chr.3: 42874397-42874397 on Build GRCh38
rs763740338 Chr.3: 42874418-42874418 on Build GRCh38
rs770317982 Chr.3: 42874543-42874543 on Build GRCh38
rs370752214 Chr.3: 42874356-42874356 on Build GRCh38
rs751401316 Chr.3: 42874491-42874491 on Build GRCh38
rs35637877 Chr.3: 42874748-42874748 on Build GRCh38
rs961903195 Chr.3: 42874618-42874618 on Build GRCh38
rs777977904 Chr.3: 42874736-42874736 on Build GRCh38
rs776931947 Chr.3: 42874387-42874387 on Build GRCh38
rs370477226 Chr.3: 42874490-42874490 on Build GRCh38
rs370565459 Chr.3: 42874451-42874451 on Build GRCh38
rs140045753 Chr.3: 42874508-42874508 on Build GRCh38
rs753449623 Chr.3: 42874569-42874569 on Build GRCh38
rs746971123 Chr.3: 42874363-42874363 on Build GRCh38
rs761338569 Chr.3: 42874482-42874482 on Build GRCh38
rs752533981 Chr.3: 42874574-42874574 on Build GRCh38
rs754959169 Chr.3: 42874364-42874364 on Build GRCh38
rs766489047 Chr.3: 42874688-42874688 on Build GRCh38
rs748697680 Chr.3: 42874538-42874538 on Build GRCh38
rs779027550 Chr.3: 42874449-42874449 on Build GRCh38
rs899077279 Chr.3: 42874605-42874605 on Build GRCh38
rs774416736 Chr.3: 42874788-42874788 on Build GRCh38
rs578084493 Chr.3: 42874623-42874623 on Build GRCh38
rs1037585674 Chr.3: 42874601-42874601 on Build GRCh38
rs749410960 Chr.3: 42874447-42874447 on Build GRCh38
rs144987882 Chr.3: 42874464-42874464 on Build GRCh38
rs735320 Chr.3: 42874386-42874386 on Build GRCh38
rs749998299 Chr.3: 42874415-42874415 on Build GRCh38
rs759681635 Chr.3: 42874438-42874438 on Build GRCh38
rs374042052 Chr.3: 42874459-42874459 on Build GRCh38
rs201949466 Chr.3: 42874350-42874350 on Build GRCh38
rs760589676 Chr.3: 42874581-42874581 on Build GRCh38
rs773966466 Chr.3: 42874653-42874653 on Build GRCh38
rs1024381303 Chr.3: 42874524-42874524 on Build GRCh38
rs373209479 Chr.3: 42874393-42874393 on Build GRCh38
rs755254367 Chr.3: 42874698-42874698 on Build GRCh38
rs767458378 Chr.3: 42874573-42874573 on Build GRCh38
rs536175564 Chr.3: 42874513-42874513 on Build GRCh38
rs1054418938 Chr.3: 42874510-42874510 on Build GRCh38
rs771278880 Chr.3: 42874382-42874382 on Build GRCh38
rs768840000 Chr.3: 42874677-42874678 on Build GRCh38
rs774096821 Chr.3: 42874570-42874570 on Build GRCh38
rs555313540 Chr.3: 42874529-42874529 on Build GRCh38
rs112066175 Chr.3: 42874613-42874613 on Build GRCh38
rs138292874 Chr.3: 42874509-42874509 on Build GRCh38
rs765149827 Chr.3: 42874777-42874777 on Build GRCh38
rs775891073 Chr.3: 42874395-42874395 on Build GRCh38
rs1041946948 Chr.3: 42874495-42874495 on Build GRCh38
rs763970646 Chr.3: 42874674-42874674 on Build GRCh38
rs372472190 Chr.3: 42874354-42874354 on Build GRCh38
rs534733049 Chr.3: 42874706-42874706 on Build GRCh38
rs758049663 Chr.3: 42874416-42874416 on Build GRCh38
rs267599827 Chr.3: 42874349-42874349 on Build GRCh38
rs770204568 Chr.3: 42874626-42874626 on Build GRCh38
rs200974379 Chr.3: 42874479-42874479 on Build GRCh38
rs762568062 Chr.3: 42874488-42874488 on Build GRCh38
rs778621588 Chr.3: 42874757-42874757 on Build GRCh38
rs149079664 Chr.3: 42874408-42874408 on Build GRCh38
rs142764338 Chr.3: 42874776-42874776 on Build GRCh38
rs745317569 Chr.3: 42874764-42874764 on Build GRCh38
rs768964905 Chr.3: 42874526-42874526 on Build GRCh38
rs375960422 Chr.3: 42874444-42874444 on Build GRCh38
rs760124342 Chr.3: 42874357-42874364 on Build GRCh38
rs777266662 Chr.3: 42874487-42874487 on Build GRCh38
rs778299923 Chr.3: 42874630-42874630 on Build GRCh38
rs764706332 Chr.3: 42874486-42874486 on Build GRCh38
rs775777382 Chr.3: 42874455-42874455 on Build GRCh38
rs138532585 Chr.3: 42874358-42874358 on Build GRCh38
rs147356860 Chr.3: 42874572-42874572 on Build GRCh38
rs139423402 Chr.3: 42874593-42874593 on Build GRCh38
rs374468470 Chr.3: 42874512-42874512 on Build GRCh38
rs776132074 Chr.3: 42874400-42874400 on Build GRCh38
rs141302264 Chr.3: 42874624-42874624 on Build GRCh38
rs764035169 Chr.3: 42874587-42874587 on Build GRCh38
rs757067156 Chr.3: 42874620-42874621 on Build GRCh38
rs772607624 Chr.3: 42874550-42874550 on Build GRCh38
rs762454550 Chr.3: 42874562-42874562 on Build GRCh38
rs766151616 Chr.3: 42874427-42874427 on Build GRCh38
rs528157119 Chr.3: 42874428-42874428 on Build GRCh38
rs754776097 Chr.3: 42874503-42874503 on Build GRCh38
rs543245438 Chr.3: 42874643-42874643 on Build GRCh38
rs751637637 Chr.3: 42874695-42874695 on Build GRCh38
rs749318881 Chr.3: 42874378-42874378 on Build GRCh38
rs146795927 Chr.3: 42874660-42874660 on Build GRCh38
rs150584658 Chr.3: 42874598-42874598 on Build GRCh38
rs920224607 Chr.3: 42874675-42874675 on Build GRCh38
rs755164557 Chr.3: 42874614-42874614 on Build GRCh38
rs764977017 Chr.3: 42874414-42874414 on Build GRCh38
rs144198819 Chr.3: 42874353-42874353 on Build GRCh38

View Full Product Details